Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 56275843-56276348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAO1
Entrez Gene ID
2775
Gene Name
G protein subunit alpha o1
Gene Aliases
EIEE17, G-ALPHA-o, GNAO, HLA-DQB1
UniGene
Hs.644524
Gene Chromosome Location
Chr.16: 56191339-56357444 on Build GRCh38
Transcripts
NM_020988.2 NM_138736.2 XM_011523003.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1742384 c.280G>A p.E94K Chr.16: 56276049-56276049 on Build GRCh38
4428796 c.256C>T p.R86W Chr.16: 56276025-56276025 on Build GRCh38
6430809 c.253G>A p.V85I Chr.16: 56276022-56276022 on Build GRCh38
6002028 c.245C>A p.A82E Chr.16: 56276014-56276014 on Build GRCh38
7123140 c.198G>A p.V66V Chr.16: 56275967-56275967 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199865766 Chr.16: 56276261-56276262 on Build GRCh38
rs201728736 Chr.16: 56276036-56276036 on Build GRCh38
rs371330886 Chr.16: 56275983-56275983 on Build GRCh38
rs577003204 Chr.16: 56276277-56276277 on Build GRCh38
rs774491938 Chr.16: 56275892-56275892 on Build GRCh38
rs992304782 Chr.16: 56276313-56276313 on Build GRCh38
rs952157881 Chr.16: 56276242-56276242 on Build GRCh38
rs374220803 Chr.16: 56276220-56276220 on Build GRCh38
rs975185492 Chr.16: 56276180-56276180 on Build GRCh38
rs930724742 Chr.16: 56275909-56275909 on Build GRCh38
rs756630839 Chr.16: 56276166-56276166 on Build GRCh38
rs767115578 Chr.16: 56276025-56276025 on Build GRCh38
rs770917900 Chr.16: 56276102-56276102 on Build GRCh38
rs940833242 Chr.16: 56275956-56275956 on Build GRCh38
rs749909537 Chr.16: 56276048-56276048 on Build GRCh38
rs903629384 Chr.16: 56276022-56276022 on Build GRCh38
rs199632176 Chr.16: 56276057-56276057 on Build GRCh38
rs757370770 Chr.16: 56276108-56276108 on Build GRCh38
rs747048547 Chr.16: 56276099-56276099 on Build GRCh38
rs766855846 Chr.16: 56275920-56275920 on Build GRCh38
rs77558236 Chr.16: 56275976-56275976 on Build GRCh38
rs559674838 Chr.16: 56276055-56276055 on Build GRCh38
rs557932562 Chr.16: 56275965-56275965 on Build GRCh38
rs756947463 Chr.16: 56276021-56276021 on Build GRCh38
rs758141746 Chr.16: 56275870-56275870 on Build GRCh38
rs773324912 Chr.16: 56276086-56276086 on Build GRCh38
rs200201305 Chr.16: 56275955-56275955 on Build GRCh38
rs867727516 Chr.16: 56276296-56276296 on Build GRCh38
rs748368535 Chr.16: 56275894-56275894 on Build GRCh38
rs745880843 Chr.16: 56276213-56276213 on Build GRCh38
rs183824697 Chr.16: 56275910-56275910 on Build GRCh38
rs746915167 Chr.16: 56276299-56276299 on Build GRCh38
rs776835586 Chr.16: 56276117-56276117 on Build GRCh38
rs896696411 Chr.16: 56275979-56275979 on Build GRCh38
rs747990383 Chr.16: 56276079-56276079 on Build GRCh38
rs78190585 Chr.16: 56275883-56275883 on Build GRCh38
rs746831543 Chr.16: 56276298-56276298 on Build GRCh38
rs773323392 Chr.16: 56275905-56275905 on Build GRCh38
rs544584257 Chr.16: 56276314-56276314 on Build GRCh38
rs368169260 Chr.16: 56275890-56275890 on Build GRCh38
rs1012333756 Chr.16: 56275991-56275991 on Build GRCh38
rs368113345 Chr.16: 56275964-56275964 on Build GRCh38
rs778669032 Chr.16: 56276059-56276059 on Build GRCh38
rs760104939 Chr.16: 56276122-56276122 on Build GRCh38
rs778169910 Chr.16: 56275982-56275982 on Build GRCh38
rs963988866 Chr.16: 56276179-56276179 on Build GRCh38
rs771820878 Chr.16: 56276222-56276222 on Build GRCh38
rs772342375 Chr.16: 56275895-56275895 on Build GRCh38
rs907940126 Chr.16: 56276297-56276297 on Build GRCh38
rs778698228 Chr.16: 56276167-56276167 on Build GRCh38
rs189990922 Chr.16: 56276056-56276056 on Build GRCh38
rs772254603 Chr.16: 56276085-56276085 on Build GRCh38

View Full Product Details