Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 56336652-56337147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAO1
Entrez Gene ID
2775
Gene Name
G protein subunit alpha o1
Gene Aliases
EIEE17, G-ALPHA-o, GNAO, HLA-DQB1
UniGene
Hs.644524
Gene Chromosome Location
Chr.16: 56191339-56357444 on Build GRCh38
Transcripts
NM_020988.2 NM_138736.2 XM_011523003.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
971646 c.681G>A p.A227A Chr.16: 56336818-56336818 on Build GRCh38
971643 c.649G>A p.E217K Chr.16: 56336786-56336786 on Build GRCh38
6055412 c.658A>C p.T220P Chr.16: 56336795-56336795 on Build GRCh38
971640 c.648C>A p.F216L Chr.16: 56336785-56336785 on Build GRCh38
971636 c.606C>T p.V202V Chr.16: 56336743-56336743 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747946596 Chr.16: 56336716-56336716 on Build GRCh38
rs757819174 Chr.16: 56336876-56336876 on Build GRCh38
rs752775959 Chr.16: 56336851-56336851 on Build GRCh38
rs759007389 Chr.16: 56337105-56337105 on Build GRCh38
rs768258979 Chr.16: 56336749-56336749 on Build GRCh38
rs765429450 Chr.16: 56336845-56336845 on Build GRCh38
rs538663993 Chr.16: 56336893-56336893 on Build GRCh38
rs189380751 Chr.16: 56336797-56336797 on Build GRCh38
rs777790786 Chr.16: 56336903-56336903 on Build GRCh38
rs977905228 Chr.16: 56337107-56337107 on Build GRCh38
rs959628768 Chr.16: 56337087-56337087 on Build GRCh38
rs751882855 Chr.16: 56336710-56336710 on Build GRCh38
rs542653987 Chr.16: 56337100-56337100 on Build GRCh38
rs754090582 Chr.16: 56336683-56336683 on Build GRCh38
rs1007566323 Chr.16: 56336677-56336677 on Build GRCh38
rs546569747 Chr.16: 56336824-56336824 on Build GRCh38
rs12721461 Chr.16: 56336793-56336793 on Build GRCh38
rs760476691 Chr.16: 56336815-56336815 on Build GRCh38
rs369026621 Chr.16: 56336704-56336704 on Build GRCh38
rs747707167 Chr.16: 56336785-56336785 on Build GRCh38
rs377573101 Chr.16: 56336906-56336906 on Build GRCh38
rs1048268406 Chr.16: 56337001-56337001 on Build GRCh38
rs886039494 Chr.16: 56336762-56336762 on Build GRCh38
rs1003670301 Chr.16: 56337045-56337045 on Build GRCh38
rs933814130 Chr.16: 56336886-56336886 on Build GRCh38
rs797044878 Chr.16: 56336763-56336763 on Build GRCh38
rs12721464 Chr.16: 56336895-56336895 on Build GRCh38
rs770769352 Chr.16: 56336818-56336818 on Build GRCh38
rs560924276 Chr.16: 56336963-56336963 on Build GRCh38
rs770509149 Chr.16: 56336996-56336996 on Build GRCh38
rs759775512 Chr.16: 56336826-56336826 on Build GRCh38
rs1033776222 Chr.16: 56337053-56337053 on Build GRCh38
rs373395629 Chr.16: 56336705-56336705 on Build GRCh38
rs797045599 Chr.16: 56336817-56336817 on Build GRCh38
rs764028554 Chr.16: 56336875-56336875 on Build GRCh38
rs1057518440 Chr.16: 56336757-56336757 on Build GRCh38
rs150641952 Chr.16: 56336743-56336743 on Build GRCh38
rs778118270 Chr.16: 56337047-56337047 on Build GRCh38
rs371451695 Chr.16: 56336869-56336869 on Build GRCh38
rs770019378 Chr.16: 56336910-56336910 on Build GRCh38
rs781616251 Chr.16: 56336879-56336879 on Build GRCh38
rs567136805 Chr.16: 56336738-56336738 on Build GRCh38
rs370692563 Chr.16: 56336880-56336880 on Build GRCh38
rs370658719 Chr.16: 56336907-56336907 on Build GRCh38
rs763053362 Chr.16: 56336860-56336860 on Build GRCh38
rs2278657 Chr.16: 56337052-56337052 on Build GRCh38
rs377359901 Chr.16: 56336894-56336894 on Build GRCh38
rs769651863 Chr.16: 56336720-56336720 on Build GRCh38
rs548233212 Chr.16: 56336725-56336725 on Build GRCh38
rs773889434 Chr.16: 56337010-56337010 on Build GRCh38
rs572151875 Chr.16: 56336905-56336905 on Build GRCh38
rs1011091535 Chr.16: 56337088-56337088 on Build GRCh38
rs899142351 Chr.16: 56336944-56336944 on Build GRCh38
rs76948070 Chr.16: 56337016-56337016 on Build GRCh38
rs139322464 Chr.16: 56336791-56336791 on Build GRCh38
rs779514851 Chr.16: 56336727-56336727 on Build GRCh38
rs760140791 Chr.16: 56336872-56336872 on Build GRCh38
rs587777057 Chr.16: 56336744-56336744 on Build GRCh38
rs1057518678 Chr.16: 56336829-56336829 on Build GRCh38
rs386791061 Chr.16: 56337074-56337075 on Build GRCh38
rs745675528 Chr.16: 56336909-56336909 on Build GRCh38
rs778453425 Chr.16: 56336767-56336767 on Build GRCh38
rs752075327 Chr.16: 56336875-56336875 on Build GRCh38
rs759739007 Chr.16: 56336684-56336684 on Build GRCh38
rs112651404 Chr.16: 56336700-56336700 on Build GRCh38
rs373093494 Chr.16: 56336881-56336881 on Build GRCh38

View Full Product Details