Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67132367-67132866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf70
Entrez Gene ID
80262
Gene Name
chromosome 16 open reading frame 70
Gene Aliases
C16orf6, LIN10, lin-10
UniGene
Hs.513666
Gene Chromosome Location
Chr.16: 67109881-67148544 on Build GRCh38
Transcripts
NM_001320540.1 NM_001320541.1 NM_001320542.1 NM_001320543.1 NM_025187.4 XM_006721284.3 XM_011523350.2 XM_017023729.1 XM_017023730.1 XM_017023731.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6614561 c.283C>T p.H95Y Chr.16: 67132453-67132453 on Build GRCh38
305929 c.293C>A p.S98Y Chr.16: 67132463-67132463 on Build GRCh38
1246535 c.389A>G p.N130S Chr.16: 67132850-67132850 on Build GRCh38
7331381 c.279C>T p.G93G Chr.16: 67132449-67132449 on Build GRCh38
6451071 c.366C>T p.S122S Chr.16: 67132827-67132827 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs957920750 Chr.16: 67132653-67132653 on Build GRCh38
rs776122798 Chr.16: 67132463-67132463 on Build GRCh38
rs754827801 Chr.16: 67132539-67132539 on Build GRCh38
rs780508699 Chr.16: 67132670-67132670 on Build GRCh38
rs748903129 Chr.16: 67132808-67132808 on Build GRCh38
rs768305771 Chr.16: 67132408-67132408 on Build GRCh38
rs745537230 Chr.16: 67132672-67132672 on Build GRCh38
rs748086725 Chr.16: 67132557-67132557 on Build GRCh38
rs751420574 Chr.16: 67132837-67132837 on Build GRCh38
rs758915292 Chr.16: 67132483-67132483 on Build GRCh38
rs962685671 Chr.16: 67132712-67132712 on Build GRCh38
rs755146317 Chr.16: 67132422-67132422 on Build GRCh38
rs780323053 Chr.16: 67132644-67132644 on Build GRCh38
rs201486467 Chr.16: 67132602-67132602 on Build GRCh38
rs778518017 Chr.16: 67132550-67132550 on Build GRCh38
rs773440272 Chr.16: 67132564-67132564 on Build GRCh38
rs772517492 Chr.16: 67132823-67132823 on Build GRCh38
rs763404145 Chr.16: 67132600-67132600 on Build GRCh38
rs775187407 Chr.16: 67132680-67132680 on Build GRCh38
rs767448775 Chr.16: 67132628-67132628 on Build GRCh38
rs766438603 Chr.16: 67132827-67132827 on Build GRCh38
rs771588108 Chr.16: 67132443-67132443 on Build GRCh38
rs766554905 Chr.16: 67132732-67132732 on Build GRCh38
rs372791486 Chr.16: 67132826-67132826 on Build GRCh38
rs752473664 Chr.16: 67132719-67132719 on Build GRCh38
rs777723677 Chr.16: 67132659-67132659 on Build GRCh38
rs765474714 Chr.16: 67132475-67132475 on Build GRCh38
rs757074411 Chr.16: 67132804-67132804 on Build GRCh38
rs942754160 Chr.16: 67132839-67132839 on Build GRCh38
rs201455515 Chr.16: 67132810-67132810 on Build GRCh38
rs757602248 Chr.16: 67132708-67132719 on Build GRCh38
rs187185848 Chr.16: 67132697-67132697 on Build GRCh38
rs62624497 Chr.16: 67132636-67132636 on Build GRCh38
rs988997046 Chr.16: 67132707-67132707 on Build GRCh38
rs772068254 Chr.16: 67132561-67132561 on Build GRCh38
rs769399429 Chr.16: 67132675-67132675 on Build GRCh38
rs753268610 Chr.16: 67132480-67132480 on Build GRCh38
rs956379836 Chr.16: 67132703-67132703 on Build GRCh38
rs752549797 Chr.16: 67132654-67132654 on Build GRCh38
rs999296518 Chr.16: 67132655-67132655 on Build GRCh38
rs750539123 Chr.16: 67132720-67132720 on Build GRCh38
rs759060393 Chr.16: 67132591-67132591 on Build GRCh38
rs763824071 Chr.16: 67132778-67132778 on Build GRCh38
rs902153664 Chr.16: 67132614-67132614 on Build GRCh38
rs1039893636 Chr.16: 67132555-67132555 on Build GRCh38
rs144951759 Chr.16: 67132569-67132569 on Build GRCh38
rs752082096 Chr.16: 67132606-67132606 on Build GRCh38
rs754040825 Chr.16: 67132402-67132402 on Build GRCh38
rs569581781 Chr.16: 67132580-67132580 on Build GRCh38
rs760800116 Chr.16: 67132721-67132721 on Build GRCh38
rs34017457 Chr.16: 67132828-67132828 on Build GRCh38
rs769103806 Chr.16: 67132696-67132696 on Build GRCh38
rs758164378 Chr.16: 67132657-67132657 on Build GRCh38
rs374172550 Chr.16: 67132805-67132805 on Build GRCh38
rs759981968 Chr.16: 67132471-67132471 on Build GRCh38
rs1032143938 Chr.16: 67132615-67132615 on Build GRCh38
rs764748595 Chr.16: 67132603-67132603 on Build GRCh38
rs755620331 Chr.16: 67132806-67132806 on Build GRCh38
rs770394433 Chr.16: 67132571-67132571 on Build GRCh38
rs747990967 Chr.16: 67132428-67132428 on Build GRCh38
rs141238675 Chr.16: 67132449-67132449 on Build GRCh38
rs537008618 Chr.16: 67132817-67132817 on Build GRCh38
rs765479476 Chr.16: 67132663-67132663 on Build GRCh38
rs754024436 Chr.16: 67132742-67132742 on Build GRCh38
rs767642547 Chr.16: 67132699-67132699 on Build GRCh38
rs764272827 Chr.16: 67132484-67132484 on Build GRCh38
rs766075035 Chr.16: 67132450-67132450 on Build GRCh38
rs772645701 Chr.16: 67132444-67132444 on Build GRCh38
rs199531709 Chr.16: 67132807-67132807 on Build GRCh38
rs769158533 Chr.16: 67132809-67132809 on Build GRCh38
rs1046492815 Chr.16: 67132843-67132843 on Build GRCh38
rs370263639 Chr.16: 67132535-67132535 on Build GRCh38
rs762016680 Chr.16: 67132621-67132621 on Build GRCh38
rs866943646 Chr.16: 67132775-67132775 on Build GRCh38
rs746907083 Chr.16: 67132660-67132660 on Build GRCh38
rs763879335 Chr.16: 67132831-67132831 on Build GRCh38
rs748544133 Chr.16: 67132818-67132818 on Build GRCh38
rs756226782 Chr.16: 67132641-67132641 on Build GRCh38
rs775650504 Chr.16: 67132595-67132595 on Build GRCh38
rs754176825 Chr.16: 67132599-67132599 on Build GRCh38
rs781520128 Chr.16: 67132522-67132522 on Build GRCh38
rs762355761 Chr.16: 67132698-67132698 on Build GRCh38
rs374143121 Chr.16: 67132398-67132398 on Build GRCh38
rs912691355 Chr.16: 67132838-67132838 on Build GRCh38
rs538481613 Chr.16: 67132688-67132688 on Build GRCh38
rs182635625 Chr.16: 67132509-67132509 on Build GRCh38
rs777670612 Chr.16: 67132568-67132568 on Build GRCh38
rs534710152 Chr.16: 67132785-67132785 on Build GRCh38
rs565686389 Chr.16: 67132757-67132757 on Build GRCh38
rs918719052 Chr.16: 67132752-67132752 on Build GRCh38
rs951443520 Chr.16: 67132754-67132754 on Build GRCh38
rs764517594 Chr.16: 67132533-67132533 on Build GRCh38

View Full Product Details