Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67132735-67133232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf70
Entrez Gene ID
80262
Gene Name
chromosome 16 open reading frame 70
Gene Aliases
C16orf6, LIN10, lin-10
UniGene
Hs.513666
Gene Chromosome Location
Chr.16: 67109881-67148544 on Build GRCh38
Transcripts
NM_001320540.1 NM_001320541.1 NM_001320542.1 NM_001320543.1 NM_025187.4 XM_006721284.3 XM_011523350.2 XM_017023729.1 XM_017023730.1 XM_017023731.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1246535 c.389A>G p.N130S Chr.16: 67132850-67132850 on Build GRCh38
6228369 c.450+2T>C p.? Chr.16: 67132913-67132913 on Build GRCh38
6451071 c.366C>T p.S122S Chr.16: 67132827-67132827 on Build GRCh38
5636065 c.387C>T p.L129L Chr.16: 67132848-67132848 on Build GRCh38
7303826 c.402G>A p.L134L Chr.16: 67132863-67132863 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1011936690 Chr.16: 67133090-67133090 on Build GRCh38
rs748956301 Chr.16: 67132886-67132886 on Build GRCh38
rs748903129 Chr.16: 67132808-67132808 on Build GRCh38
rs757061777 Chr.16: 67132844-67132844 on Build GRCh38
rs1053527717 Chr.16: 67132932-67132932 on Build GRCh38
rs751420574 Chr.16: 67132837-67132837 on Build GRCh38
rs962835777 Chr.16: 67133109-67133109 on Build GRCh38
rs1056052409 Chr.16: 67133030-67133030 on Build GRCh38
rs112316420 Chr.16: 67133064-67133064 on Build GRCh38
rs772517492 Chr.16: 67132823-67132823 on Build GRCh38
rs377148729 Chr.16: 67132895-67132895 on Build GRCh38
rs572507258 Chr.16: 67133145-67133145 on Build GRCh38
rs771009481 Chr.16: 67132952-67132952 on Build GRCh38
rs1018225434 Chr.16: 67133088-67133088 on Build GRCh38
rs766438603 Chr.16: 67132827-67132827 on Build GRCh38
rs372791486 Chr.16: 67132826-67132826 on Build GRCh38
rs998155201 Chr.16: 67133176-67133176 on Build GRCh38
rs773619275 Chr.16: 67132905-67132905 on Build GRCh38
rs757074411 Chr.16: 67132804-67132804 on Build GRCh38
rs942754160 Chr.16: 67132839-67132839 on Build GRCh38
rs893663287 Chr.16: 67132966-67132966 on Build GRCh38
rs201455515 Chr.16: 67132810-67132810 on Build GRCh38
rs755603155 Chr.16: 67132878-67132878 on Build GRCh38
rs767403797 Chr.16: 67132848-67132848 on Build GRCh38
rs769504252 Chr.16: 67133189-67133189 on Build GRCh38
rs541517621 Chr.16: 67133177-67133177 on Build GRCh38
rs947680533 Chr.16: 67132893-67132893 on Build GRCh38
rs995445193 Chr.16: 67133121-67133121 on Build GRCh38
rs763824071 Chr.16: 67132778-67132778 on Build GRCh38
rs746956981 Chr.16: 67133018-67133018 on Build GRCh38
rs754747332 Chr.16: 67132887-67132887 on Build GRCh38
rs746402490 Chr.16: 67132940-67132940 on Build GRCh38
rs902184541 Chr.16: 67132953-67132953 on Build GRCh38
rs34017457 Chr.16: 67132828-67132828 on Build GRCh38
rs1023446203 Chr.16: 67133098-67133098 on Build GRCh38
rs975472375 Chr.16: 67133209-67133209 on Build GRCh38
rs776624021 Chr.16: 67132955-67132955 on Build GRCh38
rs1039926107 Chr.16: 67132851-67132851 on Build GRCh38
rs374172550 Chr.16: 67132805-67132805 on Build GRCh38
rs200907582 Chr.16: 67132863-67132863 on Build GRCh38
rs755620331 Chr.16: 67132806-67132806 on Build GRCh38
rs750334706 Chr.16: 67132859-67132859 on Build GRCh38
rs144322662 Chr.16: 67132892-67132892 on Build GRCh38
rs986834228 Chr.16: 67133163-67133163 on Build GRCh38
rs537008618 Chr.16: 67132817-67132817 on Build GRCh38
rs775018391 Chr.16: 67133201-67133201 on Build GRCh38
rs769158533 Chr.16: 67132809-67132809 on Build GRCh38
rs199531709 Chr.16: 67132807-67132807 on Build GRCh38
rs1025945226 Chr.16: 67133125-67133125 on Build GRCh38
rs1046492815 Chr.16: 67132843-67132843 on Build GRCh38
rs756802453 Chr.16: 67133058-67133058 on Build GRCh38
rs866943646 Chr.16: 67132775-67132775 on Build GRCh38
rs779117292 Chr.16: 67132846-67132847 on Build GRCh38
rs763879335 Chr.16: 67132831-67132831 on Build GRCh38
rs748544133 Chr.16: 67132818-67132818 on Build GRCh38
rs772447538 Chr.16: 67132896-67132896 on Build GRCh38
rs565466373 Chr.16: 67133131-67133131 on Build GRCh38
rs912691355 Chr.16: 67132838-67132838 on Build GRCh38
rs912554921 Chr.16: 67133167-67133167 on Build GRCh38
rs565686389 Chr.16: 67132757-67132757 on Build GRCh38
rs534710152 Chr.16: 67132785-67132785 on Build GRCh38
rs747499328 Chr.16: 67132922-67132922 on Build GRCh38
rs779655073 Chr.16: 67132884-67132884 on Build GRCh38

View Full Product Details