Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67134048-67134548 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf70
Entrez Gene ID
80262
Gene Name
chromosome 16 open reading frame 70
Gene Aliases
C16orf6, LIN10, lin-10
UniGene
Hs.513666
Gene Chromosome Location
Chr.16: 67109881-67148544 on Build GRCh38
Transcripts
NM_001320540.1 NM_001320541.1 NM_001320542.1 NM_001320543.1 NM_025187.4 XM_006721284.3 XM_011523350.2 XM_017023729.1 XM_017023730.1 XM_017023731.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5385150 c.646C>T p.R216C Chr.16: 67134452-67134452 on Build GRCh38
4061744 c.640C>T p.R214* Chr.16: 67134446-67134446 on Build GRCh38
6614562 c.647G>A p.R216H Chr.16: 67134453-67134453 on Build GRCh38
5942643 c.463C>T p.H155Y Chr.16: 67134180-67134180 on Build GRCh38
4975020 c.632C>T p.A211V Chr.16: 67134438-67134438 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745995284 Chr.16: 67134203-67134203 on Build GRCh38
rs1038722379 Chr.16: 67134523-67134523 on Build GRCh38
rs774416684 Chr.16: 67134406-67134406 on Build GRCh38
rs922306431 Chr.16: 67134275-67134275 on Build GRCh38
rs766417901 Chr.16: 67134149-67134149 on Build GRCh38
rs201686073 Chr.16: 67134253-67134253 on Build GRCh38
rs1008162760 Chr.16: 67134098-67134098 on Build GRCh38
rs781654200 Chr.16: 67134200-67134200 on Build GRCh38
rs751390076 Chr.16: 67134420-67134420 on Build GRCh38
rs752303249 Chr.16: 67134332-67134332 on Build GRCh38
rs747472160 Chr.16: 67134181-67134181 on Build GRCh38
rs997294582 Chr.16: 67134331-67134331 on Build GRCh38
rs746326539 Chr.16: 67134368-67134368 on Build GRCh38
rs756683132 Chr.16: 67134488-67134488 on Build GRCh38
rs760595107 Chr.16: 67134146-67134146 on Build GRCh38
rs769832515 Chr.16: 67134204-67134204 on Build GRCh38
rs368249714 Chr.16: 67134276-67134276 on Build GRCh38
rs763179851 Chr.16: 67134225-67134225 on Build GRCh38
rs143610025 Chr.16: 67134499-67134499 on Build GRCh38
rs982583396 Chr.16: 67134351-67134351 on Build GRCh38
rs767294977 Chr.16: 67134130-67134130 on Build GRCh38
rs753459309 Chr.16: 67134154-67134154 on Build GRCh38
rs556798418 Chr.16: 67134517-67134517 on Build GRCh38
rs369353129 Chr.16: 67134500-67134500 on Build GRCh38
rs748327050 Chr.16: 67134506-67134506 on Build GRCh38
rs780116527 Chr.16: 67134380-67134380 on Build GRCh38
rs768982228 Chr.16: 67134396-67134396 on Build GRCh38
rs759144652 Chr.16: 67134322-67134322 on Build GRCh38
rs761545720 Chr.16: 67134128-67134128 on Build GRCh38
rs758709116 Chr.16: 67134172-67134172 on Build GRCh38
rs781694028 Chr.16: 67134367-67134367 on Build GRCh38
rs775597376 Chr.16: 67134221-67134221 on Build GRCh38
rs775130293 Chr.16: 67134452-67134452 on Build GRCh38
rs771492177 Chr.16: 67134188-67134188 on Build GRCh38
rs758000382 Chr.16: 67134339-67134339 on Build GRCh38
rs762355142 Chr.16: 67134453-67134453 on Build GRCh38
rs753800144 Chr.16: 67134311-67134311 on Build GRCh38
rs1021618679 Chr.16: 67134348-67134348 on Build GRCh38
rs750343191 Chr.16: 67134140-67134140 on Build GRCh38
rs745586099 Chr.16: 67134509-67134511 on Build GRCh38
rs969235077 Chr.16: 67134355-67134355 on Build GRCh38
rs759485004 Chr.16: 67134446-67134446 on Build GRCh38
rs529669558 Chr.16: 67134132-67134132 on Build GRCh38
rs908396005 Chr.16: 67134494-67134494 on Build GRCh38
rs540784667 Chr.16: 67134074-67134074 on Build GRCh38
rs764655660 Chr.16: 67134447-67134447 on Build GRCh38
rs771770542 Chr.16: 67134226-67134226 on Build GRCh38
rs759605326 Chr.16: 67134118-67134118 on Build GRCh38
rs539405391 Chr.16: 67134478-67134478 on Build GRCh38
rs778138583 Chr.16: 67134177-67134177 on Build GRCh38
rs543975340 Chr.16: 67134083-67134083 on Build GRCh38
rs752350817 Chr.16: 67134169-67134169 on Build GRCh38
rs751181290 Chr.16: 67134479-67134479 on Build GRCh38
rs764787769 Chr.16: 67134328-67134328 on Build GRCh38
rs766477899 Chr.16: 67134279-67134279 on Build GRCh38
rs773128784 Chr.16: 67134250-67134250 on Build GRCh38
rs757649552 Chr.16: 67134482-67134482 on Build GRCh38
rs752017823 Chr.16: 67134362-67134362 on Build GRCh38
rs201249313 Chr.16: 67134390-67134390 on Build GRCh38
rs776727335 Chr.16: 67134438-67134438 on Build GRCh38
rs760589547 Chr.16: 67134419-67134419 on Build GRCh38
rs561022910 Chr.16: 67134075-67134075 on Build GRCh38
rs778752257 Chr.16: 67134162-67134162 on Build GRCh38
rs975187802 Chr.16: 67134247-67134247 on Build GRCh38
rs765500225 Chr.16: 67134123-67134123 on Build GRCh38
rs777285067 Chr.16: 67134343-67134343 on Build GRCh38
rs769877201 Chr.16: 67134374-67134374 on Build GRCh38
rs767811516 Chr.16: 67134487-67134487 on Build GRCh38
rs754518432 Chr.16: 67134156-67134156 on Build GRCh38
rs775758356 Chr.16: 67134125-67134125 on Build GRCh38

View Full Product Details