Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67140423-67140933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf70
Entrez Gene ID
80262
Gene Name
chromosome 16 open reading frame 70
Gene Aliases
C16orf6, LIN10, lin-10
UniGene
Hs.513666
Gene Chromosome Location
Chr.16: 67109881-67148544 on Build GRCh38
Transcripts
NM_001320540.1 NM_001320541.1 NM_001320542.1 NM_001320543.1 NM_025187.4 XM_006721284.3 XM_011523350.2 XM_017023729.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3712166 c.811C>T p.P271S Chr.16: 67140526-67140526 on Build GRCh38
6905809 c.824_830delAACAAGT p.K275fs*28 Chr.16: 67140539-67140545 on Build GRCh38
5971853 c.815C>T p.S272F Chr.16: 67140530-67140530 on Build GRCh38
1378830 c.853delT p.F286fs*19 Chr.16: 67140568-67140568 on Build GRCh38
4756787 c.869C>T p.T290I Chr.16: 67140584-67140584 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576671932 Chr.16: 67140758-67140758 on Build GRCh38
rs535720215 Chr.16: 67140527-67140527 on Build GRCh38
rs760183165 Chr.16: 67140474-67140474 on Build GRCh38
rs772876962 Chr.16: 67140467-67140467 on Build GRCh38
rs386791747 Chr.16: 67140458-67140459 on Build GRCh38
rs951666300 Chr.16: 67140841-67140841 on Build GRCh38
rs1054154248 Chr.16: 67140693-67140693 on Build GRCh38
rs975986204 Chr.16: 67140839-67140839 on Build GRCh38
rs753624189 Chr.16: 67140608-67140608 on Build GRCh38
rs1056717472 Chr.16: 67140661-67140661 on Build GRCh38
rs1025799124 Chr.16: 67140801-67140801 on Build GRCh38
rs902298498 Chr.16: 67140526-67140526 on Build GRCh38
rs753226741 Chr.16: 67140552-67140552 on Build GRCh38
rs201146623 Chr.16: 67140531-67140531 on Build GRCh38
rs141621654 Chr.16: 67140697-67140697 on Build GRCh38
rs373039690 Chr.16: 67140602-67140602 on Build GRCh38
rs376549916 Chr.16: 67140631-67140631 on Build GRCh38
rs896291048 Chr.16: 67140672-67140672 on Build GRCh38
rs898408256 Chr.16: 67140700-67140700 on Build GRCh38
rs1012584858 Chr.16: 67140696-67140696 on Build GRCh38
rs776931392 Chr.16: 67140498-67140498 on Build GRCh38
rs8047207 Chr.16: 67140459-67140459 on Build GRCh38
rs986876678 Chr.16: 67140770-67140770 on Build GRCh38
rs1043363525 Chr.16: 67140490-67140490 on Build GRCh38
rs150912761 Chr.16: 67140771-67140771 on Build GRCh38
rs370677267 Chr.16: 67140585-67140585 on Build GRCh38
rs1025633418 Chr.16: 67140743-67140743 on Build GRCh38
rs764221513 Chr.16: 67140557-67140557 on Build GRCh38
rs998296120 Chr.16: 67140796-67140796 on Build GRCh38
rs765896578 Chr.16: 67140533-67140533 on Build GRCh38
rs772858888 Chr.16: 67140568-67140568 on Build GRCh38
rs565207033 Chr.16: 67140816-67140816 on Build GRCh38
rs759861367 Chr.16: 67140601-67140602 on Build GRCh38
rs139477881 Chr.16: 67140735-67140735 on Build GRCh38
rs771335608 Chr.16: 67140495-67140495 on Build GRCh38
rs543292278 Chr.16: 67140579-67140579 on Build GRCh38
rs995536084 Chr.16: 67140709-67140709 on Build GRCh38
rs574370405 Chr.16: 67140567-67140567 on Build GRCh38
rs77081283 Chr.16: 67140458-67140458 on Build GRCh38
rs754734370 Chr.16: 67140629-67140629 on Build GRCh38
rs879207045 Chr.16: 67140861-67140861 on Build GRCh38
rs748025550 Chr.16: 67140632-67140632 on Build GRCh38

View Full Product Details