Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67179730-67180238 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0895L
Entrez Gene ID
653319
Gene Name
KIAA0895 like
Gene Aliases
-
UniGene
Hs.651201
Gene Chromosome Location
Chr.16: 67175602-67183980 on Build GRCh38
Transcripts
NM_001040715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
972310 c.543C>T p.C181C Chr.16: 67180068-67180068 on Build GRCh38
6693425 c.660C>A p.C220* Chr.16: 67179869-67179869 on Build GRCh38
972309 c.680G>A p.R227H Chr.16: 67179849-67179849 on Build GRCh38
3370498 c.526G>A p.V176M Chr.16: 67180085-67180085 on Build GRCh38
5685989 c.468C>T p.F156F Chr.16: 67180143-67180143 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750871268 Chr.16: 67180213-67180213 on Build GRCh38
rs780859480 Chr.16: 67179775-67179775 on Build GRCh38
rs1021585402 Chr.16: 67179906-67179906 on Build GRCh38
rs757976589 Chr.16: 67180169-67180169 on Build GRCh38
rs200813712 Chr.16: 67179994-67179994 on Build GRCh38
rs775570742 Chr.16: 67180024-67180024 on Build GRCh38
rs751306579 Chr.16: 67179917-67179917 on Build GRCh38
rs771027914 Chr.16: 67180010-67180010 on Build GRCh38
rs772573706 Chr.16: 67179846-67179846 on Build GRCh38
rs758761151 Chr.16: 67179993-67179993 on Build GRCh38
rs922060126 Chr.16: 67180002-67180002 on Build GRCh38
rs762128967 Chr.16: 67179904-67179933 on Build GRCh38
rs775420239 Chr.16: 67179887-67179899 on Build GRCh38
rs763798258 Chr.16: 67179915-67179915 on Build GRCh38
rs779892140 Chr.16: 67179818-67179818 on Build GRCh38
rs770970595 Chr.16: 67179874-67179874 on Build GRCh38
rs367663913 Chr.16: 67179979-67179979 on Build GRCh38
rs755680640 Chr.16: 67179813-67179813 on Build GRCh38
rs759677385 Chr.16: 67179924-67179924 on Build GRCh38
rs891585403 Chr.16: 67180181-67180181 on Build GRCh38
rs375360009 Chr.16: 67179850-67179850 on Build GRCh38
rs770109018 Chr.16: 67180022-67180022 on Build GRCh38
rs767476777 Chr.16: 67179886-67179887 on Build GRCh38
rs374180537 Chr.16: 67179826-67179826 on Build GRCh38
rs115387781 Chr.16: 67180021-67180021 on Build GRCh38
rs548676920 Chr.16: 67180209-67180209 on Build GRCh38
rs775829907 Chr.16: 67180126-67180126 on Build GRCh38
rs750354324 Chr.16: 67179963-67179963 on Build GRCh38
rs757608846 Chr.16: 67180191-67180191 on Build GRCh38
rs74997855 Chr.16: 67180078-67180078 on Build GRCh38
rs757700708 Chr.16: 67180096-67180096 on Build GRCh38
rs769731448 Chr.16: 67180123-67180123 on Build GRCh38
rs200015778 Chr.16: 67180162-67180162 on Build GRCh38
rs553644761 Chr.16: 67179823-67179823 on Build GRCh38
rs759761103 Chr.16: 67179893-67179893 on Build GRCh38
rs181055064 Chr.16: 67180090-67180090 on Build GRCh38
rs750011245 Chr.16: 67179795-67179795 on Build GRCh38
rs756238696 Chr.16: 67180214-67180214 on Build GRCh38
rs747495714 Chr.16: 67179998-67179998 on Build GRCh38
rs764635260 Chr.16: 67180166-67180166 on Build GRCh38
rs759515825 Chr.16: 67179884-67179885 on Build GRCh38
rs1022685349 Chr.16: 67179761-67179761 on Build GRCh38
rs77670236 Chr.16: 67179756-67179756 on Build GRCh38
rs772022702 Chr.16: 67180137-67180137 on Build GRCh38
rs760650817 Chr.16: 67180053-67180053 on Build GRCh38
rs562504503 Chr.16: 67179941-67179941 on Build GRCh38
rs185255358 Chr.16: 67180116-67180116 on Build GRCh38
rs779160394 Chr.16: 67179985-67179985 on Build GRCh38
rs760815129 Chr.16: 67179858-67179858 on Build GRCh38
rs758814175 Chr.16: 67180082-67180082 on Build GRCh38
rs373332286 Chr.16: 67180157-67180157 on Build GRCh38
rs933389416 Chr.16: 67180011-67180011 on Build GRCh38
rs774305093 Chr.16: 67179878-67179880 on Build GRCh38
rs748603543 Chr.16: 67179986-67179986 on Build GRCh38
rs773076731 Chr.16: 67180143-67180143 on Build GRCh38
rs761707067 Chr.16: 67180029-67180029 on Build GRCh38
rs770797546 Chr.16: 67179867-67179874 on Build GRCh38
rs13339140 Chr.16: 67180020-67180020 on Build GRCh38
rs368422262 Chr.16: 67179837-67179837 on Build GRCh38
rs767396852 Chr.16: 67180036-67180036 on Build GRCh38
rs760704082 Chr.16: 67180156-67180156 on Build GRCh38
rs776890188 Chr.16: 67179882-67179882 on Build GRCh38
rs955997882 Chr.16: 67179849-67179849 on Build GRCh38
rs755733593 Chr.16: 67179967-67179967 on Build GRCh38
rs905601272 Chr.16: 67179771-67179771 on Build GRCh38
rs749472286 Chr.16: 67180132-67180132 on Build GRCh38
rs1035548823 Chr.16: 67180175-67180175 on Build GRCh38
rs540172139 Chr.16: 67179764-67179764 on Build GRCh38
rs761495006 Chr.16: 67179929-67179929 on Build GRCh38
rs773548182 Chr.16: 67180089-67180089 on Build GRCh38
rs752264533 Chr.16: 67180077-67180077 on Build GRCh38
rs750488049 Chr.16: 67180117-67180118 on Build GRCh38
rs767629652 Chr.16: 67179945-67179945 on Build GRCh38
rs781533784 Chr.16: 67180106-67180106 on Build GRCh38
rs765440760 Chr.16: 67179966-67179966 on Build GRCh38
rs754432470 Chr.16: 67180068-67180068 on Build GRCh38
rs747516597 Chr.16: 67180095-67180095 on Build GRCh38
rs765992787 Chr.16: 67180054-67180054 on Build GRCh38
rs376400226 Chr.16: 67180067-67180067 on Build GRCh38
rs779659501 Chr.16: 67179973-67179973 on Build GRCh38
rs751974883 Chr.16: 67180180-67180180 on Build GRCh38
rs370460050 Chr.16: 67180173-67180173 on Build GRCh38
rs371060075 Chr.16: 67179824-67179824 on Build GRCh38
rs552506595 Chr.16: 67179812-67179812 on Build GRCh38
rs776428830 Chr.16: 67180163-67180163 on Build GRCh38
rs765011802 Chr.16: 67180070-67180070 on Build GRCh38
rs752420325 Chr.16: 67180167-67180167 on Build GRCh38
rs754594545 Chr.16: 67179984-67179984 on Build GRCh38
rs61733768 Chr.16: 67180047-67180047 on Build GRCh38

View Full Product Details