Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71850329-71850823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN1L
Entrez Gene ID
342371
Gene Name
ataxin 1 like
Gene Aliases
BOAT, BOAT1
UniGene
Hs.743239
Gene Chromosome Location
Chr.16: 71845991-71857333 on Build GRCh38
Transcripts
NM_001137675.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5584518 c.996C>T p.D332D Chr.16: 71850736-71850736 on Build GRCh38
5578094 c.1067G>A p.R356K Chr.16: 71850807-71850807 on Build GRCh38
5468951 c.842G>C p.R281T Chr.16: 71850582-71850582 on Build GRCh38
7325046 c.1067G>T p.R356M Chr.16: 71850807-71850807 on Build GRCh38
435727 c.682C>G p.P228A Chr.16: 71850422-71850422 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775797893 Chr.16: 71850732-71850732 on Build GRCh38
rs1006015843 Chr.16: 71850635-71850635 on Build GRCh38
rs866322022 Chr.16: 71850733-71850733 on Build GRCh38
rs994115061 Chr.16: 71850560-71850560 on Build GRCh38
rs567254924 Chr.16: 71850524-71850524 on Build GRCh38
rs756919884 Chr.16: 71850578-71850578 on Build GRCh38
rs117416318 Chr.16: 71850416-71850416 on Build GRCh38
rs868348852 Chr.16: 71850566-71850566 on Build GRCh38
rs926013235 Chr.16: 71850564-71850564 on Build GRCh38
rs370198764 Chr.16: 71850756-71850756 on Build GRCh38
rs762412751 Chr.16: 71850483-71850483 on Build GRCh38
rs532592720 Chr.16: 71850753-71850753 on Build GRCh38
rs1001164571 Chr.16: 71850354-71850354 on Build GRCh38
rs887516097 Chr.16: 71850387-71850387 on Build GRCh38
rs747026925 Chr.16: 71850711-71850711 on Build GRCh38
rs547099255 Chr.16: 71850684-71850684 on Build GRCh38
rs377449268 Chr.16: 71850790-71850790 on Build GRCh38
rs370296528 Chr.16: 71850598-71850598 on Build GRCh38
rs1053958402 Chr.16: 71850479-71850479 on Build GRCh38
rs887476053 Chr.16: 71850730-71850730 on Build GRCh38
rs913333887 Chr.16: 71850606-71850606 on Build GRCh38
rs948203709 Chr.16: 71850760-71850760 on Build GRCh38
rs964632237 Chr.16: 71850409-71850409 on Build GRCh38
rs778020746 Chr.16: 71850702-71850702 on Build GRCh38
rs761644064 Chr.16: 71850519-71850519 on Build GRCh38
rs1025708967 Chr.16: 71850366-71850366 on Build GRCh38
rs1016471557 Chr.16: 71850396-71850396 on Build GRCh38
rs930196806 Chr.16: 71850789-71850789 on Build GRCh38
rs779901674 Chr.16: 71850663-71850663 on Build GRCh38
rs572866356 Chr.16: 71850370-71850370 on Build GRCh38
rs140112104 Chr.16: 71850466-71850466 on Build GRCh38
rs117676797 Chr.16: 71850698-71850698 on Build GRCh38
rs182371255 Chr.16: 71850592-71850592 on Build GRCh38
rs749420720 Chr.16: 71850459-71850459 on Build GRCh38
rs769097768 Chr.16: 71850786-71850786 on Build GRCh38
rs147505763 Chr.16: 71850417-71850417 on Build GRCh38
rs781552390 Chr.16: 71850723-71850723 on Build GRCh38
rs957666092 Chr.16: 71850597-71850597 on Build GRCh38
rs187050076 Chr.16: 71850726-71850726 on Build GRCh38
rs906566178 Chr.16: 71850538-71850538 on Build GRCh38
rs765724239 Chr.16: 71850457-71850457 on Build GRCh38
rs1016132528 Chr.16: 71850655-71850655 on Build GRCh38
rs780571858 Chr.16: 71850497-71850497 on Build GRCh38
rs529794084 Chr.16: 71850505-71850505 on Build GRCh38
rs551247048 Chr.16: 71850782-71850782 on Build GRCh38
rs888437690 Chr.16: 71850799-71850799 on Build GRCh38
rs922972397 Chr.16: 71850420-71850420 on Build GRCh38
rs746422084 Chr.16: 71850570-71850570 on Build GRCh38
rs773859541 Chr.16: 71850788-71850788 on Build GRCh38
rs761404202 Chr.16: 71850755-71850755 on Build GRCh38
rs191663318 Chr.16: 71850425-71850425 on Build GRCh38
rs34672048 Chr.16: 71850383-71850384 on Build GRCh38
rs1046658433 Chr.16: 71850781-71850781 on Build GRCh38
rs199599582 Chr.16: 71850527-71850527 on Build GRCh38
rs748089660 Chr.16: 71850406-71850408 on Build GRCh38
rs755921391 Chr.16: 71850609-71850609 on Build GRCh38
rs767085819 Chr.16: 71850586-71850586 on Build GRCh38
rs978275409 Chr.16: 71850450-71850450 on Build GRCh38
rs1004676194 Chr.16: 71850398-71850398 on Build GRCh38
rs7194407 Chr.16: 71850677-71850677 on Build GRCh38
rs540357035 Chr.16: 71850395-71850395 on Build GRCh38
rs756080250 Chr.16: 71850361-71850361 on Build GRCh38
rs923560692 Chr.16: 71850727-71850727 on Build GRCh38
rs1045935881 Chr.16: 71850665-71850665 on Build GRCh38
rs1027330670 Chr.16: 71850375-71850375 on Build GRCh38
rs941494773 Chr.16: 71850800-71850800 on Build GRCh38
rs954344660 Chr.16: 71850439-71850439 on Build GRCh38
rs771346505 Chr.16: 71850721-71850721 on Build GRCh38
rs373371385 Chr.16: 71850766-71850766 on Build GRCh38
rs1030242624 Chr.16: 71850689-71850689 on Build GRCh38
rs916700136 Chr.16: 71850489-71850489 on Build GRCh38
rs866133545 Chr.16: 71850722-71850722 on Build GRCh38

View Full Product Details