Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71850699-71851193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN1L
Entrez Gene ID
342371
Gene Name
ataxin 1 like
Gene Aliases
BOAT, BOAT1
UniGene
Hs.743239
Gene Chromosome Location
Chr.16: 71845991-71857333 on Build GRCh38
Transcripts
NM_001137675.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5584518 c.996C>T p.D332D Chr.16: 71850736-71850736 on Build GRCh38
4463291 c.1285C>T p.L429L Chr.16: 71851025-71851025 on Build GRCh38
5578094 c.1067G>A p.R356K Chr.16: 71850807-71850807 on Build GRCh38
4458436 c.1085C>T p.P362L Chr.16: 71850825-71850825 on Build GRCh38
7325046 c.1067G>T p.R356M Chr.16: 71850807-71850807 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775797893 Chr.16: 71850732-71850732 on Build GRCh38
rs866322022 Chr.16: 71850733-71850733 on Build GRCh38
rs769862624 Chr.16: 71850980-71850980 on Build GRCh38
rs533644980 Chr.16: 71850976-71850976 on Build GRCh38
rs1032238423 Chr.16: 71851113-71851113 on Build GRCh38
rs934022280 Chr.16: 71851108-71851108 on Build GRCh38
rs1030698447 Chr.16: 71850947-71850947 on Build GRCh38
rs370198764 Chr.16: 71850756-71850756 on Build GRCh38
rs548784813 Chr.16: 71850882-71850882 on Build GRCh38
rs779433425 Chr.16: 71851174-71851174 on Build GRCh38
rs532592720 Chr.16: 71850753-71850753 on Build GRCh38
rs541759206 Chr.16: 71851157-71851157 on Build GRCh38
rs957937811 Chr.16: 71850951-71850951 on Build GRCh38
rs373000860 Chr.16: 71850843-71850843 on Build GRCh38
rs772952736 Chr.16: 71851016-71851016 on Build GRCh38
rs769139829 Chr.16: 71850937-71850937 on Build GRCh38
rs765889737 Chr.16: 71851081-71851081 on Build GRCh38
rs760396672 Chr.16: 71851080-71851080 on Build GRCh38
rs937572104 Chr.16: 71851024-71851024 on Build GRCh38
rs760424394 Chr.16: 71850860-71850860 on Build GRCh38
rs377449268 Chr.16: 71850790-71850790 on Build GRCh38
rs750414804 Chr.16: 71851111-71851111 on Build GRCh38
rs767212175 Chr.16: 71850842-71850842 on Build GRCh38
rs887476053 Chr.16: 71850730-71850730 on Build GRCh38
rs749553627 Chr.16: 71850982-71850982 on Build GRCh38
rs948203709 Chr.16: 71850760-71850760 on Build GRCh38
rs567078834 Chr.16: 71850913-71850913 on Build GRCh38
rs375831093 Chr.16: 71850918-71850918 on Build GRCh38
rs574410412 Chr.16: 71851107-71851107 on Build GRCh38
rs756367353 Chr.16: 71851112-71851112 on Build GRCh38
rs781607705 Chr.16: 71850930-71850930 on Build GRCh38
rs930196806 Chr.16: 71850789-71850789 on Build GRCh38
rs767730095 Chr.16: 71851110-71851110 on Build GRCh38
rs1010206057 Chr.16: 71851131-71851131 on Build GRCh38
rs1049348972 Chr.16: 71851039-71851039 on Build GRCh38
rs370440155 Chr.16: 71851094-71851094 on Build GRCh38
rs769097768 Chr.16: 71850786-71850786 on Build GRCh38
rs749715705 Chr.16: 71851117-71851117 on Build GRCh38
rs772242971 Chr.16: 71851050-71851050 on Build GRCh38
rs989267824 Chr.16: 71850957-71850957 on Build GRCh38
rs781552390 Chr.16: 71850723-71850723 on Build GRCh38
rs866479417 Chr.16: 71851162-71851162 on Build GRCh38
rs187050076 Chr.16: 71850726-71850726 on Build GRCh38
rs755309676 Chr.16: 71851128-71851128 on Build GRCh38
rs762483877 Chr.16: 71851091-71851091 on Build GRCh38
rs758373482 Chr.16: 71851109-71851109 on Build GRCh38
rs941346391 Chr.16: 71851096-71851096 on Build GRCh38
rs551247048 Chr.16: 71850782-71850782 on Build GRCh38
rs888437690 Chr.16: 71850799-71850799 on Build GRCh38
rs969081239 Chr.16: 71851067-71851067 on Build GRCh38
rs989239184 Chr.16: 71850846-71850846 on Build GRCh38
rs1033412513 Chr.16: 71850811-71850811 on Build GRCh38
rs773859541 Chr.16: 71850788-71850788 on Build GRCh38
rs761404202 Chr.16: 71850755-71850755 on Build GRCh38
rs867886138 Chr.16: 71850870-71850870 on Build GRCh38
rs754955575 Chr.16: 71850896-71850896 on Build GRCh38
rs890383264 Chr.16: 71850922-71850922 on Build GRCh38
rs536807147 Chr.16: 71850851-71850851 on Build GRCh38
rs774823153 Chr.16: 71851009-71851009 on Build GRCh38
rs746136067 Chr.16: 71850975-71850975 on Build GRCh38
rs780139575 Chr.16: 71851116-71851116 on Build GRCh38
rs1046658433 Chr.16: 71850781-71850781 on Build GRCh38
rs770410015 Chr.16: 71850917-71850917 on Build GRCh38
rs576720943 Chr.16: 71850981-71850981 on Build GRCh38
rs753671441 Chr.16: 71851090-71851090 on Build GRCh38
rs773921353 Chr.16: 71851132-71851132 on Build GRCh38
rs769214468 Chr.16: 71850983-71850983 on Build GRCh38
rs778926253 Chr.16: 71850897-71850897 on Build GRCh38
rs772556229 Chr.16: 71851070-71851070 on Build GRCh38
rs923560692 Chr.16: 71850727-71850727 on Build GRCh38
rs556013942 Chr.16: 71851104-71851104 on Build GRCh38
rs763389131 Chr.16: 71851168-71851168 on Build GRCh38
rs376116149 Chr.16: 71851106-71851106 on Build GRCh38
rs941494773 Chr.16: 71850800-71850800 on Build GRCh38
rs554657834 Chr.16: 71851167-71851167 on Build GRCh38
rs999813474 Chr.16: 71850945-71850945 on Build GRCh38
rs375970161 Chr.16: 71851069-71851069 on Build GRCh38
rs771346505 Chr.16: 71850721-71850721 on Build GRCh38
rs373371385 Chr.16: 71850766-71850766 on Build GRCh38
rs191148831 Chr.16: 71850977-71850977 on Build GRCh38
rs981599019 Chr.16: 71850962-71850962 on Build GRCh38
rs866133545 Chr.16: 71850722-71850722 on Build GRCh38
rs766226090 Chr.16: 71850872-71850872 on Build GRCh38

View Full Product Details