Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71852992-71853512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN1L
Entrez Gene ID
342371
Gene Name
ataxin 1 like
Gene Aliases
BOAT, BOAT1
UniGene
Hs.743239
Gene Chromosome Location
Chr.16: 71845991-71857333 on Build GRCh38
Transcripts
NM_001137675.3

SNP Detail

SNP ID Chromosome Location
rs961500185 Chr.16: 71853077-71853077 on Build GRCh38
rs139847997 Chr.16: 71853457-71853457 on Build GRCh38
rs937556153 Chr.16: 71853273-71853273 on Build GRCh38
rs537244609 Chr.16: 71853224-71853224 on Build GRCh38
rs915294636 Chr.16: 71853310-71853310 on Build GRCh38
rs995357575 Chr.16: 71853329-71853329 on Build GRCh38
rs561977775 Chr.16: 71853467-71853467 on Build GRCh38
rs1057070709 Chr.16: 71853292-71853292 on Build GRCh38
rs1026780299 Chr.16: 71853336-71853336 on Build GRCh38
rs774595362 Chr.16: 71853257-71853257 on Build GRCh38
rs755428803 Chr.16: 71853348-71853348 on Build GRCh38
rs1036837754 Chr.16: 71853318-71853318 on Build GRCh38
rs771262065 Chr.16: 71853249-71853250 on Build GRCh38
rs1056689691 Chr.16: 71853379-71853379 on Build GRCh38
rs962546501 Chr.16: 71853038-71853038 on Build GRCh38
rs188754021 Chr.16: 71853045-71853045 on Build GRCh38
rs186209757 Chr.16: 71853250-71853250 on Build GRCh38
rs564452090 Chr.16: 71853437-71853437 on Build GRCh38
rs140775247 Chr.16: 71853201-71853201 on Build GRCh38
rs895905506 Chr.16: 71853046-71853046 on Build GRCh38
rs200092995 Chr.16: 71853197-71853197 on Build GRCh38
rs372388329 Chr.16: 71853269-71853269 on Build GRCh38
rs554115135 Chr.16: 71853345-71853345 on Build GRCh38
rs747601723 Chr.16: 71853307-71853307 on Build GRCh38
rs146429807 Chr.16: 71853073-71853073 on Build GRCh38
rs1038453924 Chr.16: 71853443-71853443 on Build GRCh38
rs1026916058 Chr.16: 71853114-71853114 on Build GRCh38
rs16973477 Chr.16: 71853478-71853478 on Build GRCh38
rs897016276 Chr.16: 71853323-71853323 on Build GRCh38
rs868275670 Chr.16: 71853035-71853035 on Build GRCh38
rs572396999 Chr.16: 71853419-71853419 on Build GRCh38
rs867851791 Chr.16: 71853034-71853034 on Build GRCh38
rs780253232 Chr.16: 71853233-71853233 on Build GRCh38
rs1025467795 Chr.16: 71853250-71853250 on Build GRCh38
rs965190077 Chr.16: 71853212-71853212 on Build GRCh38
rs947543235 Chr.16: 71853257-71853257 on Build GRCh38
rs911919757 Chr.16: 71853065-71853065 on Build GRCh38
rs558123582 Chr.16: 71853165-71853165 on Build GRCh38
rs946865372 Chr.16: 71853107-71853107 on Build GRCh38
rs1019315020 Chr.16: 71853355-71853355 on Build GRCh38
rs570752205 Chr.16: 71853280-71853280 on Build GRCh38
rs939624292 Chr.16: 71853185-71853185 on Build GRCh38
rs924181183 Chr.16: 71853184-71853184 on Build GRCh38
rs531828886 Chr.16: 71853025-71853025 on Build GRCh38
rs533403526 Chr.16: 71853436-71853436 on Build GRCh38
rs556665025 Chr.16: 71853229-71853229 on Build GRCh38
rs939571886 Chr.16: 71853376-71853379 on Build GRCh38
rs927433673 Chr.16: 71853474-71853474 on Build GRCh38
rs112686816 Chr.16: 71853134-71853134 on Build GRCh38
rs574893300 Chr.16: 71853302-71853302 on Build GRCh38
rs924130542 Chr.16: 71853350-71853350 on Build GRCh38
rs889714178 Chr.16: 71853347-71853347 on Build GRCh38
rs550686790 Chr.16: 71853164-71853164 on Build GRCh38
rs1046865903 Chr.16: 71853472-71853472 on Build GRCh38
rs565565132 Chr.16: 71853081-71853081 on Build GRCh38
rs955271521 Chr.16: 71853252-71853252 on Build GRCh38
rs576184678 Chr.16: 71853450-71853450 on Build GRCh38
rs539714429 Chr.16: 71853159-71853159 on Build GRCh38
rs530321566 Chr.16: 71853188-71853188 on Build GRCh38
rs180775544 Chr.16: 71853047-71853047 on Build GRCh38
rs1006803874 Chr.16: 71853354-71853354 on Build GRCh38
rs546469681 Chr.16: 71853427-71853427 on Build GRCh38
rs987429390 Chr.16: 71853059-71853059 on Build GRCh38
rs978873122 Chr.16: 71853264-71853264 on Build GRCh38
rs779294175 Chr.16: 71853020-71853020 on Build GRCh38
rs1018620086 Chr.16: 71853258-71853259 on Build GRCh38
rs965397792 Chr.16: 71853426-71853426 on Build GRCh38
rs549427936 Chr.16: 71853026-71853026 on Build GRCh38
rs964715433 Chr.16: 71853285-71853285 on Build GRCh38
rs975151075 Chr.16: 71853286-71853286 on Build GRCh38
rs915329420 Chr.16: 71853085-71853085 on Build GRCh38
rs758867760 Chr.16: 71853099-71853099 on Build GRCh38
rs541993583 Chr.16: 71853317-71853317 on Build GRCh38

View Full Product Details