Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 81045591-81046103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATMIN
Entrez Gene ID
23300
Gene Name
ATM interactor
Gene Aliases
ASCIZ, ZNF822
UniGene
Hs.16349
Gene Chromosome Location
Chr.16: 81035853-81047346 on Build GRCh38
Transcripts
NM_001300728.1 NM_015251.2

SNP Detail

SNP ID Chromosome Location
rs1029789580 Chr.16: 81045637-81045637 on Build GRCh38
rs2278025 Chr.16: 81045636-81045636 on Build GRCh38
rs138737402 Chr.16: 81045702-81045702 on Build GRCh38
rs2278024 Chr.16: 81045628-81045628 on Build GRCh38
rs564559591 Chr.16: 81045676-81045676 on Build GRCh38
rs956472082 Chr.16: 81045907-81045907 on Build GRCh38
rs1018925629 Chr.16: 81045759-81045759 on Build GRCh38
rs376517907 Chr.16: 81045985-81045987 on Build GRCh38
rs980156818 Chr.16: 81046008-81046008 on Build GRCh38
rs913480552 Chr.16: 81046075-81046075 on Build GRCh38
rs946989900 Chr.16: 81045738-81045738 on Build GRCh38
rs927708054 Chr.16: 81045718-81045718 on Build GRCh38
rs953777988 Chr.16: 81045651-81045652 on Build GRCh38
rs1029423109 Chr.16: 81045632-81045632 on Build GRCh38
rs533884367 Chr.16: 81045902-81045902 on Build GRCh38
rs187831546 Chr.16: 81045819-81045819 on Build GRCh38
rs751800936 Chr.16: 81045909-81045909 on Build GRCh38
rs993558557 Chr.16: 81045798-81045798 on Build GRCh38
rs549598996 Chr.16: 81046059-81046059 on Build GRCh38
rs541601625 Chr.16: 81046069-81046069 on Build GRCh38
rs1050900234 Chr.16: 81045806-81045806 on Build GRCh38
rs752596659 Chr.16: 81045746-81045746 on Build GRCh38
rs889155412 Chr.16: 81045753-81045753 on Build GRCh38
rs142673997 Chr.16: 81045714-81045714 on Build GRCh38
rs888314803 Chr.16: 81046053-81046053 on Build GRCh38
rs1006961980 Chr.16: 81045663-81045663 on Build GRCh38
rs13861 Chr.16: 81046076-81046076 on Build GRCh38
rs746613942 Chr.16: 81045631-81045631 on Build GRCh38
rs867396684 Chr.16: 81045955-81045955 on Build GRCh38
rs748007242 Chr.16: 81045716-81045722 on Build GRCh38
rs145841446 Chr.16: 81045694-81045694 on Build GRCh38
rs1053333336 Chr.16: 81045794-81045796 on Build GRCh38
rs972570049 Chr.16: 81045681-81045681 on Build GRCh38
rs2549890 Chr.16: 81045794-81045794 on Build GRCh38
rs115565389 Chr.16: 81045853-81045853 on Build GRCh38
rs531016238 Chr.16: 81045688-81045688 on Build GRCh38
rs749795984 Chr.16: 81045817-81045817 on Build GRCh38
rs1025476116 Chr.16: 81045927-81045927 on Build GRCh38
rs970527253 Chr.16: 81046001-81046001 on Build GRCh38
rs34093092 Chr.16: 81045977-81045978 on Build GRCh38
rs182761823 Chr.16: 81045679-81045679 on Build GRCh38
rs910814923 Chr.16: 81045991-81045991 on Build GRCh38
rs557198327 Chr.16: 81045995-81045995 on Build GRCh38
rs756000487 Chr.16: 81045908-81045908 on Build GRCh38
rs1019037141 Chr.16: 81045675-81045675 on Build GRCh38
rs574750843 Chr.16: 81045970-81045970 on Build GRCh38
rs998759835 Chr.16: 81045802-81045802 on Build GRCh38
rs984465562 Chr.16: 81045814-81045814 on Build GRCh38
rs1008897459 Chr.16: 81045754-81045754 on Build GRCh38
rs1024420845 Chr.16: 81045993-81045993 on Build GRCh38
rs952560829 Chr.16: 81045813-81045813 on Build GRCh38
rs763836216 Chr.16: 81045901-81045901 on Build GRCh38
rs936254848 Chr.16: 81045777-81045777 on Build GRCh38
rs555233207 Chr.16: 81045912-81045912 on Build GRCh38
rs75954720 Chr.16: 81045732-81045732 on Build GRCh38
rs1025494368 Chr.16: 81045805-81045805 on Build GRCh38
rs946902916 Chr.16: 81045929-81045929 on Build GRCh38
rs984024796 Chr.16: 81045695-81045695 on Build GRCh38
rs113636244 Chr.16: 81045676-81045676 on Build GRCh38
rs146931181 Chr.16: 81045725-81045725 on Build GRCh38
rs528822368 Chr.16: 81045711-81045711 on Build GRCh38
rs1040418627 Chr.16: 81045997-81045997 on Build GRCh38
rs1025445119 Chr.16: 81045682-81045682 on Build GRCh38
rs969493756 Chr.16: 81045855-81045855 on Build GRCh38
rs935046707 Chr.16: 81045721-81045721 on Build GRCh38
rs142005911 Chr.16: 81045856-81045856 on Build GRCh38
rs568103094 Chr.16: 81045723-81045723 on Build GRCh38
rs542160465 Chr.16: 81045994-81045994 on Build GRCh38
rs13185 Chr.16: 81046036-81046036 on Build GRCh38
rs776140756 Chr.16: 81045677-81045677 on Build GRCh38
rs915419376 Chr.16: 81045920-81045920 on Build GRCh38
rs150702436 Chr.16: 81046045-81046045 on Build GRCh38
rs889111860 Chr.16: 81045633-81045633 on Build GRCh38
rs552553788 Chr.16: 81045831-81045832 on Build GRCh38
rs62054592 Chr.16: 81046017-81046017 on Build GRCh38
rs960610724 Chr.16: 81045959-81045960 on Build GRCh38
rs1051154495 Chr.16: 81045935-81045935 on Build GRCh38
rs1023546836 Chr.16: 81045849-81045849 on Build GRCh38

View Full Product Details