Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 84439074-84439566 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2C2
Entrez Gene ID
9914
Gene Name
ATPase secretory pathway Ca2+ transporting 2
Gene Aliases
SPCA2
UniGene
Hs.6168
Gene Chromosome Location
Chr.16: 84368523-84465777 on Build GRCh38
Transcripts
NM_001286527.2 NM_001291454.1 NM_014861.3 XM_011523486.2 XM_011523487.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
974393 c.1157C>T p.A386V Chr.16: 84439472-84439472 on Build GRCh38
145122 c.1207G>A p.E403K Chr.16: 84439522-84439522 on Build GRCh38
4664398 c.1024G>A p.V342I Chr.16: 84439203-84439203 on Build GRCh38
5851630 c.1081G>A p.V361M Chr.16: 84439260-84439260 on Build GRCh38
5527140 c.1054C>T p.L352L Chr.16: 84439233-84439233 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374260157 Chr.16: 84439141-84439141 on Build GRCh38
rs751429844 Chr.16: 84439522-84439522 on Build GRCh38
rs996651779 Chr.16: 84439351-84439351 on Build GRCh38
rs751694377 Chr.16: 84439268-84439268 on Build GRCh38
rs777942282 Chr.16: 84439225-84439225 on Build GRCh38
rs938164502 Chr.16: 84439250-84439250 on Build GRCh38
rs200192229 Chr.16: 84439475-84439475 on Build GRCh38
rs150034273 Chr.16: 84439159-84439159 on Build GRCh38
rs141384127 Chr.16: 84439379-84439379 on Build GRCh38
rs778871153 Chr.16: 84439333-84439333 on Build GRCh38
rs762036909 Chr.16: 84439321-84439321 on Build GRCh38
rs1015715157 Chr.16: 84439365-84439365 on Build GRCh38
rs973174770 Chr.16: 84439166-84439166 on Build GRCh38
rs765718239 Chr.16: 84439216-84439216 on Build GRCh38
rs746597904 Chr.16: 84439452-84439452 on Build GRCh38
rs247905 Chr.16: 84439366-84439366 on Build GRCh38
rs765221081 Chr.16: 84439137-84439137 on Build GRCh38
rs752496416 Chr.16: 84439516-84439516 on Build GRCh38
rs766677783 Chr.16: 84439426-84439426 on Build GRCh38
rs760624027 Chr.16: 84439103-84439103 on Build GRCh38
rs538323406 Chr.16: 84439301-84439301 on Build GRCh38
rs758446158 Chr.16: 84439220-84439220 on Build GRCh38
rs1028051198 Chr.16: 84439191-84439191 on Build GRCh38
rs553122232 Chr.16: 84439356-84439356 on Build GRCh38
rs247904 Chr.16: 84439531-84439531 on Build GRCh38
rs869200159 Chr.16: 84439401-84439402 on Build GRCh38
rs755829920 Chr.16: 84439542-84439542 on Build GRCh38
rs772890980 Chr.16: 84439291-84439291 on Build GRCh38
rs199811419 Chr.16: 84439401-84439401 on Build GRCh38
rs763161332 Chr.16: 84439313-84439313 on Build GRCh38
rs145163175 Chr.16: 84439417-84439417 on Build GRCh38
rs377016980 Chr.16: 84439158-84439158 on Build GRCh38
rs367635448 Chr.16: 84439227-84439227 on Build GRCh38
rs986836539 Chr.16: 84439200-84439200 on Build GRCh38
rs377175065 Chr.16: 84439147-84439147 on Build GRCh38
rs774881764 Chr.16: 84439471-84439471 on Build GRCh38
rs201133834 Chr.16: 84439492-84439492 on Build GRCh38
rs775041439 Chr.16: 84439408-84439408 on Build GRCh38
rs368020130 Chr.16: 84439236-84439236 on Build GRCh38
rs904549110 Chr.16: 84439106-84439106 on Build GRCh38
rs761475103 Chr.16: 84439203-84439203 on Build GRCh38
rs777902681 Chr.16: 84439167-84439167 on Build GRCh38
rs558000536 Chr.16: 84439324-84439324 on Build GRCh38
rs80000075 Chr.16: 84439169-84439169 on Build GRCh38
rs371722139 Chr.16: 84439521-84439521 on Build GRCh38
rs146656709 Chr.16: 84439503-84439503 on Build GRCh38
rs762458619 Chr.16: 84439146-84439146 on Build GRCh38
rs995109753 Chr.16: 84439539-84439539 on Build GRCh38
rs776144986 Chr.16: 84439393-84439393 on Build GRCh38
rs765383247 Chr.16: 84439508-84439508 on Build GRCh38
rs760853605 Chr.16: 84439487-84439487 on Build GRCh38
rs775494516 Chr.16: 84439186-84439186 on Build GRCh38
rs372130717 Chr.16: 84439493-84439493 on Build GRCh38
rs367904279 Chr.16: 84439449-84439449 on Build GRCh38
rs769534792 Chr.16: 84439461-84439461 on Build GRCh38
rs201882619 Chr.16: 84439121-84439121 on Build GRCh38
rs374715934 Chr.16: 84439206-84439206 on Build GRCh38
rs371072369 Chr.16: 84439202-84439202 on Build GRCh38
rs770622432 Chr.16: 84439392-84439392 on Build GRCh38
rs371551354 Chr.16: 84439231-84439231 on Build GRCh38
rs201069944 Chr.16: 84439513-84439513 on Build GRCh38
rs752102779 Chr.16: 84439320-84439320 on Build GRCh38
rs373385435 Chr.16: 84439352-84439352 on Build GRCh38
rs377115423 Chr.16: 84439245-84439245 on Build GRCh38
rs180864274 Chr.16: 84439230-84439230 on Build GRCh38
rs766441764 Chr.16: 84439329-84439329 on Build GRCh38
rs753984302 Chr.16: 84439427-84439427 on Build GRCh38
rs781625415 Chr.16: 84439273-84439273 on Build GRCh38
rs780950794 Chr.16: 84439457-84439457 on Build GRCh38
rs566637685 Chr.16: 84439431-84439431 on Build GRCh38
rs771774140 Chr.16: 84439251-84439251 on Build GRCh38
rs372103955 Chr.16: 84439472-84439472 on Build GRCh38
rs893021348 Chr.16: 84439485-84439485 on Build GRCh38
rs369924720 Chr.16: 84439149-84439149 on Build GRCh38
rs386385280 Chr.16: 84439399-84439400 on Build GRCh38
rs980304486 Chr.16: 84439311-84439311 on Build GRCh38
rs374714353 Chr.16: 84439233-84439233 on Build GRCh38
rs753730436 Chr.16: 84439331-84439331 on Build GRCh38
rs776751824 Chr.16: 84439124-84439124 on Build GRCh38
rs935938382 Chr.16: 84439368-84439368 on Build GRCh38
rs62640931 Chr.16: 84439181-84439181 on Build GRCh38
rs186501679 Chr.16: 84439517-84439517 on Build GRCh38
rs779919296 Chr.16: 84439278-84439278 on Build GRCh38
rs200849669 Chr.16: 84439286-84439286 on Build GRCh38
rs577967745 Chr.16: 84439326-84439326 on Build GRCh38
rs145373170 Chr.16: 84439362-84439362 on Build GRCh38
rs750654957 Chr.16: 84439277-84439277 on Build GRCh38
rs943221481 Chr.16: 84439363-84439363 on Build GRCh38
rs374834608 Chr.16: 84439398-84439398 on Build GRCh38
rs376382339 Chr.16: 84439198-84439198 on Build GRCh38
rs892990969 Chr.16: 84439349-84439349 on Build GRCh38
rs200651927 Chr.16: 84439403-84439403 on Build GRCh38
rs1032908960 Chr.16: 84439339-84439339 on Build GRCh38
rs369305687 Chr.16: 84439305-84439305 on Build GRCh38
rs767097973 Chr.16: 84439205-84439205 on Build GRCh38
rs201256097 Chr.16: 84439252-84439252 on Build GRCh38
rs370140600 Chr.16: 84439424-84439424 on Build GRCh38
rs776593054 Chr.16: 84439177-84439177 on Build GRCh38
rs377058044 Chr.16: 84439430-84439430 on Build GRCh38
rs373098892 Chr.16: 84439154-84439154 on Build GRCh38
rs773656054 Chr.16: 84439396-84439397 on Build GRCh38
rs370709927 Chr.16: 84439162-84439162 on Build GRCh38
rs1017586814 Chr.16: 84439101-84439101 on Build GRCh38
rs974945488 Chr.16: 84439506-84439506 on Build GRCh38
rs748974485 Chr.16: 84439285-84439285 on Build GRCh38
rs911675471 Chr.16: 84439497-84439497 on Build GRCh38
rs746834286 Chr.16: 84439120-84439120 on Build GRCh38
rs773474885 Chr.16: 84439118-84439118 on Build GRCh38
rs911707967 Chr.16: 84439361-84439361 on Build GRCh38
rs752437549 Chr.16: 84439443-84439443 on Build GRCh38
rs762429097 Chr.16: 84439188-84439188 on Build GRCh38
rs199936161 Chr.16: 84439269-84439269 on Build GRCh38
rs774881251 Chr.16: 84439144-84439144 on Build GRCh38
rs3085208 Chr.16: 84439398-84439399 on Build GRCh38
rs373034436 Chr.16: 84439323-84439323 on Build GRCh38
rs771547390 Chr.16: 84439290-84439290 on Build GRCh38
rs779845321 Chr.16: 84439545-84439545 on Build GRCh38
rs745482175 Chr.16: 84439458-84439458 on Build GRCh38
rs759581070 Chr.16: 84439130-84439130 on Build GRCh38
rs377104847 Chr.16: 84439289-84439289 on Build GRCh38
rs778219893 Chr.16: 84439288-84439288 on Build GRCh38
rs902511446 Chr.16: 84439307-84439307 on Build GRCh38
rs1000265893 Chr.16: 84439107-84439107 on Build GRCh38
rs748045886 Chr.16: 84439378-84439378 on Build GRCh38
rs554219991 Chr.16: 84439358-84439358 on Build GRCh38
rs758346688 Chr.16: 84439445-84439445 on Build GRCh38
rs561833219 Chr.16: 84439367-84439367 on Build GRCh38
rs754712148 Chr.16: 84439332-84439332 on Build GRCh38
rs745605167 Chr.16: 84439179-84439179 on Build GRCh38
rs759942295 Chr.16: 84439257-84439257 on Build GRCh38
rs768426478 Chr.16: 84439238-84439238 on Build GRCh38
rs749901458 Chr.16: 84439535-84439535 on Build GRCh38
rs999214585 Chr.16: 84439322-84439322 on Build GRCh38
rs752961744 Chr.16: 84439260-84439260 on Build GRCh38
rs564070721 Chr.16: 84439423-84439423 on Build GRCh38
rs34912793 Chr.16: 84439104-84439105 on Build GRCh38
rs771241885 Chr.16: 84439176-84439176 on Build GRCh38
rs1024432931 Chr.16: 84439132-84439132 on Build GRCh38
rs950385339 Chr.16: 84439490-84439490 on Build GRCh38
rs763539539 Chr.16: 84439265-84439265 on Build GRCh38
rs79209156 Chr.16: 84439185-84439185 on Build GRCh38
rs147201753 Chr.16: 84439177-84439178 on Build GRCh38
rs764765052 Chr.16: 84439315-84439315 on Build GRCh38
rs564091352 Chr.16: 84439160-84439160 on Build GRCh38
rs202026876 Chr.16: 84439438-84439438 on Build GRCh38
rs906039982 Chr.16: 84439422-84439422 on Build GRCh38
rs769124014 Chr.16: 84439407-84439407 on Build GRCh38
rs756834716 Chr.16: 84439524-84439524 on Build GRCh38
rs769900943 Chr.16: 84439394-84439395 on Build GRCh38
rs970262523 Chr.16: 84439133-84439133 on Build GRCh38
rs1032210135 Chr.16: 84439110-84439110 on Build GRCh38
rs372231003 Chr.16: 84439237-84439237 on Build GRCh38
rs186295467 Chr.16: 84439115-84439115 on Build GRCh38
rs753223933 Chr.16: 84439217-84439217 on Build GRCh38
rs201782040 Chr.16: 84439243-84439243 on Build GRCh38
rs375518718 Chr.16: 84439387-84439387 on Build GRCh38
rs918947430 Chr.16: 84439350-84439350 on Build GRCh38
rs375931622 Chr.16: 84439259-84439259 on Build GRCh38

View Full Product Details