Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 84872253-84872777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRISPLD2
Entrez Gene ID
83716
Gene Name
cysteine rich secretory protein LCCL domain containing 2
Gene Aliases
CRISP11, LCRISP2
UniGene
Hs.513779
Gene Chromosome Location
Chr.16: 84819981-84909510 on Build GRCh38
Transcripts
NM_031476.3 XM_005256190.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5894605 c.915-6C>T p.? Chr.16: 84872436-84872436 on Build GRCh38
6482504 c.921G>T p.Q307H Chr.16: 84872448-84872448 on Build GRCh38
6482505 c.921G>T p.Q307H Chr.16: 84872448-84872448 on Build GRCh38
6635542 c.957C>T p.I319I Chr.16: 84872484-84872484 on Build GRCh38
5894604 c.915-6C>T p.? Chr.16: 84872436-84872436 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs721005 Chr.16: 84872492-84872492 on Build GRCh38
rs777013270 Chr.16: 84872727-84872727 on Build GRCh38
rs192547375 Chr.16: 84872420-84872420 on Build GRCh38
rs751616098 Chr.16: 84872408-84872408 on Build GRCh38
rs568906326 Chr.16: 84872402-84872402 on Build GRCh38
rs541850470 Chr.16: 84872395-84872395 on Build GRCh38
rs922026360 Chr.16: 84872362-84872362 on Build GRCh38
rs1009139293 Chr.16: 84872371-84872371 on Build GRCh38
rs1003197762 Chr.16: 84872476-84872476 on Build GRCh38
rs548950008 Chr.16: 84872530-84872530 on Build GRCh38
rs564309410 Chr.16: 84872452-84872452 on Build GRCh38
rs369203821 Chr.16: 84872557-84872557 on Build GRCh38
rs757970559 Chr.16: 84872517-84872517 on Build GRCh38
rs745653374 Chr.16: 84872606-84872606 on Build GRCh38
rs970025329 Chr.16: 84872633-84872633 on Build GRCh38
rs1023837138 Chr.16: 84872430-84872430 on Build GRCh38
rs902451759 Chr.16: 84872453-84872453 on Build GRCh38
rs948182242 Chr.16: 84872281-84872281 on Build GRCh38
rs1023619393 Chr.16: 84872605-84872605 on Build GRCh38
rs990960124 Chr.16: 84872284-84872284 on Build GRCh38
rs929149918 Chr.16: 84872682-84872682 on Build GRCh38
rs984063106 Chr.16: 84872526-84872526 on Build GRCh38
rs372522087 Chr.16: 84872484-84872484 on Build GRCh38
rs759944232 Chr.16: 84872401-84872401 on Build GRCh38
rs749176858 Chr.16: 84872392-84872392 on Build GRCh38
rs894823197 Chr.16: 84872675-84872675 on Build GRCh38
rs560611349 Chr.16: 84872501-84872501 on Build GRCh38
rs935123347 Chr.16: 84872313-84872313 on Build GRCh38
rs920782102 Chr.16: 84872298-84872298 on Build GRCh38
rs771433504 Chr.16: 84872700-84872700 on Build GRCh38
rs746467916 Chr.16: 84872520-84872520 on Build GRCh38
rs757377904 Chr.16: 84872410-84872410 on Build GRCh38
rs1015520321 Chr.16: 84872747-84872747 on Build GRCh38
rs927693949 Chr.16: 84872311-84872311 on Build GRCh38
rs905163215 Chr.16: 84872715-84872715 on Build GRCh38
rs887546993 Chr.16: 84872719-84872719 on Build GRCh38
rs139614090 Chr.16: 84872396-84872396 on Build GRCh38
rs201194618 Chr.16: 84872499-84872499 on Build GRCh38
rs908153058 Chr.16: 84872351-84872351 on Build GRCh38
rs781124473 Chr.16: 84872511-84872511 on Build GRCh38
rs990186627 Chr.16: 84872575-84872575 on Build GRCh38
rs753604192 Chr.16: 84872509-84872509 on Build GRCh38
rs201861816 Chr.16: 84872477-84872477 on Build GRCh38
rs764140927 Chr.16: 84872509-84872510 on Build GRCh38
rs889024885 Chr.16: 84872525-84872525 on Build GRCh38
rs377233935 Chr.16: 84872534-84872534 on Build GRCh38
rs544274712 Chr.16: 84872686-84872686 on Build GRCh38
rs721004 Chr.16: 84872374-84872374 on Build GRCh38
rs776310093 Chr.16: 84872524-84872524 on Build GRCh38
rs1005036578 Chr.16: 84872745-84872745 on Build GRCh38
rs180786700 Chr.16: 84872693-84872693 on Build GRCh38
rs764547273 Chr.16: 84872489-84872489 on Build GRCh38
rs367754049 Chr.16: 84872679-84872679 on Build GRCh38
rs1010590193 Chr.16: 84872325-84872325 on Build GRCh38
rs370188694 Chr.16: 84872441-84872441 on Build GRCh38
rs747697897 Chr.16: 84872459-84872459 on Build GRCh38
rs368001426 Chr.16: 84872478-84872478 on Build GRCh38
rs151045407 Chr.16: 84872674-84872674 on Build GRCh38
rs368398329 Chr.16: 84872573-84872573 on Build GRCh38
rs573913693 Chr.16: 84872394-84872394 on Build GRCh38
rs555303283 Chr.16: 84872669-84872669 on Build GRCh38
rs1040799855 Chr.16: 84872361-84872361 on Build GRCh38
rs778872270 Chr.16: 84872445-84872445 on Build GRCh38
rs531398640 Chr.16: 84872537-84872537 on Build GRCh38
rs370539314 Chr.16: 84872442-84872442 on Build GRCh38
rs760765774 Chr.16: 84872541-84872541 on Build GRCh38
rs779717506 Chr.16: 84872427-84872427 on Build GRCh38
rs914700154 Chr.16: 84872577-84872577 on Build GRCh38
rs1052512545 Chr.16: 84872314-84872314 on Build GRCh38
rs762410863 Chr.16: 84872535-84872535 on Build GRCh38
rs754070036 Chr.16: 84872548-84872548 on Build GRCh38
rs988348045 Chr.16: 84872645-84872645 on Build GRCh38
rs748430936 Chr.16: 84872707-84872707 on Build GRCh38
rs188468122 Chr.16: 84872538-84872538 on Build GRCh38
rs142773203 Chr.16: 84872656-84872656 on Build GRCh38
rs983710378 Chr.16: 84872339-84872339 on Build GRCh38
rs368558267 Chr.16: 84872528-84872528 on Build GRCh38
rs116554813 Chr.16: 84872644-84872644 on Build GRCh38
rs571367690 Chr.16: 84872359-84872359 on Build GRCh38
rs767898029 Chr.16: 84872418-84872418 on Build GRCh38
rs762038885 Chr.16: 84872494-84872494 on Build GRCh38
rs540463871 Chr.16: 84872698-84872698 on Build GRCh38
rs759308168 Chr.16: 84872550-84872550 on Build GRCh38
rs772821614 Chr.16: 84872398-84872398 on Build GRCh38
rs770515424 Chr.16: 84872522-84872522 on Build GRCh38
rs763378252 Chr.16: 84872405-84872405 on Build GRCh38
rs144130319 Chr.16: 84872561-84872561 on Build GRCh38
rs1018809198 Chr.16: 84872372-84872372 on Build GRCh38
rs533386261 Chr.16: 84872486-84872486 on Build GRCh38
rs921955374 Chr.16: 84872563-84872563 on Build GRCh38
rs966296165 Chr.16: 84872753-84872753 on Build GRCh38
rs578076706 Chr.16: 84872694-84872694 on Build GRCh38
rs544311455 Chr.16: 84872437-84872437 on Build GRCh38
rs148703123 Chr.16: 84872643-84872643 on Build GRCh38
rs994242490 Chr.16: 84872385-84872385 on Build GRCh38
rs772393787 Chr.16: 84872716-84872716 on Build GRCh38
rs777301231 Chr.16: 84872421-84872421 on Build GRCh38
rs376224145 Chr.16: 84872542-84872542 on Build GRCh38
rs748066080 Chr.16: 84872514-84872514 on Build GRCh38
rs553761148 Chr.16: 84872393-84872393 on Build GRCh38
rs1046597584 Chr.16: 84872309-84872309 on Build GRCh38
rs533732838 Chr.16: 84872614-84872614 on Build GRCh38
rs76393375 Chr.16: 84872322-84872322 on Build GRCh38
rs184877327 Chr.16: 84872508-84872508 on Build GRCh38
rs777274346 Chr.16: 84872519-84872519 on Build GRCh38
rs985776390 Chr.16: 84872665-84872665 on Build GRCh38
rs377006441 Chr.16: 84872487-84872487 on Build GRCh38
rs112893966 Chr.16: 84872350-84872350 on Build GRCh38

View Full Product Details