Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89098501-89098993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSF3
Entrez Gene ID
197322
Gene Name
acyl-CoA synthetase family member 3
Gene Aliases
-
UniGene
Hs.461727
Gene Chromosome Location
Chr.16: 89093809-89160556 on Build GRCh38
Transcripts
NM_001127214.3 NM_001243279.2 NM_001284316.1 NM_174917.4 NR_104293.1 XM_005256293.2 XM_011522942.1 XM_011522943.1 XM_017023018.1 XM_017023019.1 XM_017023021.1

SNP Detail

SNP ID Chromosome Location
rs550756859 Chr.16: 89098847-89098847 on Build GRCh38
rs548417953 Chr.16: 89098754-89098754 on Build GRCh38
rs918052620 Chr.16: 89098680-89098680 on Build GRCh38
rs1000822118 Chr.16: 89098916-89098916 on Build GRCh38
rs552532864 Chr.16: 89098824-89098824 on Build GRCh38
rs775664048 Chr.16: 89098713-89098713 on Build GRCh38
rs770477265 Chr.16: 89098963-89098963 on Build GRCh38
rs941671761 Chr.16: 89098725-89098725 on Build GRCh38
rs993327777 Chr.16: 89098598-89098598 on Build GRCh38
rs1018294858 Chr.16: 89098635-89098635 on Build GRCh38
rs184589439 Chr.16: 89098608-89098608 on Build GRCh38
rs996050431 Chr.16: 89098602-89098602 on Build GRCh38
rs538740715 Chr.16: 89098535-89098535 on Build GRCh38
rs1020117919 Chr.16: 89098641-89098641 on Build GRCh38
rs575978814 Chr.16: 89098762-89098762 on Build GRCh38
rs748831431 Chr.16: 89098949-89098949 on Build GRCh38
rs529807641 Chr.16: 89098733-89098733 on Build GRCh38
rs931727000 Chr.16: 89098829-89098829 on Build GRCh38
rs758890096 Chr.16: 89098872-89098872 on Build GRCh38
rs747409821 Chr.16: 89098894-89098894 on Build GRCh38
rs983041055 Chr.16: 89098738-89098738 on Build GRCh38
rs917533898 Chr.16: 89098581-89098581 on Build GRCh38
rs768965227 Chr.16: 89098913-89098913 on Build GRCh38
rs144570053 Chr.16: 89098586-89098586 on Build GRCh38
rs749130601 Chr.16: 89098648-89098648 on Build GRCh38
rs573137492 Chr.16: 89098927-89098927 on Build GRCh38
rs148444044 Chr.16: 89098755-89098755 on Build GRCh38
rs935489018 Chr.16: 89098555-89098555 on Build GRCh38
rs1040179959 Chr.16: 89098768-89098768 on Build GRCh38
rs980898782 Chr.16: 89098691-89098691 on Build GRCh38
rs890292233 Chr.16: 89098862-89098862 on Build GRCh38
rs577925407 Chr.16: 89098597-89098597 on Build GRCh38
rs774075134 Chr.16: 89098544-89098545 on Build GRCh38
rs534214572 Chr.16: 89098943-89098943 on Build GRCh38
rs978870742 Chr.16: 89098550-89098550 on Build GRCh38
rs1024361215 Chr.16: 89098958-89098958 on Build GRCh38
rs571540361 Chr.16: 89098838-89098838 on Build GRCh38
rs968755021 Chr.16: 89098646-89098646 on Build GRCh38
rs757372814 Chr.16: 89098709-89098709 on Build GRCh38
rs759095896 Chr.16: 89098808-89098808 on Build GRCh38
rs554723752 Chr.16: 89098921-89098921 on Build GRCh38
rs996923846 Chr.16: 89098685-89098685 on Build GRCh38
rs1037607037 Chr.16: 89098596-89098596 on Build GRCh38
rs543967889 Chr.16: 89098666-89098666 on Build GRCh38
rs545395061 Chr.16: 89098618-89098618 on Build GRCh38
rs770653368 Chr.16: 89098814-89098814 on Build GRCh38
rs1041649431 Chr.16: 89098880-89098880 on Build GRCh38
rs181386231 Chr.16: 89098577-89098577 on Build GRCh38
rs180876717 Chr.16: 89098901-89098901 on Build GRCh38
rs189625890 Chr.16: 89098761-89098761 on Build GRCh38
rs774356361 Chr.16: 89098559-89098559 on Build GRCh38
rs964362215 Chr.16: 89098650-89098650 on Build GRCh38
rs1008687830 Chr.16: 89098617-89098617 on Build GRCh38
rs772194969 Chr.16: 89098710-89098710 on Build GRCh38
rs759049458 Chr.16: 89098893-89098893 on Build GRCh38
rs1010105244 Chr.16: 89098636-89098636 on Build GRCh38
rs959029403 Chr.16: 89098687-89098687 on Build GRCh38
rs988262022 Chr.16: 89098675-89098675 on Build GRCh38
rs558866583 Chr.16: 89098959-89098959 on Build GRCh38
rs773650209 Chr.16: 89098775-89098775 on Build GRCh38
rs778787971 Chr.16: 89098674-89098674 on Build GRCh38
rs899855894 Chr.16: 89098851-89098851 on Build GRCh38
rs539028189 Chr.16: 89098846-89098846 on Build GRCh38
rs921734547 Chr.16: 89098798-89098798 on Build GRCh38
rs776429283 Chr.16: 89098843-89098843 on Build GRCh38
rs576815618 Chr.16: 89098619-89098619 on Build GRCh38
rs1047469979 Chr.16: 89098583-89098583 on Build GRCh38
rs1008325440 Chr.16: 89098763-89098763 on Build GRCh38
rs890404124 Chr.16: 89098628-89098628 on Build GRCh38
rs759497869 Chr.16: 89098742-89098745 on Build GRCh38
rs36099289 Chr.16: 89098566-89098566 on Build GRCh38
rs1037470461 Chr.16: 89098600-89098600 on Build GRCh38
rs780980210 Chr.16: 89098589-89098589 on Build GRCh38
rs371292822 Chr.16: 89098601-89098601 on Build GRCh38
rs375918363 Chr.16: 89098541-89098541 on Build GRCh38
rs971289460 Chr.16: 89098964-89098964 on Build GRCh38
rs569349231 Chr.16: 89098854-89098854 on Build GRCh38
rs917420878 Chr.16: 89098727-89098727 on Build GRCh38
rs930204329 Chr.16: 89098582-89098582 on Build GRCh38
rs562604822 Chr.16: 89098699-89098699 on Build GRCh38
rs745951848 Chr.16: 89098684-89098684 on Build GRCh38
rs780521687 Chr.16: 89098892-89098892 on Build GRCh38
rs1032538435 Chr.16: 89098669-89098669 on Build GRCh38
rs575704599 Chr.16: 89098546-89098546 on Build GRCh38
rs1017207847 Chr.16: 89098972-89098972 on Build GRCh38

View Full Product Details