Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89100581-89101075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSF3
Entrez Gene ID
197322
Gene Name
acyl-CoA synthetase family member 3
Gene Aliases
-
UniGene
Hs.461727
Gene Chromosome Location
Chr.16: 89093809-89160556 on Build GRCh38
Transcripts
NM_001127214.3 NM_001243279.2 NM_001284316.1 NM_174917.4 NR_104293.1 XM_005256293.2 XM_011522942.1 XM_011522943.1 XM_017023018.1 XM_017023019.1 XM_017023020.1 XM_017023021.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3378025 c.189C>T p.H63H Chr.16: 89100870-89100870 on Build GRCh38
7234594 c.297C>T p.S99S Chr.16: 89100978-89100978 on Build GRCh38
308835 c.194G>A p.R65H Chr.16: 89100875-89100875 on Build GRCh38
4129507 c.342G>C p.A114A Chr.16: 89101023-89101023 on Build GRCh38
3513246 c.80G>A p.R27K Chr.16: 89100761-89100761 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs931176162 Chr.16: 89100756-89100756 on Build GRCh38
rs779775679 Chr.16: 89100796-89100796 on Build GRCh38
rs770572252 Chr.16: 89100873-89100873 on Build GRCh38
rs745684193 Chr.16: 89100833-89100839 on Build GRCh38
rs767348758 Chr.16: 89100815-89100815 on Build GRCh38
rs879113913 Chr.16: 89101024-89101024 on Build GRCh38
rs751461445 Chr.16: 89100747-89100747 on Build GRCh38
rs562260243 Chr.16: 89100870-89100870 on Build GRCh38
rs141490092 Chr.16: 89100666-89100666 on Build GRCh38
rs745852838 Chr.16: 89100642-89100642 on Build GRCh38
rs763738880 Chr.16: 89100637-89100637 on Build GRCh38
rs373476048 Chr.16: 89100932-89100932 on Build GRCh38
rs543359504 Chr.16: 89101050-89101051 on Build GRCh38
rs532903196 Chr.16: 89100949-89100949 on Build GRCh38
rs754161182 Chr.16: 89100767-89100767 on Build GRCh38
rs776314400 Chr.16: 89100697-89100697 on Build GRCh38
rs746564011 Chr.16: 89100837-89100837 on Build GRCh38
rs775152026 Chr.16: 89100659-89100659 on Build GRCh38
rs746400895 Chr.16: 89100731-89100731 on Build GRCh38
rs779450250 Chr.16: 89100786-89100786 on Build GRCh38
rs769716004 Chr.16: 89101001-89101001 on Build GRCh38
rs781244439 Chr.16: 89100997-89100997 on Build GRCh38
rs141518662 Chr.16: 89100826-89100826 on Build GRCh38
rs147915828 Chr.16: 89100810-89100810 on Build GRCh38
rs1028917212 Chr.16: 89100814-89100814 on Build GRCh38
rs981914227 Chr.16: 89100974-89100974 on Build GRCh38
rs778403709 Chr.16: 89100934-89100934 on Build GRCh38
rs11547019 Chr.16: 89100730-89100730 on Build GRCh38
rs768265862 Chr.16: 89101008-89101009 on Build GRCh38
rs774117419 Chr.16: 89100874-89100874 on Build GRCh38
rs767573178 Chr.16: 89100750-89100750 on Build GRCh38
rs375071176 Chr.16: 89100699-89100699 on Build GRCh38
rs370382601 Chr.16: 89100682-89100682 on Build GRCh38
rs750225037 Chr.16: 89100700-89100700 on Build GRCh38
rs6500527 Chr.16: 89101023-89101023 on Build GRCh38
rs761400921 Chr.16: 89100698-89100698 on Build GRCh38
rs753339356 Chr.16: 89100920-89100920 on Build GRCh38
rs746787960 Chr.16: 89100995-89100995 on Build GRCh38
rs760248047 Chr.16: 89100696-89100696 on Build GRCh38
rs764393364 Chr.16: 89100764-89100764 on Build GRCh38
rs1035298569 Chr.16: 89100752-89100752 on Build GRCh38
rs757051211 Chr.16: 89100663-89100663 on Build GRCh38
rs982369014 Chr.16: 89100610-89100610 on Build GRCh38
rs775060040 Chr.16: 89100687-89100687 on Build GRCh38
rs564576843 Chr.16: 89100805-89100805 on Build GRCh38
rs6500528 Chr.16: 89101035-89101035 on Build GRCh38
rs759448696 Chr.16: 89100811-89100811 on Build GRCh38
rs770106565 Chr.16: 89100651-89100651 on Build GRCh38
rs749994320 Chr.16: 89100993-89100993 on Build GRCh38
rs372534270 Chr.16: 89100638-89100638 on Build GRCh38
rs530503915 Chr.16: 89100961-89100961 on Build GRCh38
rs562298049 Chr.16: 89100628-89100628 on Build GRCh38
rs749751522 Chr.16: 89100685-89100685 on Build GRCh38
rs771946908 Chr.16: 89100881-89100881 on Build GRCh38
rs780402535 Chr.16: 89100860-89100860 on Build GRCh38
rs200703917 Chr.16: 89101039-89101039 on Build GRCh38
rs938384928 Chr.16: 89100626-89100626 on Build GRCh38
rs758439131 Chr.16: 89100643-89100643 on Build GRCh38
rs771182485 Chr.16: 89101006-89101006 on Build GRCh38
rs760759040 Chr.16: 89100986-89100986 on Build GRCh38
rs772372919 Chr.16: 89101054-89101054 on Build GRCh38
rs145819902 Chr.16: 89100641-89100641 on Build GRCh38
rs571472953 Chr.16: 89101007-89101007 on Build GRCh38
rs189821127 Chr.16: 89101009-89101009 on Build GRCh38
rs202182978 Chr.16: 89100709-89100709 on Build GRCh38
rs781163359 Chr.16: 89100713-89100713 on Build GRCh38
rs772073893 Chr.16: 89100927-89100927 on Build GRCh38
rs751551226 Chr.16: 89100710-89100710 on Build GRCh38
rs11273288 Chr.16: 89100667-89100668 on Build GRCh38
rs7193255 Chr.16: 89100996-89100996 on Build GRCh38
rs770158205 Chr.16: 89100677-89100677 on Build GRCh38
rs762053293 Chr.16: 89100899-89100899 on Build GRCh38
rs545886514 Chr.16: 89100803-89100803 on Build GRCh38
rs138156311 Chr.16: 89100919-89100919 on Build GRCh38
rs763583313 Chr.16: 89100656-89100656 on Build GRCh38
rs529560098 Chr.16: 89100892-89100892 on Build GRCh38
rs776862565 Chr.16: 89100898-89100898 on Build GRCh38
rs7188200 Chr.16: 89100686-89100686 on Build GRCh38
rs766109997 Chr.16: 89101034-89101034 on Build GRCh38
rs750707295 Chr.16: 89100902-89100902 on Build GRCh38
rs758856054 Chr.16: 89100853-89100853 on Build GRCh38
rs775551806 Chr.16: 89100816-89100816 on Build GRCh38
rs549664473 Chr.16: 89100791-89100791 on Build GRCh38
rs143605434 Chr.16: 89100959-89100959 on Build GRCh38
rs756279730 Chr.16: 89100761-89100761 on Build GRCh38
rs539069474 Chr.16: 89100664-89100664 on Build GRCh38
rs182147718 Chr.16: 89101038-89101038 on Build GRCh38
rs754782061 Chr.16: 89101037-89101037 on Build GRCh38
rs147044602 Chr.16: 89100852-89100852 on Build GRCh38
rs748533635 Chr.16: 89100650-89100650 on Build GRCh38
rs773740285 Chr.16: 89100652-89100652 on Build GRCh38
rs780132599 Chr.16: 89100644-89100644 on Build GRCh38
rs751637826 Chr.16: 89100670-89100670 on Build GRCh38
rs145969050 Chr.16: 89100994-89100994 on Build GRCh38
rs747180472 Chr.16: 89100645-89100645 on Build GRCh38
rs375374971 Chr.16: 89100724-89100724 on Build GRCh38
rs369499144 Chr.16: 89100827-89100827 on Build GRCh38
rs763197756 Chr.16: 89100906-89100906 on Build GRCh38
rs775259772 Chr.16: 89100889-89100889 on Build GRCh38
rs748128555 Chr.16: 89100998-89100998 on Build GRCh38
rs1040960753 Chr.16: 89100711-89100711 on Build GRCh38
rs753488764 Chr.16: 89100662-89100662 on Build GRCh38
rs769957763 Chr.16: 89100958-89100958 on Build GRCh38
rs761267255 Chr.16: 89101021-89101021 on Build GRCh38
rs753248595 Chr.16: 89100970-89100970 on Build GRCh38
rs59213357 Chr.16: 89100978-89100978 on Build GRCh38
rs749371364 Chr.16: 89101047-89101047 on Build GRCh38
rs572357895 Chr.16: 89100746-89100746 on Build GRCh38
rs373274928 Chr.16: 89100839-89100839 on Build GRCh38
rs57558972 Chr.16: 89100658-89100658 on Build GRCh38
rs757664305 Chr.16: 89100780-89100780 on Build GRCh38
rs745633046 Chr.16: 89100875-89100875 on Build GRCh38
rs187962814 Chr.16: 89100966-89100966 on Build GRCh38
rs148895267 Chr.16: 89100950-89100950 on Build GRCh38
rs778741714 Chr.16: 89100670-89100671 on Build GRCh38
rs764592417 Chr.16: 89101022-89101022 on Build GRCh38
rs748883934 Chr.16: 89100872-89100872 on Build GRCh38
rs747598756 Chr.16: 89100800-89100800 on Build GRCh38
rs752720474 Chr.16: 89100753-89100753 on Build GRCh38
rs746334624 Chr.16: 89100789-89100789 on Build GRCh38
rs901081235 Chr.16: 89100712-89100712 on Build GRCh38
rs7201122 Chr.16: 89100732-89100732 on Build GRCh38
rs756300619 Chr.16: 89100723-89100723 on Build GRCh38
rs767178224 Chr.16: 89100625-89100625 on Build GRCh38
rs6500526 Chr.16: 89100987-89100987 on Build GRCh38
rs775569136 Chr.16: 89100940-89100940 on Build GRCh38
rs746883692 Chr.16: 89100948-89100948 on Build GRCh38
rs781454948 Chr.16: 89100672-89100672 on Build GRCh38
rs779919537 Chr.16: 89100942-89100942 on Build GRCh38
rs144711526 Chr.16: 89100797-89100797 on Build GRCh38
rs755491777 Chr.16: 89100917-89100917 on Build GRCh38
rs762719449 Chr.16: 89100740-89100740 on Build GRCh38
rs777642279 Chr.16: 89101044-89101044 on Build GRCh38
rs757905943 Chr.16: 89101029-89101029 on Build GRCh38
rs766802453 Chr.16: 89100908-89100908 on Build GRCh38
rs756085657 Chr.16: 89101042-89101042 on Build GRCh38
rs202121474 Chr.16: 89100864-89100864 on Build GRCh38
rs537218566 Chr.16: 89100689-89100689 on Build GRCh38
rs201113468 Chr.16: 89100951-89100951 on Build GRCh38
rs755852664 Chr.16: 89100629-89100629 on Build GRCh38
rs751929678 Chr.16: 89100964-89100964 on Build GRCh38
rs115469156 Chr.16: 89100667-89100667 on Build GRCh38
rs534594073 Chr.16: 89100734-89100734 on Build GRCh38
rs552894451 Chr.16: 89100737-89100737 on Build GRCh38
rs771110203 Chr.16: 89100669-89100670 on Build GRCh38
rs901883302 Chr.16: 89100804-89100804 on Build GRCh38
rs771723461 Chr.16: 89100940-89100940 on Build GRCh38
rs776789917 Chr.16: 89100648-89100648 on Build GRCh38
rs6500529 Chr.16: 89101050-89101050 on Build GRCh38
rs999563327 Chr.16: 89100719-89100719 on Build GRCh38
rs765633249 Chr.16: 89100840-89100840 on Build GRCh38
rs771481057 Chr.16: 89100726-89100726 on Build GRCh38
rs763021932 Chr.16: 89101005-89101005 on Build GRCh38
rs755071079 Chr.16: 89100671-89100671 on Build GRCh38
rs67522353 Chr.16: 89100668-89100669 on Build GRCh38
rs551417698 Chr.16: 89100669-89100669 on Build GRCh38
rs1041386545 Chr.16: 89100624-89100624 on Build GRCh38
rs200536797 Chr.16: 89100871-89100871 on Build GRCh38
rs541228443 Chr.16: 89100611-89100611 on Build GRCh38
rs922581606 Chr.16: 89100841-89100841 on Build GRCh38
rs34972688 Chr.16: 89100801-89100801 on Build GRCh38
rs752913078 Chr.16: 89100718-89100718 on Build GRCh38
rs768930982 Chr.16: 89100647-89100647 on Build GRCh38
rs141517318 Chr.16: 89101008-89101008 on Build GRCh38
rs749661738 Chr.16: 89100725-89100725 on Build GRCh38
rs767399752 Chr.16: 89100664-89100665 on Build GRCh38
rs772907663 Chr.16: 89100739-89100739 on Build GRCh38
rs994160606 Chr.16: 89100952-89100952 on Build GRCh38
rs901932797 Chr.16: 89101002-89101002 on Build GRCh38
rs766597613 Chr.16: 89100962-89100962 on Build GRCh38
rs746501634 Chr.16: 89100911-89100911 on Build GRCh38
rs763794563 Chr.16: 89100660-89100660 on Build GRCh38
rs142006073 Chr.16: 89101049-89101050 on Build GRCh38
rs775104550 Chr.16: 89100666-89100667 on Build GRCh38
rs560101290 Chr.16: 89100939-89100939 on Build GRCh38

View Full Product Details