Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89267253-89267793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD11
Entrez Gene ID
29123
Gene Name
ankyrin repeat domain 11
Gene Aliases
ANCO-1, ANCO1, LZ16, T13
UniGene
Hs.335003
Gene Chromosome Location
Chr.16: 89267619-89490561 on Build GRCh38
Transcripts
NM_001256182.1 NM_001256183.1 NM_013275.5 XM_006721181.1 XM_006721184.2 XM_011523051.2 XM_011523053.2 XM_011523054.2 XM_011523055.2 XM_011523056.2 XM_017023182.1 XM_017023183.1 XM_017023184.1 XM_017023185.1 XM_017023186.1 XM_017023187.1

SNP Detail

SNP ID Chromosome Location
rs554067405 Chr.16: 89267632-89267635 on Build GRCh38
rs28404961 Chr.16: 89267459-89267459 on Build GRCh38
rs925841756 Chr.16: 89267282-89267282 on Build GRCh38
rs550297860 Chr.16: 89267423-89267423 on Build GRCh38
rs558548887 Chr.16: 89267441-89267441 on Build GRCh38
rs998485069 Chr.16: 89267543-89267543 on Build GRCh38
rs977142308 Chr.16: 89267574-89267574 on Build GRCh38
rs977033371 Chr.16: 89267405-89267405 on Build GRCh38
rs894558372 Chr.16: 89267320-89267320 on Build GRCh38
rs906936772 Chr.16: 89267302-89267302 on Build GRCh38
rs117652610 Chr.16: 89267429-89267429 on Build GRCh38
rs751136608 Chr.16: 89267639-89267642 on Build GRCh38
rs186604464 Chr.16: 89267344-89267344 on Build GRCh38
rs9652637 Chr.16: 89267696-89267696 on Build GRCh38
rs1018634237 Chr.16: 89267387-89267387 on Build GRCh38
rs545743434 Chr.16: 89267626-89267626 on Build GRCh38
rs948241796 Chr.16: 89267638-89267639 on Build GRCh38
rs540061542 Chr.16: 89267383-89267383 on Build GRCh38
rs1055592887 Chr.16: 89267315-89267315 on Build GRCh38
rs1018667106 Chr.16: 89267521-89267521 on Build GRCh38
rs1005284569 Chr.16: 89267347-89267347 on Build GRCh38
rs908632575 Chr.16: 89267613-89267613 on Build GRCh38
rs989790098 Chr.16: 89267282-89267283 on Build GRCh38
rs908599609 Chr.16: 89267432-89267432 on Build GRCh38
rs966510095 Chr.16: 89267360-89267360 on Build GRCh38
rs915787032 Chr.16: 89267419-89267419 on Build GRCh38
rs536892343 Chr.16: 89267404-89267404 on Build GRCh38
rs997143901 Chr.16: 89267488-89267488 on Build GRCh38
rs1038458280 Chr.16: 89267625-89267625 on Build GRCh38
rs145479059 Chr.16: 89267291-89267294 on Build GRCh38
rs997428295 Chr.16: 89267374-89267374 on Build GRCh38
rs898728343 Chr.16: 89267363-89267363 on Build GRCh38
rs915802120 Chr.16: 89267575-89267575 on Build GRCh38
rs1008781116 Chr.16: 89267497-89267497 on Build GRCh38
rs981249849 Chr.16: 89267612-89267612 on Build GRCh38
rs1012475708 Chr.16: 89267334-89267334 on Build GRCh38
rs1004910286 Chr.16: 89267522-89267522 on Build GRCh38
rs940912986 Chr.16: 89267642-89267642 on Build GRCh38
rs1049677721 Chr.16: 89267638-89267638 on Build GRCh38
rs759624724 Chr.16: 89267427-89267427 on Build GRCh38
rs940055474 Chr.16: 89267618-89267618 on Build GRCh38
rs191560896 Chr.16: 89267536-89267536 on Build GRCh38
rs564784737 Chr.16: 89267406-89267406 on Build GRCh38
rs565289288 Chr.16: 89267651-89267651 on Build GRCh38
rs991447031 Chr.16: 89267561-89267561 on Build GRCh38
rs938417375 Chr.16: 89267453-89267454 on Build GRCh38
rs935854407 Chr.16: 89267296-89267296 on Build GRCh38
rs1036385658 Chr.16: 89267659-89267659 on Build GRCh38
rs957257997 Chr.16: 89267560-89267560 on Build GRCh38
rs1046230752 Chr.16: 89267332-89267332 on Build GRCh38
rs1011355865 Chr.16: 89267556-89267556 on Build GRCh38
rs901803808 Chr.16: 89267674-89267674 on Build GRCh38
rs1028459496 Chr.16: 89267380-89267380 on Build GRCh38
rs931888809 Chr.16: 89267280-89267282 on Build GRCh38
rs953180084 Chr.16: 89267549-89267551 on Build GRCh38
rs4785558 Chr.16: 89267578-89267578 on Build GRCh38
rs939574760 Chr.16: 89267321-89267321 on Build GRCh38
rs866046925 Chr.16: 89267333-89267333 on Build GRCh38
rs901942310 Chr.16: 89267530-89267530 on Build GRCh38
rs559900679 Chr.16: 89267548-89267548 on Build GRCh38
rs1038646213 Chr.16: 89267362-89267362 on Build GRCh38
rs957275677 Chr.16: 89267408-89267408 on Build GRCh38
rs868090329 Chr.16: 89267384-89267384 on Build GRCh38
rs1027660120 Chr.16: 89267340-89267340 on Build GRCh38
rs555817968 Chr.16: 89267295-89267296 on Build GRCh38
rs928426490 Chr.16: 89267442-89267442 on Build GRCh38
rs868402459 Chr.16: 89267475-89267475 on Build GRCh38
rs182900009 Chr.16: 89267547-89267547 on Build GRCh38
rs4785650 Chr.16: 89267409-89267409 on Build GRCh38
rs55681374 Chr.16: 89267752-89267752 on Build GRCh38
rs950195345 Chr.16: 89267329-89267329 on Build GRCh38
rs989984866 Chr.16: 89267389-89267389 on Build GRCh38
rs760895886 Chr.16: 89267682-89267682 on Build GRCh38
rs997562240 Chr.16: 89267710-89267710 on Build GRCh38
rs1038342192 Chr.16: 89267460-89267460 on Build GRCh38
rs567192174 Chr.16: 89267665-89267665 on Build GRCh38
rs146990114 Chr.16: 89267356-89267356 on Build GRCh38
rs891321416 Chr.16: 89267492-89267492 on Build GRCh38
rs746197372 Chr.16: 89267592-89267593 on Build GRCh38
rs530397883 Chr.16: 89267489-89267489 on Build GRCh38
rs555939564 Chr.16: 89267494-89267494 on Build GRCh38
rs201829721 Chr.16: 89267413-89267414 on Build GRCh38

View Full Product Details