Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89268574-89269083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD11
Entrez Gene ID
29123
Gene Name
ankyrin repeat domain 11
Gene Aliases
ANCO-1, ANCO1, LZ16, T13
UniGene
Hs.335003
Gene Chromosome Location
Chr.16: 89267619-89490561 on Build GRCh38
Transcripts
NM_001256182.1 NM_001256183.1 NM_013275.5 XM_006721181.1 XM_006721184.2 XM_011523051.2 XM_011523053.2 XM_011523054.2 XM_011523055.2 XM_011523056.2 XM_011523057.2 XM_017023182.1 XM_017023183.1 XM_017023184.1 XM_017023185.1 XM_017023186.1 XM_017023187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
255829 c.7823G>A p.R2608Q Chr.16: 89268647-89268647 on Build GRCh38
6977749 c.7807-1G>C p.? Chr.16: 89268664-89268664 on Build GRCh38
1380497 c.7840G>A p.A2614T Chr.16: 89268630-89268630 on Build GRCh38
5963249 c.7834G>A p.E2612K Chr.16: 89268636-89268636 on Build GRCh38
4508706 c.7853C>T p.A2618V Chr.16: 89268617-89268617 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs997898675 Chr.16: 89268943-89268943 on Build GRCh38
rs1042499301 Chr.16: 89268719-89268719 on Build GRCh38
rs1032705191 Chr.16: 89268752-89268752 on Build GRCh38
rs114121452 Chr.16: 89269003-89269003 on Build GRCh38
rs896778396 Chr.16: 89268939-89268939 on Build GRCh38
rs979185942 Chr.16: 89268851-89268851 on Build GRCh38
rs552734531 Chr.16: 89268781-89268781 on Build GRCh38
rs895613597 Chr.16: 89268685-89268687 on Build GRCh38
rs186947934 Chr.16: 89268751-89268751 on Build GRCh38
rs753365602 Chr.16: 89268691-89268693 on Build GRCh38
rs757833973 Chr.16: 89268911-89268911 on Build GRCh38
rs553403498 Chr.16: 89268879-89268879 on Build GRCh38
rs377589207 Chr.16: 89268724-89268724 on Build GRCh38
rs1032098943 Chr.16: 89269018-89269018 on Build GRCh38
rs530241255 Chr.16: 89268712-89268712 on Build GRCh38
rs746391051 Chr.16: 89268984-89268984 on Build GRCh38
rs773162819 Chr.16: 89268622-89268622 on Build GRCh38
rs761051391 Chr.16: 89268679-89268679 on Build GRCh38
rs16965520 Chr.16: 89269020-89269020 on Build GRCh38
rs749885956 Chr.16: 89268685-89268685 on Build GRCh38
rs763570873 Chr.16: 89268683-89268684 on Build GRCh38
rs1036686419 Chr.16: 89268740-89268740 on Build GRCh38
rs143733186 Chr.16: 89268873-89268873 on Build GRCh38
rs969395888 Chr.16: 89268995-89268995 on Build GRCh38
rs1037122196 Chr.16: 89268900-89268900 on Build GRCh38
rs892854408 Chr.16: 89268941-89268941 on Build GRCh38
rs143287644 Chr.16: 89268631-89268631 on Build GRCh38
rs549395265 Chr.16: 89268853-89268853 on Build GRCh38
rs532018283 Chr.16: 89268827-89268827 on Build GRCh38
rs948628508 Chr.16: 89268698-89268698 on Build GRCh38
rs756083090 Chr.16: 89268671-89268672 on Build GRCh38
rs941258153 Chr.16: 89268894-89268894 on Build GRCh38
rs370800421 Chr.16: 89268680-89268680 on Build GRCh38
rs774722465 Chr.16: 89268655-89268655 on Build GRCh38
rs1015503029 Chr.16: 89269019-89269019 on Build GRCh38
rs797044900 Chr.16: 89268645-89268645 on Build GRCh38
rs1017744432 Chr.16: 89268983-89268983 on Build GRCh38
rs970115960 Chr.16: 89269040-89269040 on Build GRCh38
rs757939041 Chr.16: 89268700-89268700 on Build GRCh38
rs1012968593 Chr.16: 89268761-89268761 on Build GRCh38
rs1015471920 Chr.16: 89268767-89268767 on Build GRCh38
rs560680331 Chr.16: 89268760-89268760 on Build GRCh38
rs1044369839 Chr.16: 89268717-89268717 on Build GRCh38
rs565432078 Chr.16: 89268932-89268932 on Build GRCh38
rs968985287 Chr.16: 89268843-89268843 on Build GRCh38
rs891346399 Chr.16: 89268707-89268707 on Build GRCh38
rs759943360 Chr.16: 89268661-89268661 on Build GRCh38
rs59204820 Chr.16: 89268967-89268967 on Build GRCh38
rs74033705 Chr.16: 89268804-89268804 on Build GRCh38
rs981282734 Chr.16: 89268766-89268766 on Build GRCh38
rs1022900336 Chr.16: 89268764-89268764 on Build GRCh38
rs1012587161 Chr.16: 89268953-89268953 on Build GRCh38
rs527837033 Chr.16: 89268770-89268770 on Build GRCh38
rs1050834832 Chr.16: 89268946-89268946 on Build GRCh38
rs944222329 Chr.16: 89268709-89268709 on Build GRCh38
rs902680755 Chr.16: 89268734-89268734 on Build GRCh38
rs912822054 Chr.16: 89268833-89268833 on Build GRCh38
rs56011166 Chr.16: 89268627-89268627 on Build GRCh38
rs567682785 Chr.16: 89268857-89268857 on Build GRCh38
rs868039749 Chr.16: 89268653-89268653 on Build GRCh38
rs1007679539 Chr.16: 89268820-89268822 on Build GRCh38
rs985822981 Chr.16: 89268828-89268828 on Build GRCh38
rs917333144 Chr.16: 89268871-89268871 on Build GRCh38
rs888365349 Chr.16: 89268722-89268722 on Build GRCh38
rs191465180 Chr.16: 89269035-89269035 on Build GRCh38
rs1027070744 Chr.16: 89269028-89269028 on Build GRCh38
rs902711863 Chr.16: 89268870-89268870 on Build GRCh38
rs377601454 Chr.16: 89268628-89268628 on Build GRCh38
rs909816893 Chr.16: 89268887-89268887 on Build GRCh38
rs562845696 Chr.16: 89268667-89268667 on Build GRCh38
rs971683946 Chr.16: 89268765-89268765 on Build GRCh38
rs141842419 Chr.16: 89269021-89269021 on Build GRCh38
rs1005651026 Chr.16: 89268726-89268726 on Build GRCh38
rs148332563 Chr.16: 89268637-89268637 on Build GRCh38
rs557917714 Chr.16: 89268975-89268975 on Build GRCh38
rs761282274 Chr.16: 89269017-89269018 on Build GRCh38
rs776041550 Chr.16: 89268668-89268668 on Build GRCh38

View Full Product Details