Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89274722-89275222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD11
Entrez Gene ID
29123
Gene Name
ankyrin repeat domain 11
Gene Aliases
ANCO-1, ANCO1, LZ16, T13
UniGene
Hs.335003
Gene Chromosome Location
Chr.16: 89267619-89490561 on Build GRCh38
Transcripts
NM_001256182.1 NM_001256183.1 NM_013275.5 XM_006721181.1 XM_006721184.2 XM_011523051.2 XM_011523053.2 XM_011523054.2 XM_011523055.2 XM_011523056.2 XM_011523057.2 XM_017023182.1 XM_017023183.1 XM_017023184.1 XM_017023185.1 XM_017023186.1 XM_017023187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
259430 c.7599G>T p.E2533D Chr.16: 89274928-89274928 on Build GRCh38
6969477 c.7699C>T p.P2567S Chr.16: 89274828-89274828 on Build GRCh38
6600316 c.7622C>T p.A2541V Chr.16: 89274905-89274905 on Build GRCh38
7303009 c.7531G>A p.V2511I Chr.16: 89275131-89275131 on Build GRCh38
6927999 c.7591G>C p.E2531Q Chr.16: 89274936-89274936 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754093852 Chr.16: 89275164-89275164 on Build GRCh38
rs946312469 Chr.16: 89275191-89275191 on Build GRCh38
rs765196702 Chr.16: 89274814-89274814 on Build GRCh38
rs143429968 Chr.16: 89274850-89274850 on Build GRCh38
rs769512532 Chr.16: 89275116-89275116 on Build GRCh38
rs147434505 Chr.16: 89274845-89274845 on Build GRCh38
rs757570717 Chr.16: 89275065-89275065 on Build GRCh38
rs753798424 Chr.16: 89275174-89275174 on Build GRCh38
rs760761901 Chr.16: 89275150-89275150 on Build GRCh38
rs890731174 Chr.16: 89275027-89275027 on Build GRCh38
rs375665514 Chr.16: 89274781-89274781 on Build GRCh38
rs368616338 Chr.16: 89275060-89275060 on Build GRCh38
rs182655301 Chr.16: 89274759-89274759 on Build GRCh38
rs753555053 Chr.16: 89275153-89275153 on Build GRCh38
rs1047816097 Chr.16: 89274754-89274754 on Build GRCh38
rs190789698 Chr.16: 89274904-89274904 on Build GRCh38
rs907721683 Chr.16: 89275126-89275126 on Build GRCh38
rs777571791 Chr.16: 89275131-89275131 on Build GRCh38
rs773189154 Chr.16: 89274905-89274905 on Build GRCh38
rs764362118 Chr.16: 89275061-89275061 on Build GRCh38
rs751657071 Chr.16: 89274838-89274838 on Build GRCh38
rs763557063 Chr.16: 89274999-89274999 on Build GRCh38
rs182946384 Chr.16: 89275021-89275021 on Build GRCh38
rs954918239 Chr.16: 89274785-89274785 on Build GRCh38
rs893851146 Chr.16: 89274755-89274755 on Build GRCh38
rs747782392 Chr.16: 89274998-89274998 on Build GRCh38
rs371515678 Chr.16: 89274986-89274986 on Build GRCh38
rs766284510 Chr.16: 89274919-89274919 on Build GRCh38
rs12709112 Chr.16: 89274760-89274760 on Build GRCh38
rs369656239 Chr.16: 89275003-89275003 on Build GRCh38
rs983273950 Chr.16: 89274928-89274928 on Build GRCh38
rs1052222126 Chr.16: 89275169-89275169 on Build GRCh38
rs745569506 Chr.16: 89274978-89274979 on Build GRCh38
rs751599226 Chr.16: 89274946-89274946 on Build GRCh38
rs200826191 Chr.16: 89275168-89275168 on Build GRCh38
rs761658762 Chr.16: 89274895-89274895 on Build GRCh38
rs758994547 Chr.16: 89275079-89275079 on Build GRCh38
rs549163436 Chr.16: 89274795-89274795 on Build GRCh38
rs770230623 Chr.16: 89274767-89274767 on Build GRCh38
rs188800705 Chr.16: 89274776-89274776 on Build GRCh38
rs755368384 Chr.16: 89275046-89275047 on Build GRCh38
rs559756815 Chr.16: 89274868-89274868 on Build GRCh38
rs548065999 Chr.16: 89274765-89274765 on Build GRCh38
rs1027527236 Chr.16: 89274786-89274786 on Build GRCh38
rs886041889 Chr.16: 89275110-89275110 on Build GRCh38
rs770834630 Chr.16: 89275005-89275005 on Build GRCh38
rs199966235 Chr.16: 89274894-89274894 on Build GRCh38
rs749777331 Chr.16: 89274859-89274859 on Build GRCh38
rs780945391 Chr.16: 89274996-89274996 on Build GRCh38
rs747727451 Chr.16: 89275114-89275114 on Build GRCh38
rs368773165 Chr.16: 89274934-89274934 on Build GRCh38
rs759996108 Chr.16: 89275053-89275053 on Build GRCh38
rs372665809 Chr.16: 89275132-89275132 on Build GRCh38
rs371943158 Chr.16: 89275189-89275189 on Build GRCh38
rs774916088 Chr.16: 89274782-89274782 on Build GRCh38
rs377763770 Chr.16: 89274985-89274985 on Build GRCh38
rs754147309 Chr.16: 89275064-89275064 on Build GRCh38
rs147131178 Chr.16: 89274994-89274994 on Build GRCh38
rs767469593 Chr.16: 89275055-89275055 on Build GRCh38
rs142845301 Chr.16: 89274832-89274832 on Build GRCh38
rs775056744 Chr.16: 89274889-89274889 on Build GRCh38
rs375954198 Chr.16: 89275044-89275044 on Build GRCh38
rs372534900 Chr.16: 89275041-89275041 on Build GRCh38
rs1037708608 Chr.16: 89274771-89274771 on Build GRCh38
rs371485455 Chr.16: 89275002-89275002 on Build GRCh38
rs778645389 Chr.16: 89274962-89274964 on Build GRCh38
rs375761059 Chr.16: 89274992-89274992 on Build GRCh38
rs767736770 Chr.16: 89274955-89274955 on Build GRCh38
rs764237472 Chr.16: 89275152-89275152 on Build GRCh38
rs139657234 Chr.16: 89274865-89274865 on Build GRCh38
rs544309810 Chr.16: 89275134-89275134 on Build GRCh38
rs990753494 Chr.16: 89274886-89274886 on Build GRCh38
rs1012018771 Chr.16: 89275008-89275008 on Build GRCh38
rs143679902 Chr.16: 89274949-89274949 on Build GRCh38
rs750415803 Chr.16: 89274817-89274817 on Build GRCh38
rs568821799 Chr.16: 89275045-89275045 on Build GRCh38
rs779438523 Chr.16: 89275070-89275070 on Build GRCh38
rs772110406 Chr.16: 89275144-89275144 on Build GRCh38
rs775530757 Chr.16: 89275145-89275145 on Build GRCh38
rs924000018 Chr.16: 89275195-89275195 on Build GRCh38
rs1010790884 Chr.16: 89274787-89274787 on Build GRCh38
rs201416489 Chr.16: 89274804-89274804 on Build GRCh38
rs863223319 Chr.16: 89274958-89274958 on Build GRCh38
rs758502813 Chr.16: 89274825-89274825 on Build GRCh38
rs750841875 Chr.16: 89275177-89275177 on Build GRCh38
rs367974049 Chr.16: 89275197-89275197 on Build GRCh38
rs572878194 Chr.16: 89275143-89275143 on Build GRCh38
rs931953835 Chr.16: 89275000-89275000 on Build GRCh38
rs371135572 Chr.16: 89274841-89274841 on Build GRCh38
rs797044890 Chr.16: 89275092-89275092 on Build GRCh38
rs752186603 Chr.16: 89275200-89275200 on Build GRCh38
rs983326501 Chr.16: 89275074-89275074 on Build GRCh38
rs779305495 Chr.16: 89274990-89274990 on Build GRCh38
rs767743130 Chr.16: 89275071-89275071 on Build GRCh38
rs569975517 Chr.16: 89275173-89275173 on Build GRCh38
rs887675042 Chr.16: 89275033-89275033 on Build GRCh38
rs371586812 Chr.16: 89275073-89275073 on Build GRCh38
rs373574984 Chr.16: 89275043-89275043 on Build GRCh38
rs757773624 Chr.16: 89274989-89274989 on Build GRCh38
rs762952786 Chr.16: 89274910-89274910 on Build GRCh38
rs541250965 Chr.16: 89274824-89274824 on Build GRCh38
rs756374514 Chr.16: 89274967-89274967 on Build GRCh38
rs143942711 Chr.16: 89275124-89275124 on Build GRCh38
rs781616497 Chr.16: 89274763-89274763 on Build GRCh38
rs781285799 Chr.16: 89274844-89274844 on Build GRCh38
rs752811231 Chr.16: 89275059-89275059 on Build GRCh38
rs748520500 Chr.16: 89274766-89274766 on Build GRCh38
rs748645496 Chr.16: 89275069-89275070 on Build GRCh38
rs373811394 Chr.16: 89274976-89274976 on Build GRCh38
rs558439504 Chr.16: 89274879-89274879 on Build GRCh38
rs781453809 Chr.16: 89275065-89275066 on Build GRCh38
rs778206999 Chr.16: 89274853-89274853 on Build GRCh38
rs200599560 Chr.16: 89274806-89274806 on Build GRCh38
rs374764021 Chr.16: 89274892-89274892 on Build GRCh38
rs28603039 Chr.16: 89274913-89274913 on Build GRCh38
rs745344760 Chr.16: 89274780-89274780 on Build GRCh38
rs780818897 Chr.16: 89275080-89275080 on Build GRCh38
rs758827352 Chr.16: 89275186-89275186 on Build GRCh38
rs575149376 Chr.16: 89275040-89275040 on Build GRCh38
rs752936167 Chr.16: 89274963-89274963 on Build GRCh38
rs770056407 Chr.16: 89274925-89274925 on Build GRCh38
rs751892768 Chr.16: 89275022-89275022 on Build GRCh38
rs776947170 Chr.16: 89274805-89274805 on Build GRCh38

View Full Product Details