Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89279589-89280088 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD11
Entrez Gene ID
29123
Gene Name
ankyrin repeat domain 11
Gene Aliases
ANCO-1, ANCO1, LZ16, T13
UniGene
Hs.335003
Gene Chromosome Location
Chr.16: 89267619-89490561 on Build GRCh38
Transcripts
NM_001256182.1 NM_001256183.1 NM_013275.5 XM_006721181.1 XM_006721184.2 XM_011523051.2 XM_011523053.2 XM_011523054.2 XM_011523055.2 XM_011523056.2 XM_011523057.2 XM_017023182.1 XM_017023183.1 XM_017023184.1 XM_017023185.1 XM_017023186.1 XM_017023187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6600315 c.6514G>A p.G2172R Chr.16: 89280028-89280028 on Build GRCh38
6977975 c.6788C>G p.P2263R Chr.16: 89279754-89279754 on Build GRCh38
6953192 c.6883G>A p.E2295K Chr.16: 89279659-89279659 on Build GRCh38
1324227 c.6787C>T p.P2263S Chr.16: 89279755-89279755 on Build GRCh38
1563442 c.6535G>A p.V2179I Chr.16: 89280007-89280007 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746379317 Chr.16: 89279929-89279929 on Build GRCh38
rs888918193 Chr.16: 89279610-89279610 on Build GRCh38
rs767106606 Chr.16: 89279725-89279725 on Build GRCh38
rs763538904 Chr.16: 89279783-89279783 on Build GRCh38
rs759279703 Chr.16: 89280008-89280008 on Build GRCh38
rs993671847 Chr.16: 89279933-89279933 on Build GRCh38
rs761444335 Chr.16: 89279861-89279861 on Build GRCh38
rs954622305 Chr.16: 89279670-89279670 on Build GRCh38
rs769271809 Chr.16: 89279993-89279993 on Build GRCh38
rs748372503 Chr.16: 89279828-89279828 on Build GRCh38
rs200852197 Chr.16: 89279812-89279812 on Build GRCh38
rs918154229 Chr.16: 89279616-89279616 on Build GRCh38
rs907454686 Chr.16: 89279770-89279770 on Build GRCh38
rs1013086722 Chr.16: 89279664-89279664 on Build GRCh38
rs532695477 Chr.16: 89279752-89279752 on Build GRCh38
rs771395581 Chr.16: 89279922-89279922 on Build GRCh38
rs772878608 Chr.16: 89279944-89279944 on Build GRCh38
rs371524118 Chr.16: 89279940-89279940 on Build GRCh38
rs1045721935 Chr.16: 89279733-89279733 on Build GRCh38
rs924433707 Chr.16: 89279827-89279827 on Build GRCh38
rs866708710 Chr.16: 89280004-89280004 on Build GRCh38
rs775584042 Chr.16: 89279683-89279683 on Build GRCh38
rs772115194 Chr.16: 89279680-89279680 on Build GRCh38
rs772567539 Chr.16: 89279981-89279981 on Build GRCh38
rs747612507 Chr.16: 89279988-89279988 on Build GRCh38
rs1014039214 Chr.16: 89279931-89279931 on Build GRCh38
rs778171762 Chr.16: 89279920-89279920 on Build GRCh38
rs1003478010 Chr.16: 89279782-89279782 on Build GRCh38
rs1054443690 Chr.16: 89279833-89279833 on Build GRCh38
rs753352190 Chr.16: 89279855-89279855 on Build GRCh38
rs747483763 Chr.16: 89280038-89280038 on Build GRCh38
rs993290174 Chr.16: 89279662-89279662 on Build GRCh38
rs754931132 Chr.16: 89279951-89279951 on Build GRCh38
rs764436784 Chr.16: 89279714-89279714 on Build GRCh38
rs999284780 Chr.16: 89279646-89279646 on Build GRCh38
rs539129565 Chr.16: 89279921-89279921 on Build GRCh38
rs1023089135 Chr.16: 89279666-89279666 on Build GRCh38
rs537642535 Chr.16: 89279867-89279867 on Build GRCh38
rs536681113 Chr.16: 89279909-89279909 on Build GRCh38
rs781179335 Chr.16: 89279824-89279824 on Build GRCh38
rs904421745 Chr.16: 89279893-89279893 on Build GRCh38
rs768698196 Chr.16: 89279756-89279756 on Build GRCh38
rs201957371 Chr.16: 89279716-89279716 on Build GRCh38
rs943160633 Chr.16: 89279853-89279853 on Build GRCh38
rs146575027 Chr.16: 89279618-89279618 on Build GRCh38
rs769218762 Chr.16: 89280039-89280039 on Build GRCh38
rs774615667 Chr.16: 89279924-89279924 on Build GRCh38
rs753501960 Chr.16: 89279803-89279803 on Build GRCh38
rs760673104 Chr.16: 89280054-89280054 on Build GRCh38
rs752780335 Chr.16: 89279954-89279954 on Build GRCh38
rs184871095 Chr.16: 89279887-89279887 on Build GRCh38
rs918068073 Chr.16: 89279723-89279723 on Build GRCh38
rs1007876265 Chr.16: 89279672-89279672 on Build GRCh38
rs776150975 Chr.16: 89279935-89279935 on Build GRCh38
rs367564163 Chr.16: 89279900-89279900 on Build GRCh38
rs764885268 Chr.16: 89279862-89279862 on Build GRCh38
rs903253722 Chr.16: 89279643-89279643 on Build GRCh38
rs561404796 Chr.16: 89279691-89279691 on Build GRCh38
rs752311335 Chr.16: 89279726-89279726 on Build GRCh38
rs778726288 Chr.16: 89279750-89279750 on Build GRCh38
rs149834477 Chr.16: 89280046-89280046 on Build GRCh38
rs570265865 Chr.16: 89279844-89279844 on Build GRCh38
rs1044358936 Chr.16: 89279852-89279852 on Build GRCh38
rs897670799 Chr.16: 89279644-89279644 on Build GRCh38
rs377447921 Chr.16: 89279678-89279678 on Build GRCh38
rs776944723 Chr.16: 89279757-89279757 on Build GRCh38
rs1038929796 Chr.16: 89279879-89279879 on Build GRCh38
rs201020132 Chr.16: 89280053-89280053 on Build GRCh38
rs565435346 Chr.16: 89279751-89279751 on Build GRCh38
rs770209899 Chr.16: 89279830-89279830 on Build GRCh38
rs766007820 Chr.16: 89279999-89279999 on Build GRCh38
rs910495344 Chr.16: 89279821-89279821 on Build GRCh38
rs897282716 Chr.16: 89279923-89279923 on Build GRCh38
rs148730499 Chr.16: 89279720-89279720 on Build GRCh38
rs1000057912 Chr.16: 89279897-89279897 on Build GRCh38
rs749486628 Chr.16: 89279970-89279970 on Build GRCh38
rs770262960 Chr.16: 89279761-89279761 on Build GRCh38
rs1003120636 Chr.16: 89279873-89279873 on Build GRCh38
rs767358927 Chr.16: 89279952-89279952 on Build GRCh38
rs748996527 Chr.16: 89279745-89279745 on Build GRCh38
rs966362122 Chr.16: 89279984-89279984 on Build GRCh38
rs750062555 Chr.16: 89279870-89279875 on Build GRCh38
rs757094468 Chr.16: 89279748-89279748 on Build GRCh38
rs752658811 Chr.16: 89280013-89280013 on Build GRCh38
rs754914414 Chr.16: 89279905-89279905 on Build GRCh38
rs893421448 Chr.16: 89279656-89279656 on Build GRCh38
rs767004467 Chr.16: 89279789-89279789 on Build GRCh38
rs1015762202 Chr.16: 89279790-89279790 on Build GRCh38
rs199952368 Chr.16: 89279963-89279963 on Build GRCh38
rs74033733 Chr.16: 89280045-89280045 on Build GRCh38
rs1033011026 Chr.16: 89279655-89279655 on Build GRCh38
rs975984811 Chr.16: 89280005-89280005 on Build GRCh38
rs978514758 Chr.16: 89279690-89279690 on Build GRCh38
rs781330205 Chr.16: 89279907-89279907 on Build GRCh38
rs372632879 Chr.16: 89279817-89279817 on Build GRCh38
rs777253893 Chr.16: 89280042-89280042 on Build GRCh38
rs202216051 Chr.16: 89279982-89279982 on Build GRCh38
rs1040188778 Chr.16: 89279746-89279746 on Build GRCh38
rs758276955 Chr.16: 89279813-89279813 on Build GRCh38
rs757440179 Chr.16: 89280029-89280029 on Build GRCh38
rs773653308 Chr.16: 89279832-89279832 on Build GRCh38
rs201589586 Chr.16: 89279962-89279962 on Build GRCh38
rs202219869 Chr.16: 89280058-89280058 on Build GRCh38
rs768804170 Chr.16: 89279819-89279819 on Build GRCh38
rs202081612 Chr.16: 89279797-89279797 on Build GRCh38
rs551302848 Chr.16: 89279753-89279753 on Build GRCh38
rs759281274 Chr.16: 89280047-89280047 on Build GRCh38
rs575642464 Chr.16: 89279623-89279623 on Build GRCh38
rs763412320 Chr.16: 89279835-89279835 on Build GRCh38
rs34069598 Chr.16: 89279627-89279627 on Build GRCh38
rs529727438 Chr.16: 89279779-89279779 on Build GRCh38
rs369545274 Chr.16: 89280028-89280028 on Build GRCh38
rs763625090 Chr.16: 89279849-89279849 on Build GRCh38
rs764722888 Chr.16: 89279865-89279866 on Build GRCh38
rs772750104 Chr.16: 89279997-89279997 on Build GRCh38
rs981323228 Chr.16: 89279948-89279948 on Build GRCh38
rs758715592 Chr.16: 89280033-89280033 on Build GRCh38
rs1031398540 Chr.16: 89279806-89279806 on Build GRCh38
rs764833948 Chr.16: 89279943-89279943 on Build GRCh38
rs760007187 Chr.16: 89279848-89279848 on Build GRCh38
rs756897044 Chr.16: 89279809-89279809 on Build GRCh38
rs905177395 Chr.16: 89279632-89279632 on Build GRCh38
rs929429805 Chr.16: 89279847-89279847 on Build GRCh38
rs929288226 Chr.16: 89279969-89279969 on Build GRCh38
rs755663536 Chr.16: 89279738-89279738 on Build GRCh38
rs574631245 Chr.16: 89279892-89279892 on Build GRCh38
rs944636891 Chr.16: 89279822-89279822 on Build GRCh38
rs772511220 Chr.16: 89279930-89279930 on Build GRCh38
rs752188262 Chr.16: 89279792-89279792 on Build GRCh38
rs878855327 Chr.16: 89279756-89279756 on Build GRCh38
rs750990942 Chr.16: 89279719-89279719 on Build GRCh38
rs1027326983 Chr.16: 89279798-89279798 on Build GRCh38
rs993750197 Chr.16: 89279754-89279754 on Build GRCh38
rs746235129 Chr.16: 89279979-89279979 on Build GRCh38
rs762034600 Chr.16: 89279759-89279759 on Build GRCh38
rs374592487 Chr.16: 89279834-89279834 on Build GRCh38
rs771524336 Chr.16: 89279842-89279842 on Build GRCh38
rs767440106 Chr.16: 89280011-89280011 on Build GRCh38
rs779319208 Chr.16: 89279972-89279972 on Build GRCh38
rs756336381 Chr.16: 89279917-89279917 on Build GRCh38
rs926645829 Chr.16: 89279740-89279740 on Build GRCh38
rs751378641 Chr.16: 89279949-89279949 on Build GRCh38
rs1006348798 Chr.16: 89279635-89279635 on Build GRCh38
rs973372197 Chr.16: 89279839-89279839 on Build GRCh38
rs573355746 Chr.16: 89279715-89279715 on Build GRCh38
rs773753588 Chr.16: 89279780-89279780 on Build GRCh38
rs369149353 Chr.16: 89280061-89280061 on Build GRCh38
rs573380956 Chr.16: 89280036-89280036 on Build GRCh38
rs748812288 Chr.16: 89280044-89280044 on Build GRCh38
rs764137649 Chr.16: 89280055-89280055 on Build GRCh38
rs750764062 Chr.16: 89280032-89280032 on Build GRCh38
rs777485683 Chr.16: 89279741-89279741 on Build GRCh38
rs757615498 Chr.16: 89279971-89279971 on Build GRCh38
rs374668457 Chr.16: 89279674-89279674 on Build GRCh38
rs942026944 Chr.16: 89279837-89279837 on Build GRCh38
rs758225743 Chr.16: 89279883-89279883 on Build GRCh38
rs201043388 Chr.16: 89279796-89279796 on Build GRCh38
rs764262189 Chr.16: 89280025-89280025 on Build GRCh38
rs76793093 Chr.16: 89279755-89279755 on Build GRCh38
rs753900347 Chr.16: 89280057-89280057 on Build GRCh38
rs559089022 Chr.16: 89280049-89280049 on Build GRCh38
rs537338393 Chr.16: 89279874-89279874 on Build GRCh38
rs941977575 Chr.16: 89279742-89279742 on Build GRCh38

View Full Product Details