Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89280325-89280823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD11
Entrez Gene ID
29123
Gene Name
ankyrin repeat domain 11
Gene Aliases
ANCO-1, ANCO1, LZ16, T13
UniGene
Hs.335003
Gene Chromosome Location
Chr.16: 89267619-89490561 on Build GRCh38
Transcripts
NM_001256182.1 NM_001256183.1 NM_013275.5 XM_006721181.1 XM_006721184.2 XM_011523051.2 XM_011523053.2 XM_011523054.2 XM_011523055.2 XM_011523056.2 XM_011523057.2 XM_017023182.1 XM_017023183.1 XM_017023184.1 XM_017023185.1 XM_017023186.1 XM_017023187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3691266 c.6085G>A p.V2029I Chr.16: 89280457-89280457 on Build GRCh38
4553184 c.5835G>A p.E1945E Chr.16: 89280707-89280707 on Build GRCh38
3889144 c.5962C>T p.P1988S Chr.16: 89280580-89280580 on Build GRCh38
6259714 c.5961C>A p.F1987L Chr.16: 89280581-89280581 on Build GRCh38
6980120 c.6032C>T p.S2011L Chr.16: 89280510-89280510 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763152116 Chr.16: 89280603-89280603 on Build GRCh38
rs759965356 Chr.16: 89280608-89280608 on Build GRCh38
rs1005592913 Chr.16: 89280633-89280633 on Build GRCh38
rs748478065 Chr.16: 89280365-89280365 on Build GRCh38
rs745685018 Chr.16: 89280504-89280504 on Build GRCh38
rs777257218 Chr.16: 89280725-89280725 on Build GRCh38
rs746078253 Chr.16: 89280473-89280481 on Build GRCh38
rs749270925 Chr.16: 89280701-89280701 on Build GRCh38
rs767084927 Chr.16: 89280743-89280743 on Build GRCh38
rs142373563 Chr.16: 89280804-89280804 on Build GRCh38
rs372148958 Chr.16: 89280667-89280667 on Build GRCh38
rs113264953 Chr.16: 89280547-89280547 on Build GRCh38
rs763207005 Chr.16: 89280568-89280568 on Build GRCh38
rs776243147 Chr.16: 89280377-89280377 on Build GRCh38
rs762574237 Chr.16: 89280679-89280679 on Build GRCh38
rs774995181 Chr.16: 89280371-89280371 on Build GRCh38
rs748086945 Chr.16: 89280601-89280601 on Build GRCh38
rs373393326 Chr.16: 89280776-89280776 on Build GRCh38
rs199691161 Chr.16: 89280402-89280402 on Build GRCh38
rs371829075 Chr.16: 89280744-89280744 on Build GRCh38
rs926985506 Chr.16: 89280351-89280351 on Build GRCh38
rs376468392 Chr.16: 89280431-89280431 on Build GRCh38
rs554071391 Chr.16: 89280740-89280740 on Build GRCh38
rs755603365 Chr.16: 89280484-89280484 on Build GRCh38
rs747409293 Chr.16: 89280721-89280721 on Build GRCh38
rs976983352 Chr.16: 89280521-89280521 on Build GRCh38
rs763862589 Chr.16: 89280803-89280803 on Build GRCh38
rs750094695 Chr.16: 89280582-89280582 on Build GRCh38
rs771142694 Chr.16: 89280604-89280604 on Build GRCh38
rs757242595 Chr.16: 89280764-89280764 on Build GRCh38
rs749397444 Chr.16: 89280656-89280656 on Build GRCh38
rs373782987 Chr.16: 89280354-89280354 on Build GRCh38
rs151329092 Chr.16: 89280514-89280514 on Build GRCh38
rs553002021 Chr.16: 89280389-89280389 on Build GRCh38
rs748892371 Chr.16: 89280494-89280494 on Build GRCh38
rs777385823 Chr.16: 89280485-89280485 on Build GRCh38
rs761671642 Chr.16: 89280805-89280805 on Build GRCh38
rs891424039 Chr.16: 89280433-89280433 on Build GRCh38
rs777705464 Chr.16: 89280695-89280695 on Build GRCh38
rs542077760 Chr.16: 89280470-89280470 on Build GRCh38
rs564238731 Chr.16: 89280481-89280481 on Build GRCh38
rs953261599 Chr.16: 89280488-89280488 on Build GRCh38
rs765328496 Chr.16: 89280561-89280561 on Build GRCh38
rs766038361 Chr.16: 89280680-89280680 on Build GRCh38
rs140586593 Chr.16: 89280644-89280644 on Build GRCh38
rs759003189 Chr.16: 89280787-89280787 on Build GRCh38
rs771089110 Chr.16: 89280658-89280658 on Build GRCh38
rs775544970 Chr.16: 89280510-89280510 on Build GRCh38
rs534220503 Chr.16: 89280580-89280580 on Build GRCh38
rs760383897 Chr.16: 89280800-89280800 on Build GRCh38
rs747612277 Chr.16: 89280480-89280480 on Build GRCh38
rs60520302 Chr.16: 89280475-89280475 on Build GRCh38
rs560402669 Chr.16: 89280473-89280473 on Build GRCh38
rs929624389 Chr.16: 89280581-89280581 on Build GRCh38
rs200351209 Chr.16: 89280349-89280349 on Build GRCh38
rs200724087 Chr.16: 89280430-89280430 on Build GRCh38
rs189297725 Chr.16: 89280458-89280458 on Build GRCh38
rs768438701 Chr.16: 89280594-89280594 on Build GRCh38
rs372141194 Chr.16: 89280412-89280412 on Build GRCh38
rs775221712 Chr.16: 89280482-89280490 on Build GRCh38
rs1046804092 Chr.16: 89280482-89280482 on Build GRCh38
rs753215685 Chr.16: 89280615-89280615 on Build GRCh38
rs994275260 Chr.16: 89280620-89280620 on Build GRCh38
rs768652993 Chr.16: 89280555-89280555 on Build GRCh38
rs111596062 Chr.16: 89280455-89280455 on Build GRCh38
rs748114867 Chr.16: 89280653-89280653 on Build GRCh38
rs759714831 Chr.16: 89280665-89280665 on Build GRCh38
rs775491352 Chr.16: 89280553-89280553 on Build GRCh38
rs748832367 Chr.16: 89280545-89280545 on Build GRCh38
rs1001824505 Chr.16: 89280557-89280557 on Build GRCh38
rs772993940 Chr.16: 89280424-89280424 on Build GRCh38
rs947129807 Chr.16: 89280689-89280689 on Build GRCh38
rs574973974 Chr.16: 89280711-89280711 on Build GRCh38
rs370006772 Chr.16: 89280429-89280429 on Build GRCh38
rs779372391 Chr.16: 89280445-89280445 on Build GRCh38
rs117997391 Chr.16: 89280366-89280366 on Build GRCh38
rs765986335 Chr.16: 89280736-89280736 on Build GRCh38
rs563584451 Chr.16: 89280512-89280512 on Build GRCh38
rs745564262 Chr.16: 89280548-89280548 on Build GRCh38
rs767562546 Chr.16: 89280465-89280465 on Build GRCh38
rs753973951 Chr.16: 89280474-89280474 on Build GRCh38
rs749785439 Chr.16: 89280369-89280369 on Build GRCh38
rs766233080 Chr.16: 89280392-89280392 on Build GRCh38
rs761984613 Chr.16: 89280560-89280560 on Build GRCh38
rs1025761111 Chr.16: 89280626-89280626 on Build GRCh38
rs767333781 Chr.16: 89280685-89280685 on Build GRCh38
rs775090763 Chr.16: 89280799-89280799 on Build GRCh38
rs201636725 Chr.16: 89280436-89280436 on Build GRCh38
rs907255990 Chr.16: 89280700-89280700 on Build GRCh38
rs777776688 Chr.16: 89280655-89280655 on Build GRCh38
rs201174017 Chr.16: 89280779-89280779 on Build GRCh38
rs863223321 Chr.16: 89280588-89280589 on Build GRCh38
rs565960089 Chr.16: 89280571-89280571 on Build GRCh38
rs556725847 Chr.16: 89280686-89280686 on Build GRCh38
rs772203828 Chr.16: 89280476-89280477 on Build GRCh38
rs753767970 Chr.16: 89280759-89280759 on Build GRCh38
rs761151327 Chr.16: 89280617-89280617 on Build GRCh38
rs368770538 Chr.16: 89280722-89280722 on Build GRCh38
rs1026865529 Chr.16: 89280751-89280751 on Build GRCh38
rs758181637 Chr.16: 89280585-89280585 on Build GRCh38
rs757540026 Chr.16: 89280439-89280439 on Build GRCh38
rs748553966 Chr.16: 89280410-89280430 on Build GRCh38
rs746355740 Chr.16: 89280447-89280447 on Build GRCh38
rs747661700 Chr.16: 89280454-89280454 on Build GRCh38
rs780570915 Chr.16: 89280478-89280478 on Build GRCh38
rs762691968 Chr.16: 89280425-89280425 on Build GRCh38
rs746928698 Chr.16: 89280554-89280554 on Build GRCh38
rs773848887 Chr.16: 89280786-89280786 on Build GRCh38
rs778167926 Chr.16: 89280438-89280438 on Build GRCh38
rs886041574 Chr.16: 89280752-89280752 on Build GRCh38
rs780892836 Chr.16: 89280648-89280648 on Build GRCh38
rs1053257756 Chr.16: 89280531-89280531 on Build GRCh38
rs143712083 Chr.16: 89280671-89280671 on Build GRCh38
rs1032995945 Chr.16: 89280542-89280542 on Build GRCh38
rs773403872 Chr.16: 89280565-89280565 on Build GRCh38
rs766783032 Chr.16: 89280529-89280529 on Build GRCh38
rs772192522 Chr.16: 89280390-89280390 on Build GRCh38
rs767818765 Chr.16: 89280611-89280611 on Build GRCh38
rs774653782 Chr.16: 89280607-89280607 on Build GRCh38
rs375457115 Chr.16: 89280599-89280599 on Build GRCh38
rs199914958 Chr.16: 89280769-89280769 on Build GRCh38
rs377471988 Chr.16: 89280506-89280506 on Build GRCh38
rs756815181 Chr.16: 89280546-89280546 on Build GRCh38
rs761444143 Chr.16: 89280413-89280413 on Build GRCh38
rs780624241 Chr.16: 89280449-89280449 on Build GRCh38
rs768098760 Chr.16: 89280578-89280578 on Build GRCh38
rs746434534 Chr.16: 89280408-89280408 on Build GRCh38
rs765013102 Chr.16: 89280384-89280384 on Build GRCh38
rs776509679 Chr.16: 89280556-89280556 on Build GRCh38
rs778437406 Chr.16: 89280430-89280431 on Build GRCh38
rs763914011 Chr.16: 89280745-89280745 on Build GRCh38
rs760463786 Chr.16: 89280751-89280765 on Build GRCh38
rs373218212 Chr.16: 89280477-89280477 on Build GRCh38
rs549967314 Chr.16: 89280515-89280515 on Build GRCh38
rs755165956 Chr.16: 89280362-89280362 on Build GRCh38
rs760134096 Chr.16: 89280372-89280372 on Build GRCh38
rs746088479 Chr.16: 89280710-89280710 on Build GRCh38
rs748373867 Chr.16: 89280403-89280403 on Build GRCh38
rs547762539 Chr.16: 89280533-89280533 on Build GRCh38
rs761021002 Chr.16: 89280551-89280551 on Build GRCh38
rs200531141 Chr.16: 89280522-89280522 on Build GRCh38
rs750331558 Chr.16: 89280352-89280352 on Build GRCh38
rs764701804 Chr.16: 89280673-89280673 on Build GRCh38
rs147093946 Chr.16: 89280794-89280794 on Build GRCh38
rs751194157 Chr.16: 89280683-89280683 on Build GRCh38
rs749615651 Chr.16: 89280406-89280406 on Build GRCh38
rs778221106 Chr.16: 89280405-89280405 on Build GRCh38
rs368118594 Chr.16: 89280558-89280558 on Build GRCh38
rs376679880 Chr.16: 89280427-89280427 on Build GRCh38
rs199785661 Chr.16: 89280457-89280457 on Build GRCh38
rs760859183 Chr.16: 89280471-89280471 on Build GRCh38
rs771351601 Chr.16: 89280370-89280370 on Build GRCh38
rs747284349 Chr.16: 89280778-89280778 on Build GRCh38
rs368315956 Chr.16: 89280383-89280383 on Build GRCh38
rs752834526 Chr.16: 89280434-89280434 on Build GRCh38
rs775856441 Chr.16: 89280719-89280719 on Build GRCh38
rs779884158 Chr.16: 89280635-89280635 on Build GRCh38
rs138255986 Chr.16: 89280684-89280684 on Build GRCh38
rs199737737 Chr.16: 89280631-89280631 on Build GRCh38
rs752781169 Chr.16: 89280466-89280466 on Build GRCh38
rs774332859 Chr.16: 89280472-89280473 on Build GRCh38
rs922813408 Chr.16: 89280672-89280672 on Build GRCh38
rs771037147 Chr.16: 89280471-89280479 on Build GRCh38
rs759486921 Chr.16: 89280463-89280463 on Build GRCh38
rs755371317 Chr.16: 89280577-89280577 on Build GRCh38
rs535648640 Chr.16: 89280729-89280729 on Build GRCh38
rs771427810 Chr.16: 89280407-89280407 on Build GRCh38
rs762451759 Chr.16: 89280732-89280732 on Build GRCh38
rs1046926715 Chr.16: 89280746-89280746 on Build GRCh38
rs534182728 Chr.16: 89280417-89280417 on Build GRCh38
rs768122420 Chr.16: 89280411-89280411 on Build GRCh38
rs375589721 Chr.16: 89280518-89280518 on Build GRCh38
rs778991139 Chr.16: 89280765-89280765 on Build GRCh38
rs773948438 Chr.16: 89280501-89280501 on Build GRCh38
rs370272709 Chr.16: 89280552-89280552 on Build GRCh38
rs372396960 Chr.16: 89280590-89280590 on Build GRCh38
rs867065526 Chr.16: 89280612-89280612 on Build GRCh38
rs756460819 Chr.16: 89280404-89280404 on Build GRCh38
rs377249565 Chr.16: 89280670-89280670 on Build GRCh38
rs555015817 Chr.16: 89280396-89280396 on Build GRCh38
rs199964892 Chr.16: 89280660-89280660 on Build GRCh38
rs749868016 Chr.16: 89280466-89280467 on Build GRCh38
rs770628197 Chr.16: 89280498-89280498 on Build GRCh38
rs758335433 Chr.16: 89280353-89280353 on Build GRCh38
rs202044071 Chr.16: 89280634-89280634 on Build GRCh38
rs779935496 Chr.16: 89280588-89280588 on Build GRCh38
rs749913159 Chr.16: 89280674-89280674 on Build GRCh38
rs755982261 Chr.16: 89280691-89280691 on Build GRCh38
rs766728029 Chr.16: 89280570-89280570 on Build GRCh38
rs777086823 Chr.16: 89280544-89280544 on Build GRCh38
rs769888687 Chr.16: 89280602-89280602 on Build GRCh38
rs757293729 Chr.16: 89280703-89280703 on Build GRCh38
rs200550273 Chr.16: 89280785-89280785 on Build GRCh38
rs777204084 Chr.16: 89280782-89280782 on Build GRCh38
rs778925150 Chr.16: 89280705-89280705 on Build GRCh38
rs756624276 Chr.16: 89280431-89280439 on Build GRCh38
rs535355840 Chr.16: 89280374-89280374 on Build GRCh38
rs751266006 Chr.16: 89280643-89280643 on Build GRCh38
rs879253753 Chr.16: 89280526-89280527 on Build GRCh38
rs933089649 Chr.16: 89280525-89280525 on Build GRCh38

View Full Product Details