Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89583607-89584109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE7
Entrez Gene ID
27132
Gene Name
copine 7
Gene Aliases
-
UniGene
Hs.461775
Gene Chromosome Location
Chr.16: 89575768-89597246 on Build GRCh38
Transcripts
NM_014427.4 NM_153636.2 XM_011523000.1 XM_011523001.2 XM_017023138.1 XM_017023139.1 XM_017023140.1 XM_017023141.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4341489 c.611C>T p.P204L Chr.16: 89583725-89583725 on Build GRCh38
705303 c.677C>T p.S226L Chr.16: 89584047-89584047 on Build GRCh38
4557178 c.717G>A p.R239R Chr.16: 89584087-89584087 on Build GRCh38
4652807 c.612G>A p.P204P Chr.16: 89583726-89583726 on Build GRCh38
6634696 c.636C>T p.N212N Chr.16: 89583750-89583750 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768130590 Chr.16: 89583806-89583806 on Build GRCh38
rs12932758 Chr.16: 89583638-89583638 on Build GRCh38
rs141398937 Chr.16: 89583974-89583974 on Build GRCh38
rs897542705 Chr.16: 89583846-89583846 on Build GRCh38
rs191828877 Chr.16: 89583681-89583681 on Build GRCh38
rs945008812 Chr.16: 89583629-89583629 on Build GRCh38
rs772502780 Chr.16: 89584034-89584034 on Build GRCh38
rs774246153 Chr.16: 89584043-89584043 on Build GRCh38
rs376818326 Chr.16: 89583683-89583683 on Build GRCh38
rs1015999986 Chr.16: 89583635-89583635 on Build GRCh38
rs761407667 Chr.16: 89583764-89583764 on Build GRCh38
rs771257559 Chr.16: 89583799-89583799 on Build GRCh38
rs964595049 Chr.16: 89583885-89583885 on Build GRCh38
rs377030837 Chr.16: 89584000-89584000 on Build GRCh38
rs529539909 Chr.16: 89583999-89583999 on Build GRCh38
rs12931740 Chr.16: 89583652-89583652 on Build GRCh38
rs755079585 Chr.16: 89583785-89583785 on Build GRCh38
rs375428392 Chr.16: 89583786-89583786 on Build GRCh38
rs770099552 Chr.16: 89583790-89583790 on Build GRCh38
rs917664196 Chr.16: 89584025-89584025 on Build GRCh38
rs745675640 Chr.16: 89583691-89583691 on Build GRCh38
rs760303105 Chr.16: 89583668-89583668 on Build GRCh38
rs925355633 Chr.16: 89583783-89583783 on Build GRCh38
rs186909019 Chr.16: 89583901-89583901 on Build GRCh38
rs139532630 Chr.16: 89584041-89584041 on Build GRCh38
rs201266571 Chr.16: 89583670-89583670 on Build GRCh38
rs567272288 Chr.16: 89584080-89584080 on Build GRCh38
rs896754386 Chr.16: 89583707-89583707 on Build GRCh38
rs770263088 Chr.16: 89583723-89583723 on Build GRCh38
rs780534297 Chr.16: 89584072-89584072 on Build GRCh38
rs1029926421 Chr.16: 89583856-89583856 on Build GRCh38
rs532836570 Chr.16: 89583630-89583630 on Build GRCh38
rs774730044 Chr.16: 89583740-89583740 on Build GRCh38
rs766815520 Chr.16: 89583661-89583661 on Build GRCh38
rs761318109 Chr.16: 89583818-89583818 on Build GRCh38
rs548676643 Chr.16: 89584070-89584070 on Build GRCh38
rs372418637 Chr.16: 89583982-89583982 on Build GRCh38
rs955171010 Chr.16: 89583810-89583810 on Build GRCh38
rs747643353 Chr.16: 89584036-89584036 on Build GRCh38
rs904958591 Chr.16: 89583628-89583628 on Build GRCh38
rs569594957 Chr.16: 89583665-89583665 on Build GRCh38
rs779193206 Chr.16: 89584021-89584021 on Build GRCh38
rs945060968 Chr.16: 89583830-89583830 on Build GRCh38
rs375204346 Chr.16: 89584035-89584035 on Build GRCh38
rs926555905 Chr.16: 89583942-89583942 on Build GRCh38
rs546278189 Chr.16: 89583909-89583909 on Build GRCh38
rs534942196 Chr.16: 89583839-89583839 on Build GRCh38
rs752736185 Chr.16: 89584049-89584049 on Build GRCh38
rs146394111 Chr.16: 89584010-89584010 on Build GRCh38
rs961718719 Chr.16: 89583641-89583641 on Build GRCh38
rs958399880 Chr.16: 89583656-89583656 on Build GRCh38
rs751614658 Chr.16: 89583784-89583784 on Build GRCh38
rs115469922 Chr.16: 89584063-89584063 on Build GRCh38
rs558284865 Chr.16: 89583918-89583918 on Build GRCh38
rs567442000 Chr.16: 89583838-89583838 on Build GRCh38
rs1018515798 Chr.16: 89583881-89583881 on Build GRCh38
rs376389386 Chr.16: 89584015-89584015 on Build GRCh38
rs779864662 Chr.16: 89583699-89583699 on Build GRCh38
rs758319422 Chr.16: 89583698-89583698 on Build GRCh38
rs764831303 Chr.16: 89583770-89583770 on Build GRCh38
rs751523943 Chr.16: 89583990-89583990 on Build GRCh38
rs191373359 Chr.16: 89584064-89584064 on Build GRCh38
rs145568288 Chr.16: 89583752-89583752 on Build GRCh38
rs762377005 Chr.16: 89584011-89584011 on Build GRCh38
rs986627081 Chr.16: 89583978-89583978 on Build GRCh38
rs576526147 Chr.16: 89583921-89583921 on Build GRCh38
rs941862934 Chr.16: 89583952-89583952 on Build GRCh38
rs147558775 Chr.16: 89584066-89584066 on Build GRCh38
rs917945014 Chr.16: 89583913-89583913 on Build GRCh38
rs979585229 Chr.16: 89583897-89583898 on Build GRCh38
rs770545030 Chr.16: 89583864-89583864 on Build GRCh38
rs886653276 Chr.16: 89583915-89583915 on Build GRCh38
rs762806593 Chr.16: 89583823-89583823 on Build GRCh38
rs368893675 Chr.16: 89583822-89583822 on Build GRCh38
rs408800 Chr.16: 89583963-89583963 on Build GRCh38
rs760774452 Chr.16: 89583884-89583884 on Build GRCh38
rs924916324 Chr.16: 89584012-89584012 on Build GRCh38
rs764118471 Chr.16: 89584058-89584058 on Build GRCh38
rs763256835 Chr.16: 89583725-89583725 on Build GRCh38
rs770421296 Chr.16: 89584085-89584085 on Build GRCh38
rs369886930 Chr.16: 89584037-89584037 on Build GRCh38
rs780027856 Chr.16: 89583781-89583781 on Build GRCh38
rs1016652491 Chr.16: 89583922-89583922 on Build GRCh38
rs925366874 Chr.16: 89583899-89583899 on Build GRCh38
rs974111557 Chr.16: 89583916-89583916 on Build GRCh38
rs201835948 Chr.16: 89583726-89583726 on Build GRCh38
rs371647265 Chr.16: 89583732-89583732 on Build GRCh38
rs373955797 Chr.16: 89584033-89584033 on Build GRCh38
rs748422849 Chr.16: 89583722-89583722 on Build GRCh38
rs993132802 Chr.16: 89583847-89583847 on Build GRCh38
rs879861161 Chr.16: 89583839-89583839 on Build GRCh38
rs1013816422 Chr.16: 89583714-89583714 on Build GRCh38
rs773563880 Chr.16: 89583654-89583654 on Build GRCh38
rs1037466253 Chr.16: 89583953-89583953 on Build GRCh38
rs747514745 Chr.16: 89584079-89584079 on Build GRCh38
rs777005929 Chr.16: 89583649-89583649 on Build GRCh38
rs903597574 Chr.16: 89583664-89583664 on Build GRCh38
rs763674742 Chr.16: 89583671-89583671 on Build GRCh38
rs749539380 Chr.16: 89584014-89584014 on Build GRCh38
rs529101968 Chr.16: 89584047-89584047 on Build GRCh38
rs1055666923 Chr.16: 89583685-89583685 on Build GRCh38
rs925064733 Chr.16: 89583860-89583860 on Build GRCh38
rs867859258 Chr.16: 89583660-89583660 on Build GRCh38
rs987002059 Chr.16: 89583682-89583682 on Build GRCh38
rs748259218 Chr.16: 89583787-89583787 on Build GRCh38
rs942865544 Chr.16: 89583711-89583711 on Build GRCh38
rs776989532 Chr.16: 89583721-89583721 on Build GRCh38
rs775620919 Chr.16: 89583801-89583801 on Build GRCh38
rs200302953 Chr.16: 89583780-89583780 on Build GRCh38
rs772894474 Chr.16: 89584038-89584038 on Build GRCh38
rs149729126 Chr.16: 89583719-89583719 on Build GRCh38
rs533359433 Chr.16: 89584057-89584057 on Build GRCh38
rs368560530 Chr.16: 89583687-89583687 on Build GRCh38
rs776080905 Chr.16: 89583812-89583812 on Build GRCh38
rs551078270 Chr.16: 89583655-89583655 on Build GRCh38
rs182566268 Chr.16: 89583797-89583797 on Build GRCh38
rs750169106 Chr.16: 89583776-89583776 on Build GRCh38
rs760161077 Chr.16: 89583750-89583750 on Build GRCh38
rs748863066 Chr.16: 89584084-89584084 on Build GRCh38
rs764886872 Chr.16: 89583819-89583819 on Build GRCh38
rs949399309 Chr.16: 89583929-89583929 on Build GRCh38
rs145150175 Chr.16: 89584048-89584048 on Build GRCh38
rs201485428 Chr.16: 89584059-89584059 on Build GRCh38
rs1030406384 Chr.16: 89583973-89583973 on Build GRCh38
rs781307228 Chr.16: 89584003-89584003 on Build GRCh38
rs534559082 Chr.16: 89583861-89583861 on Build GRCh38
rs762255729 Chr.16: 89583650-89583650 on Build GRCh38
rs375506546 Chr.16: 89583803-89583803 on Build GRCh38
rs900604604 Chr.16: 89583932-89583932 on Build GRCh38
rs1016354033 Chr.16: 89583788-89583788 on Build GRCh38
rs367582915 Chr.16: 89583970-89583970 on Build GRCh38

View Full Product Details