Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89584637-89585165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE7
Entrez Gene ID
27132
Gene Name
copine 7
Gene Aliases
-
UniGene
Hs.461775
Gene Chromosome Location
Chr.16: 89575768-89597246 on Build GRCh38
Transcripts
NM_014427.4 NM_153636.2 XM_011523000.1 XM_011523001.2 XM_017023138.1 XM_017023139.1 XM_017023140.1 XM_017023141.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3889154 c.784G>A p.D262N Chr.16: 89584825-89584825 on Build GRCh38
133472 c.763_764GA>TC p.E255S Chr.16: 89584804-89584805 on Build GRCh38
1709520 c.781G>A p.D261N Chr.16: 89584822-89584822 on Build GRCh38
5887433 c.816+7G>A p.? Chr.16: 89584864-89584864 on Build GRCh38
6634700 c.809G>C p.R270T Chr.16: 89584850-89584850 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1019216114 Chr.16: 89584672-89584672 on Build GRCh38
rs746768725 Chr.16: 89584822-89584822 on Build GRCh38
rs1018863244 Chr.16: 89584702-89584702 on Build GRCh38
rs999428891 Chr.16: 89585036-89585036 on Build GRCh38
rs564608544 Chr.16: 89584849-89584849 on Build GRCh38
rs747972308 Chr.16: 89584875-89584875 on Build GRCh38
rs977843955 Chr.16: 89584705-89584705 on Build GRCh38
rs921309999 Chr.16: 89584678-89584678 on Build GRCh38
rs927824258 Chr.16: 89584964-89584964 on Build GRCh38
rs367918809 Chr.16: 89584868-89584868 on Build GRCh38
rs752433834 Chr.16: 89585135-89585135 on Build GRCh38
rs755380868 Chr.16: 89584769-89584769 on Build GRCh38
rs745393139 Chr.16: 89584809-89584809 on Build GRCh38
rs773239465 Chr.16: 89584831-89584831 on Build GRCh38
rs953560501 Chr.16: 89584670-89584670 on Build GRCh38
rs557646009 Chr.16: 89584878-89584878 on Build GRCh38
rs775957418 Chr.16: 89584902-89584902 on Build GRCh38
rs3830555 Chr.16: 89584767-89584768 on Build GRCh38
rs761792156 Chr.16: 89584825-89584825 on Build GRCh38
rs900436236 Chr.16: 89584946-89584946 on Build GRCh38
rs540345767 Chr.16: 89584940-89584940 on Build GRCh38
rs761123364 Chr.16: 89584903-89584903 on Build GRCh38
rs773149702 Chr.16: 89584879-89584879 on Build GRCh38
rs1040135490 Chr.16: 89585030-89585030 on Build GRCh38
rs780995446 Chr.16: 89584874-89584874 on Build GRCh38
rs371576080 Chr.16: 89584730-89584730 on Build GRCh38
rs1033271776 Chr.16: 89585037-89585037 on Build GRCh38
rs564846957 Chr.16: 89585027-89585027 on Build GRCh38
rs1040444315 Chr.16: 89585099-89585099 on Build GRCh38
rs1054415516 Chr.16: 89585117-89585117 on Build GRCh38
rs529989121 Chr.16: 89585121-89585121 on Build GRCh38
rs926001774 Chr.16: 89584709-89584709 on Build GRCh38
rs981475694 Chr.16: 89584797-89584797 on Build GRCh38
rs532135321 Chr.16: 89585072-89585072 on Build GRCh38
rs751863155 Chr.16: 89584850-89584850 on Build GRCh38
rs775098018 Chr.16: 89584727-89584727 on Build GRCh38
rs781533919 Chr.16: 89584776-89584776 on Build GRCh38
rs577640750 Chr.16: 89584863-89584863 on Build GRCh38
rs768563080 Chr.16: 89584824-89584824 on Build GRCh38
rs369203889 Chr.16: 89584888-89584888 on Build GRCh38
rs779772134 Chr.16: 89584821-89584821 on Build GRCh38
rs756712449 Chr.16: 89584882-89584883 on Build GRCh38
rs756792230 Chr.16: 89584792-89584792 on Build GRCh38
rs981144186 Chr.16: 89584991-89584991 on Build GRCh38
rs971352407 Chr.16: 89585077-89585077 on Build GRCh38
rs1040747019 Chr.16: 89584914-89584914 on Build GRCh38
rs766527793 Chr.16: 89584848-89584848 on Build GRCh38
rs1050060394 Chr.16: 89585014-89585014 on Build GRCh38
rs991065599 Chr.16: 89585029-89585029 on Build GRCh38
rs199865735 Chr.16: 89584762-89584762 on Build GRCh38
rs546919500 Chr.16: 89585112-89585112 on Build GRCh38
rs75059901 Chr.16: 89584913-89584913 on Build GRCh38
rs947891332 Chr.16: 89584773-89584773 on Build GRCh38
rs866984421 Chr.16: 89584843-89584843 on Build GRCh38
rs930813403 Chr.16: 89585056-89585056 on Build GRCh38
rs188808451 Chr.16: 89584915-89584915 on Build GRCh38
rs971521908 Chr.16: 89584990-89584990 on Build GRCh38
rs527950295 Chr.16: 89585129-89585129 on Build GRCh38
rs575891638 Chr.16: 89584885-89584885 on Build GRCh38
rs940504638 Chr.16: 89584966-89584966 on Build GRCh38
rs761795396 Chr.16: 89584746-89584746 on Build GRCh38
rs200930124 Chr.16: 89584862-89584862 on Build GRCh38
rs1022896118 Chr.16: 89585073-89585073 on Build GRCh38
rs776690752 Chr.16: 89584733-89584733 on Build GRCh38
rs769748617 Chr.16: 89584829-89584829 on Build GRCh38
rs376182274 Chr.16: 89584851-89584851 on Build GRCh38
rs920504537 Chr.16: 89584989-89584989 on Build GRCh38
rs766832730 Chr.16: 89585013-89585013 on Build GRCh38
rs907964275 Chr.16: 89585085-89585085 on Build GRCh38
rs181270783 Chr.16: 89585034-89585034 on Build GRCh38
rs957819428 Chr.16: 89585055-89585055 on Build GRCh38
rs112368769 Chr.16: 89585136-89585136 on Build GRCh38
rs967594791 Chr.16: 89584686-89584686 on Build GRCh38
rs1010071277 Chr.16: 89584976-89584976 on Build GRCh38
rs957434752 Chr.16: 89584883-89584883 on Build GRCh38
rs200261865 Chr.16: 89584867-89584867 on Build GRCh38
rs375453188 Chr.16: 89584864-89584864 on Build GRCh38
rs536455318 Chr.16: 89584761-89584761 on Build GRCh38
rs767961893 Chr.16: 89584853-89584853 on Build GRCh38
rs753234982 Chr.16: 89584867-89584868 on Build GRCh38
rs1036315386 Chr.16: 89584967-89584967 on Build GRCh38
rs753121212 Chr.16: 89584854-89584854 on Build GRCh38
rs397740527 Chr.16: 89584771-89584772 on Build GRCh38
rs532413867 Chr.16: 89584671-89584671 on Build GRCh38
rs763027012 Chr.16: 89584836-89584836 on Build GRCh38
rs774400337 Chr.16: 89584896-89584896 on Build GRCh38
rs888888539 Chr.16: 89585020-89585020 on Build GRCh38
rs767655218 Chr.16: 89584901-89584901 on Build GRCh38
rs771808309 Chr.16: 89584820-89584820 on Build GRCh38
rs377666069 Chr.16: 89584719-89584719 on Build GRCh38
rs981927235 Chr.16: 89584929-89584929 on Build GRCh38
rs769654999 Chr.16: 89584876-89584876 on Build GRCh38
rs765341922 Chr.16: 89584755-89584755 on Build GRCh38
rs964950537 Chr.16: 89584703-89584703 on Build GRCh38
rs961943533 Chr.16: 89585090-89585090 on Build GRCh38
rs114838488 Chr.16: 89584791-89584791 on Build GRCh38
rs777867654 Chr.16: 89584892-89584893 on Build GRCh38
rs1009307925 Chr.16: 89584684-89584684 on Build GRCh38
rs1010081259 Chr.16: 89585138-89585138 on Build GRCh38
rs1046179589 Chr.16: 89584774-89584774 on Build GRCh38
rs148404104 Chr.16: 89584729-89584729 on Build GRCh38
rs920536806 Chr.16: 89585124-89585124 on Build GRCh38
rs900531797 Chr.16: 89585102-89585102 on Build GRCh38
rs750625522 Chr.16: 89584757-89584757 on Build GRCh38
rs773554090 Chr.16: 89584897-89584897 on Build GRCh38
rs751806482 Chr.16: 89584768-89584768 on Build GRCh38
rs572960212 Chr.16: 89584928-89584928 on Build GRCh38
rs73264926 Chr.16: 89584800-89584800 on Build GRCh38
rs908625098 Chr.16: 89585087-89585087 on Build GRCh38
rs981519539 Chr.16: 89585086-89585086 on Build GRCh38

View Full Product Details