Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89657715-89658215 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHMP1A
Entrez Gene ID
5119
Gene Name
charged multivesicular body protein 1A
Gene Aliases
CHMP1, PCH8, PCOLN3, PRSM1, VPS46-1, VPS46A
UniGene
Hs.589427
Gene Chromosome Location
Chr.16: 89644431-89657785 on Build GRCh38
Transcripts
NM_001083314.3 NM_002768.4 NR_046418.2 NM_001271907.1 NM_001271908.1 NM_001271909.1 NM_001271910.1 NM_153025.2 NR_073515.1 NR_073516.1 XM_006721133.2

Gene Symbol
SPATA33
Entrez Gene ID
124045
Gene Name
spermatogenesis associated 33
Gene Aliases
C16orf55
UniGene
Hs.655171
Gene Chromosome Location
Chr.16: 89657744-89670458 on Build GRCh38
Transcripts
NM_001083314.3 NM_002768.4 NR_046418.2 NM_001271907.1 NM_001271908.1 NM_001271909.1 NM_001271910.1 NM_153025.2 NR_073515.1 NR_073516.1 XM_006721133.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5811209 c.26G>A p.R9K Chr.16: 89657827-89657827 on Build GRCh38
6436101 c.25A>T p.R9W Chr.16: 89657826-89657826 on Build GRCh38
4957590 c.30C>T p.T10T Chr.16: 89657831-89657831 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765301069 Chr.16: 89657768-89657768 on Build GRCh38
rs762450417 Chr.16: 89657858-89657858 on Build GRCh38
rs888058382 Chr.16: 89658002-89658002 on Build GRCh38
rs531084822 Chr.16: 89657874-89657874 on Build GRCh38
rs1054894733 Chr.16: 89658159-89658159 on Build GRCh38
rs1004783815 Chr.16: 89658185-89658185 on Build GRCh38
rs142356394 Chr.16: 89657805-89657805 on Build GRCh38
rs769610233 Chr.16: 89658062-89658062 on Build GRCh38
rs1003324162 Chr.16: 89657887-89657887 on Build GRCh38
rs1056220910 Chr.16: 89657982-89657982 on Build GRCh38
rs571246014 Chr.16: 89658089-89658089 on Build GRCh38
rs755235146 Chr.16: 89658149-89658149 on Build GRCh38
rs771791780 Chr.16: 89657753-89657753 on Build GRCh38
rs201134136 Chr.16: 89657758-89657758 on Build GRCh38
rs771642101 Chr.16: 89657848-89657848 on Build GRCh38
rs900757626 Chr.16: 89657792-89657792 on Build GRCh38
rs980683078 Chr.16: 89658063-89658063 on Build GRCh38
rs973366483 Chr.16: 89657831-89657832 on Build GRCh38
rs779651553 Chr.16: 89658029-89658029 on Build GRCh38
rs1007756565 Chr.16: 89658004-89658004 on Build GRCh38
rs180983213 Chr.16: 89658173-89658173 on Build GRCh38
rs753224788 Chr.16: 89657827-89657827 on Build GRCh38
rs773086491 Chr.16: 89658072-89658072 on Build GRCh38
rs1002005247 Chr.16: 89658177-89658177 on Build GRCh38
rs956296284 Chr.16: 89658042-89658042 on Build GRCh38
rs778437378 Chr.16: 89657829-89657829 on Build GRCh38
rs780840564 Chr.16: 89658057-89658057 on Build GRCh38
rs1033547620 Chr.16: 89657825-89657825 on Build GRCh38
rs756550783 Chr.16: 89658010-89658010 on Build GRCh38
rs140600782 Chr.16: 89658115-89658116 on Build GRCh38
rs553132334 Chr.16: 89658079-89658079 on Build GRCh38
rs939125219 Chr.16: 89657981-89657981 on Build GRCh38
rs915181196 Chr.16: 89657931-89657931 on Build GRCh38
rs967993763 Chr.16: 89658061-89658061 on Build GRCh38
rs781658601 Chr.16: 89657818-89657818 on Build GRCh38
rs960415413 Chr.16: 89658080-89658080 on Build GRCh38
rs746720860 Chr.16: 89657867-89657867 on Build GRCh38
rs915546606 Chr.16: 89658097-89658097 on Build GRCh38
rs154664 Chr.16: 89658164-89658164 on Build GRCh38
rs763037005 Chr.16: 89657782-89657782 on Build GRCh38
rs760344651 Chr.16: 89657757-89657757 on Build GRCh38
rs966085380 Chr.16: 89658000-89658000 on Build GRCh38
rs776406371 Chr.16: 89657760-89657760 on Build GRCh38
rs929033226 Chr.16: 89658092-89658092 on Build GRCh38
rs750563817 Chr.16: 89657769-89657769 on Build GRCh38
rs754050314 Chr.16: 89658106-89658106 on Build GRCh38
rs756388492 Chr.16: 89657924-89657924 on Build GRCh38
rs757822051 Chr.16: 89658024-89658024 on Build GRCh38
rs1053511487 Chr.16: 89658152-89658152 on Build GRCh38
rs946379265 Chr.16: 89657775-89657775 on Build GRCh38
rs901853186 Chr.16: 89658023-89658023 on Build GRCh38
rs939071595 Chr.16: 89657790-89657790 on Build GRCh38
rs950737585 Chr.16: 89657988-89657988 on Build GRCh38
rs939058359 Chr.16: 89658156-89658156 on Build GRCh38
rs943500011 Chr.16: 89658111-89658111 on Build GRCh38
rs933356348 Chr.16: 89658150-89658150 on Build GRCh38
rs1003374909 Chr.16: 89657824-89657824 on Build GRCh38
rs957570649 Chr.16: 89658043-89658043 on Build GRCh38
rs769665039 Chr.16: 89657891-89657891 on Build GRCh38
rs747875399 Chr.16: 89658058-89658058 on Build GRCh38
rs570063284 Chr.16: 89657864-89657864 on Build GRCh38
rs929363357 Chr.16: 89657747-89657747 on Build GRCh38
rs774414582 Chr.16: 89657970-89657970 on Build GRCh38
rs562172129 Chr.16: 89657736-89657736 on Build GRCh38
rs1039268879 Chr.16: 89658122-89658122 on Build GRCh38
rs1020726946 Chr.16: 89657811-89657811 on Build GRCh38
rs983465542 Chr.16: 89657833-89657833 on Build GRCh38
rs920636388 Chr.16: 89657738-89657738 on Build GRCh38
rs929508012 Chr.16: 89657990-89657990 on Build GRCh38
rs903693132 Chr.16: 89658175-89658175 on Build GRCh38
rs759842731 Chr.16: 89657985-89657985 on Build GRCh38
rs956575719 Chr.16: 89657917-89657917 on Build GRCh38
rs1033495248 Chr.16: 89657936-89657936 on Build GRCh38
rs766504120 Chr.16: 89657954-89657954 on Build GRCh38
rs922149965 Chr.16: 89657855-89657855 on Build GRCh38
rs926271028 Chr.16: 89657964-89657964 on Build GRCh38
rs894996446 Chr.16: 89657979-89657979 on Build GRCh38
rs1042063078 Chr.16: 89658099-89658099 on Build GRCh38
rs959539672 Chr.16: 89657830-89657830 on Build GRCh38
rs372275615 Chr.16: 89657998-89657998 on Build GRCh38
rs891907733 Chr.16: 89658155-89658155 on Build GRCh38
rs1027580236 Chr.16: 89657986-89657986 on Build GRCh38
rs754721627 Chr.16: 89658045-89658045 on Build GRCh38
rs748068250 Chr.16: 89657886-89657886 on Build GRCh38
rs980465325 Chr.16: 89657958-89657958 on Build GRCh38
rs151272435 Chr.16: 89658182-89658182 on Build GRCh38
rs1049329414 Chr.16: 89657846-89657846 on Build GRCh38
rs1022179614 Chr.16: 89658170-89658170 on Build GRCh38
rs764654628 Chr.16: 89658014-89658014 on Build GRCh38
rs567317637 Chr.16: 89657992-89657992 on Build GRCh38
rs762981927 Chr.16: 89657906-89657906 on Build GRCh38
rs11545432 Chr.16: 89657905-89657905 on Build GRCh38
rs918971303 Chr.16: 89657984-89657984 on Build GRCh38
rs936299533 Chr.16: 89657803-89657803 on Build GRCh38
rs374976099 Chr.16: 89658162-89658162 on Build GRCh38
rs1039048987 Chr.16: 89658021-89658021 on Build GRCh38
rs746399057 Chr.16: 89658009-89658010 on Build GRCh38
rs575460448 Chr.16: 89658138-89658138 on Build GRCh38
rs967939661 Chr.16: 89657946-89657946 on Build GRCh38
rs1042115767 Chr.16: 89657882-89657882 on Build GRCh38
rs533191161 Chr.16: 89657853-89657853 on Build GRCh38
rs988352468 Chr.16: 89657923-89657923 on Build GRCh38
rs994036251 Chr.16: 89657821-89657821 on Build GRCh38
rs1009427002 Chr.16: 89658046-89658046 on Build GRCh38
rs756742875 Chr.16: 89657828-89657828 on Build GRCh38
rs911977627 Chr.16: 89657835-89657835 on Build GRCh38
rs960888258 Chr.16: 89658190-89658190 on Build GRCh38
rs1031769674 Chr.16: 89658032-89658032 on Build GRCh38
rs774913820 Chr.16: 89657997-89657998 on Build GRCh38
rs35415928 Chr.16: 89657860-89657860 on Build GRCh38
rs202019177 Chr.16: 89657755-89657755 on Build GRCh38
rs936228584 Chr.16: 89657857-89657857 on Build GRCh38
rs894938244 Chr.16: 89658163-89658163 on Build GRCh38
rs372295056 Chr.16: 89657892-89657892 on Build GRCh38
rs538677252 Chr.16: 89658091-89658091 on Build GRCh38
rs1007536100 Chr.16: 89657850-89657850 on Build GRCh38
rs966133536 Chr.16: 89657872-89657872 on Build GRCh38
rs992210535 Chr.16: 89658085-89658085 on Build GRCh38
rs761774054 Chr.16: 89657764-89657764 on Build GRCh38
rs997892716 Chr.16: 89658028-89658028 on Build GRCh38
rs766741888 Chr.16: 89658022-89658022 on Build GRCh38
rs369619180 Chr.16: 89657996-89657996 on Build GRCh38
rs559303642 Chr.16: 89657831-89657831 on Build GRCh38
rs754416340 Chr.16: 89658020-89658020 on Build GRCh38
rs557247042 Chr.16: 89658109-89658109 on Build GRCh38
rs746633803 Chr.16: 89658041-89658041 on Build GRCh38
rs893431424 Chr.16: 89657862-89657862 on Build GRCh38
rs770885525 Chr.16: 89658076-89658076 on Build GRCh38
rs751840485 Chr.16: 89657809-89657809 on Build GRCh38
rs867655591 Chr.16: 89658100-89658100 on Build GRCh38
rs926386907 Chr.16: 89657788-89657788 on Build GRCh38
rs542772990 Chr.16: 89658142-89658142 on Build GRCh38

View Full Product Details