Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89957330-89957819 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEF8
Entrez Gene ID
54849
Gene Name
differentially expressed in FDCP 8 homolog (mouse)
Gene Aliases
-
UniGene
Hs.62771
Gene Chromosome Location
Chr.16: 89948697-89968060 on Build GRCh38
Transcripts
NM_001242816.1 NM_001242817.1 NM_001242818.1 NM_001242819.1 NM_001242820.1 NM_001242821.1 NM_001242822.1 NM_017702.3 NM_207514.2 XM_005256318.2 XM_011523156.2 XM_011523157.2 XM_011523158.1 XM_011523159.2 XM_011523160.2 XM_011523161.2 XM_011523162.2 XM_017023358.1 XM_017023359.1 XM_017023360.1 XM_017023361.1 XM_017023362.1 XM_017023363.1 XM_017023364.1 XM_017023365.1 XM_017023366.1 XM_017023367.1 XM_017023368.1 XM_017023369.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4593393 c.301G>A p.E101K Chr.16: 89957589-89957589 on Build GRCh38
4886035 c.491C>T p.S164L Chr.16: 89957596-89957596 on Build GRCh38
6398501 c.511G>T p.V171L Chr.16: 89957616-89957616 on Build GRCh38
4921359 c.483C>A p.P161P Chr.16: 89957588-89957588 on Build GRCh38
4886036 c.308C>T p.S103L Chr.16: 89957596-89957596 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1019027660 Chr.16: 89957695-89957695 on Build GRCh38
rs778842562 Chr.16: 89957556-89957556 on Build GRCh38
rs562216716 Chr.16: 89957754-89957754 on Build GRCh38
rs769964230 Chr.16: 89957489-89957489 on Build GRCh38
rs757957570 Chr.16: 89957596-89957596 on Build GRCh38
rs746487233 Chr.16: 89957448-89957448 on Build GRCh38
rs376045841 Chr.16: 89957701-89957701 on Build GRCh38
rs914973279 Chr.16: 89957567-89957567 on Build GRCh38
rs1029172471 Chr.16: 89957744-89957745 on Build GRCh38
rs749246062 Chr.16: 89957474-89957474 on Build GRCh38
rs757055290 Chr.16: 89957636-89957636 on Build GRCh38
rs759640026 Chr.16: 89957492-89957492 on Build GRCh38
rs1001026075 Chr.16: 89957720-89957720 on Build GRCh38
rs34573241 Chr.16: 89957783-89957783 on Build GRCh38
rs141475941 Chr.16: 89957534-89957534 on Build GRCh38
rs778609903 Chr.16: 89957637-89957637 on Build GRCh38
rs773516794 Chr.16: 89957614-89957614 on Build GRCh38
rs990440482 Chr.16: 89957756-89957756 on Build GRCh38
rs763553148 Chr.16: 89957611-89957611 on Build GRCh38
rs770829787 Chr.16: 89957476-89957476 on Build GRCh38
rs978242401 Chr.16: 89957773-89957773 on Build GRCh38
rs894976969 Chr.16: 89957489-89957489 on Build GRCh38
rs139092099 Chr.16: 89957577-89957577 on Build GRCh38
rs763939045 Chr.16: 89957374-89957374 on Build GRCh38
rs372563628 Chr.16: 89957555-89957555 on Build GRCh38
rs565607436 Chr.16: 89957410-89957410 on Build GRCh38
rs200393892 Chr.16: 89957583-89957583 on Build GRCh38
rs554253945 Chr.16: 89957551-89957551 on Build GRCh38
rs748316515 Chr.16: 89957685-89957685 on Build GRCh38
rs576368067 Chr.16: 89957682-89957682 on Build GRCh38
rs867935630 Chr.16: 89957499-89957499 on Build GRCh38
rs775078371 Chr.16: 89957799-89957799 on Build GRCh38
rs149889201 Chr.16: 89957654-89957654 on Build GRCh38
rs764432840 Chr.16: 89957501-89957501 on Build GRCh38
rs922099435 Chr.16: 89957392-89957392 on Build GRCh38
rs775586977 Chr.16: 89957494-89957494 on Build GRCh38
rs375968022 Chr.16: 89957372-89957372 on Build GRCh38
rs566835264 Chr.16: 89957366-89957366 on Build GRCh38
rs138165651 Chr.16: 89957615-89957615 on Build GRCh38
rs931941285 Chr.16: 89957622-89957622 on Build GRCh38
rs945037883 Chr.16: 89957444-89957444 on Build GRCh38
rs938963506 Chr.16: 89957787-89957787 on Build GRCh38
rs189965578 Chr.16: 89957450-89957450 on Build GRCh38
rs917611342 Chr.16: 89957535-89957535 on Build GRCh38
rs780688966 Chr.16: 89957544-89957544 on Build GRCh38
rs749047512 Chr.16: 89957552-89957552 on Build GRCh38
rs773702392 Chr.16: 89957689-89957689 on Build GRCh38
rs1042460825 Chr.16: 89957446-89957446 on Build GRCh38
rs34006432 Chr.16: 89957779-89957779 on Build GRCh38
rs200998494 Chr.16: 89957640-89957640 on Build GRCh38
rs766997295 Chr.16: 89957614-89957614 on Build GRCh38
rs779471894 Chr.16: 89957616-89957616 on Build GRCh38
rs982640610 Chr.16: 89957727-89957727 on Build GRCh38
rs570012864 Chr.16: 89957441-89957441 on Build GRCh38
rs967605281 Chr.16: 89957771-89957771 on Build GRCh38
rs753754903 Chr.16: 89957420-89957420 on Build GRCh38
rs760882356 Chr.16: 89957498-89957498 on Build GRCh38
rs747144290 Chr.16: 89957680-89957680 on Build GRCh38
rs893549212 Chr.16: 89957430-89957430 on Build GRCh38
rs754500133 Chr.16: 89957449-89957449 on Build GRCh38
rs541226021 Chr.16: 89957793-89957793 on Build GRCh38
rs753437162 Chr.16: 89957623-89957623 on Build GRCh38
rs551349186 Chr.16: 89957440-89957440 on Build GRCh38
rs965979547 Chr.16: 89957700-89957700 on Build GRCh38
rs555306507 Chr.16: 89957451-89957451 on Build GRCh38
rs12596872 Chr.16: 89957493-89957493 on Build GRCh38
rs760864565 Chr.16: 89957585-89957585 on Build GRCh38
rs922191560 Chr.16: 89957538-89957538 on Build GRCh38
rs148317300 Chr.16: 89957513-89957513 on Build GRCh38
rs983581194 Chr.16: 89957512-89957512 on Build GRCh38
rs995446867 Chr.16: 89957383-89957383 on Build GRCh38
rs951257261 Chr.16: 89957668-89957668 on Build GRCh38
rs773059351 Chr.16: 89957472-89957472 on Build GRCh38
rs151177056 Chr.16: 89957417-89957417 on Build GRCh38
rs1033636042 Chr.16: 89957466-89957466 on Build GRCh38
rs1049699868 Chr.16: 89957638-89957638 on Build GRCh38
rs8049897 Chr.16: 89957794-89957794 on Build GRCh38
rs371680963 Chr.16: 89957757-89957757 on Build GRCh38
rs377269615 Chr.16: 89957588-89957588 on Build GRCh38
rs769483328 Chr.16: 89957462-89957462 on Build GRCh38
rs967556605 Chr.16: 89957777-89957803 on Build GRCh38
rs762449946 Chr.16: 89957515-89957515 on Build GRCh38
rs368298082 Chr.16: 89957531-89957531 on Build GRCh38
rs1003443966 Chr.16: 89957464-89957464 on Build GRCh38
rs8051733 Chr.16: 89957798-89957798 on Build GRCh38
rs749904744 Chr.16: 89957589-89957589 on Build GRCh38
rs868039790 Chr.16: 89957761-89957761 on Build GRCh38
rs755781969 Chr.16: 89957549-89957549 on Build GRCh38
rs547493603 Chr.16: 89957375-89957375 on Build GRCh38
rs774449903 Chr.16: 89957483-89957483 on Build GRCh38
rs529506375 Chr.16: 89957778-89957778 on Build GRCh38
rs572167104 Chr.16: 89957571-89957571 on Build GRCh38
rs367591039 Chr.16: 89957597-89957597 on Build GRCh38
rs780046628 Chr.16: 89957674-89957674 on Build GRCh38
rs765876348 Chr.16: 89957525-89957525 on Build GRCh38
rs544074238 Chr.16: 89957683-89957683 on Build GRCh38

View Full Product Details