Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 3924172-3924673 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2A3
Entrez Gene ID
489
Gene Name
ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 3
Gene Aliases
SERCA3
UniGene
Hs.513870
Gene Chromosome Location
Chr.17: 3923869-3964464 on Build GRCh38
Transcripts
NM_005173.3 NM_174953.2 NM_174954.2 NM_174955.2 NM_174956.2 NM_174957.2 NM_174958.2 XM_011523885.1 XM_011523888.2

SNP Detail

SNP ID Chromosome Location
rs974033673 Chr.17: 3924585-3924585 on Build GRCh38
rs1034978639 Chr.17: 3924428-3924428 on Build GRCh38
rs562992516 Chr.17: 3924536-3924536 on Build GRCh38
rs1013214493 Chr.17: 3924550-3924550 on Build GRCh38
rs958837453 Chr.17: 3924421-3924421 on Build GRCh38
rs917605 Chr.17: 3924208-3924208 on Build GRCh38
rs967636314 Chr.17: 3924214-3924214 on Build GRCh38
rs1000551788 Chr.17: 3924530-3924530 on Build GRCh38
rs917356106 Chr.17: 3924271-3924271 on Build GRCh38
rs535699504 Chr.17: 3924486-3924486 on Build GRCh38
rs747663304 Chr.17: 3924501-3924508 on Build GRCh38
rs979461367 Chr.17: 3924219-3924219 on Build GRCh38
rs934326976 Chr.17: 3924655-3924656 on Build GRCh38
rs566998762 Chr.17: 3924508-3924508 on Build GRCh38
rs1024472458 Chr.17: 3924553-3924553 on Build GRCh38
rs756644381 Chr.17: 3924498-3924498 on Build GRCh38
rs920667775 Chr.17: 3924462-3924462 on Build GRCh38
rs926100920 Chr.17: 3924231-3924231 on Build GRCh38
rs190227750 Chr.17: 3924349-3924349 on Build GRCh38
rs567622696 Chr.17: 3924617-3924617 on Build GRCh38
rs554696338 Chr.17: 3924487-3924487 on Build GRCh38
rs761834790 Chr.17: 3924414-3924414 on Build GRCh38
rs17846883 Chr.17: 3924199-3924199 on Build GRCh38
rs560326758 Chr.17: 3924533-3924533 on Build GRCh38
rs781708978 Chr.17: 3924426-3924426 on Build GRCh38
rs527493782 Chr.17: 3924608-3924608 on Build GRCh38
rs930934790 Chr.17: 3924291-3924291 on Build GRCh38
rs557428564 Chr.17: 3924412-3924412 on Build GRCh38
rs537437239 Chr.17: 3924375-3924375 on Build GRCh38
rs912361805 Chr.17: 3924618-3924618 on Build GRCh38
rs962182593 Chr.17: 3924452-3924453 on Build GRCh38
rs41283377 Chr.17: 3924583-3924583 on Build GRCh38
rs1038259835 Chr.17: 3924332-3924332 on Build GRCh38
rs917447066 Chr.17: 3924436-3924436 on Build GRCh38
rs143751516 Chr.17: 3924581-3924581 on Build GRCh38
rs890861608 Chr.17: 3924369-3924369 on Build GRCh38
rs530952721 Chr.17: 3924586-3924586 on Build GRCh38
rs955020022 Chr.17: 3924198-3924198 on Build GRCh38
rs768512816 Chr.17: 3924256-3924256 on Build GRCh38
rs540892880 Chr.17: 3924507-3924507 on Build GRCh38
rs950161728 Chr.17: 3924473-3924473 on Build GRCh38
rs542874491 Chr.17: 3924561-3924561 on Build GRCh38
rs1047040186 Chr.17: 3924307-3924307 on Build GRCh38
rs1038229132 Chr.17: 3924500-3924500 on Build GRCh38
rs746824929 Chr.17: 3924233-3924233 on Build GRCh38
rs972540265 Chr.17: 3924461-3924461 on Build GRCh38
rs528862440 Chr.17: 3924541-3924541 on Build GRCh38
rs572174095 Chr.17: 3924504-3924504 on Build GRCh38
rs1009752670 Chr.17: 3924404-3924404 on Build GRCh38
rs983306551 Chr.17: 3924479-3924479 on Build GRCh38
rs932566067 Chr.17: 3924607-3924607 on Build GRCh38
rs534920744 Chr.17: 3924251-3924251 on Build GRCh38
rs543005354 Chr.17: 3924535-3924535 on Build GRCh38
rs138079189 Chr.17: 3924420-3924420 on Build GRCh38
rs887147326 Chr.17: 3924310-3924310 on Build GRCh38
rs749664962 Chr.17: 3924505-3924505 on Build GRCh38
rs183134677 Chr.17: 3924370-3924370 on Build GRCh38
rs1020614884 Chr.17: 3924411-3924411 on Build GRCh38
rs373023676 Chr.17: 3924502-3924502 on Build GRCh38
rs932597573 Chr.17: 3924503-3924503 on Build GRCh38
rs866492175 Chr.17: 3924623-3924623 on Build GRCh38
rs903479309 Chr.17: 3924512-3924512 on Build GRCh38
rs1042115118 Chr.17: 3924622-3924622 on Build GRCh38
rs17846887 Chr.17: 3924493-3924493 on Build GRCh38
rs376351678 Chr.17: 3924323-3924323 on Build GRCh38
rs1000646616 Chr.17: 3924639-3924639 on Build GRCh38
rs941281115 Chr.17: 3924331-3924331 on Build GRCh38
rs369908741 Chr.17: 3924340-3924340 on Build GRCh38
rs536662009 Chr.17: 3924646-3924646 on Build GRCh38
rs551095245 Chr.17: 3924613-3924613 on Build GRCh38
rs996655481 Chr.17: 3924339-3924339 on Build GRCh38
rs899638473 Chr.17: 3924336-3924336 on Build GRCh38
rs188596441 Chr.17: 3924578-3924578 on Build GRCh38
rs192830154 Chr.17: 3924584-3924584 on Build GRCh38
rs907165245 Chr.17: 3924573-3924573 on Build GRCh38
rs565889210 Chr.17: 3924211-3924211 on Build GRCh38
rs551823635 Chr.17: 3924273-3924273 on Build GRCh38
rs559640562 Chr.17: 3924562-3924562 on Build GRCh38
rs993484558 Chr.17: 3924596-3924596 on Build GRCh38
rs998995347 Chr.17: 3924384-3924384 on Build GRCh38
rs950208723 Chr.17: 3924281-3924281 on Build GRCh38
rs945205388 Chr.17: 3924621-3924621 on Build GRCh38

View Full Product Details