Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4967639-4968142 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMTA2
Entrez Gene ID
23125
Gene Name
calmodulin binding transcription activator 2
Gene Aliases
-
UniGene
Hs.632242
Gene Chromosome Location
Chr.17: 4967992-4988297 on Build GRCh38
Transcripts
NM_001171166.1 NM_001171167.1 NM_001171168.1 NM_015099.3 XM_006721478.3 XM_006721481.3 XM_006721482.3 XM_011523746.2 XM_011523747.2 XM_011523748.2 XM_011523749.2 NM_004890.2

Gene Symbol
SPAG7
Entrez Gene ID
9552
Gene Name
sperm associated antigen 7
Gene Aliases
ACRP, FSA-1
UniGene
Hs.90436
Gene Chromosome Location
Chr.17: 4959226-4967837 on Build GRCh38
Transcripts
NM_001171166.1 NM_001171167.1 NM_001171168.1 NM_015099.3 XM_006721478.3 XM_006721481.3 XM_006721482.3 XM_011523746.2 XM_011523747.2 XM_011523748.2 XM_011523749.2 NM_004890.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3989147 c.69T>G p.T23T Chr.17: 4967736-4967736 on Build GRCh38
7283852 c.3G>A p.M1I Chr.17: 4967802-4967802 on Build GRCh38
3192749 c.42G>A p.K14K Chr.17: 4967763-4967763 on Build GRCh38
4579839 c.66G>C p.E22D Chr.17: 4967739-4967739 on Build GRCh38
7332104 c.54C>T p.L18L Chr.17: 4967751-4967751 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs375724808 Chr.17: 4967672-4967672 on Build GRCh38
rs759100237 Chr.17: 4967799-4967799 on Build GRCh38
rs910352321 Chr.17: 4967783-4967783 on Build GRCh38
rs759411991 Chr.17: 4967974-4967974 on Build GRCh38
rs542593996 Chr.17: 4967880-4967880 on Build GRCh38
rs1023913367 Chr.17: 4967965-4967965 on Build GRCh38
rs1003541417 Chr.17: 4967690-4967690 on Build GRCh38
rs775928608 Chr.17: 4967810-4967810 on Build GRCh38
rs117836374 Chr.17: 4967993-4967993 on Build GRCh38
rs777906513 Chr.17: 4967691-4967691 on Build GRCh38
rs1035053524 Chr.17: 4967894-4967894 on Build GRCh38
rs747938106 Chr.17: 4967704-4967704 on Build GRCh38
rs748894036 Chr.17: 4967847-4967847 on Build GRCh38
rs779163281 Chr.17: 4967757-4967757 on Build GRCh38
rs533082203 Chr.17: 4968107-4968107 on Build GRCh38
rs1027367432 Chr.17: 4967870-4967870 on Build GRCh38
rs538844587 Chr.17: 4967934-4967934 on Build GRCh38
rs760071088 Chr.17: 4967742-4967742 on Build GRCh38
rs748451952 Chr.17: 4968021-4968021 on Build GRCh38
rs748946978 Chr.17: 4967795-4967795 on Build GRCh38
rs201057121 Chr.17: 4967679-4967679 on Build GRCh38
rs781027670 Chr.17: 4967770-4967770 on Build GRCh38
rs531921314 Chr.17: 4967867-4967867 on Build GRCh38
rs934303967 Chr.17: 4967825-4967825 on Build GRCh38
rs527939798 Chr.17: 4967976-4967976 on Build GRCh38
rs992382984 Chr.17: 4967964-4967964 on Build GRCh38
rs752587540 Chr.17: 4967842-4967842 on Build GRCh38
rs925343664 Chr.17: 4967997-4967997 on Build GRCh38
rs1015134949 Chr.17: 4967670-4967670 on Build GRCh38
rs3178201 Chr.17: 4968000-4968000 on Build GRCh38
rs944554485 Chr.17: 4968102-4968102 on Build GRCh38
rs563134740 Chr.17: 4967791-4967791 on Build GRCh38
rs373404400 Chr.17: 4967812-4967812 on Build GRCh38
rs774776318 Chr.17: 4967741-4967741 on Build GRCh38
rs1003616481 Chr.17: 4967892-4967892 on Build GRCh38
rs1045640642 Chr.17: 4967876-4967876 on Build GRCh38
rs764557632 Chr.17: 4967822-4967832 on Build GRCh38
rs946953743 Chr.17: 4967871-4967871 on Build GRCh38
rs986383569 Chr.17: 4967781-4967781 on Build GRCh38
rs369468423 Chr.17: 4967827-4967827 on Build GRCh38
rs767473110 Chr.17: 4967828-4967828 on Build GRCh38
rs770911339 Chr.17: 4967931-4967931 on Build GRCh38
rs944606632 Chr.17: 4967817-4967817 on Build GRCh38
rs562375792 Chr.17: 4967916-4967916 on Build GRCh38
rs368421421 Chr.17: 4967711-4967711 on Build GRCh38
rs754942977 Chr.17: 4967759-4967759 on Build GRCh38
rs775783314 Chr.17: 4967817-4967818 on Build GRCh38
rs979519720 Chr.17: 4967743-4967743 on Build GRCh38
rs990229337 Chr.17: 4968035-4968035 on Build GRCh38
rs867291045 Chr.17: 4967999-4967999 on Build GRCh38
rs988206297 Chr.17: 4968029-4968029 on Build GRCh38
rs975859899 Chr.17: 4967986-4967986 on Build GRCh38
rs747967498 Chr.17: 4967773-4967773 on Build GRCh38
rs760986079 Chr.17: 4967749-4967749 on Build GRCh38
rs577290737 Chr.17: 4967702-4967702 on Build GRCh38
rs962010565 Chr.17: 4967734-4967734 on Build GRCh38
rs762147440 Chr.17: 4967822-4967822 on Build GRCh38
rs780080741 Chr.17: 4967698-4967698 on Build GRCh38
rs951974942 Chr.17: 4967780-4967780 on Build GRCh38
rs772803800 Chr.17: 4967719-4967719 on Build GRCh38
rs776871352 Chr.17: 4967819-4967819 on Build GRCh38
rs1015082464 Chr.17: 4967940-4967940 on Build GRCh38
rs754068212 Chr.17: 4967754-4967754 on Build GRCh38
rs756385699 Chr.17: 4967683-4967683 on Build GRCh38
rs767825449 Chr.17: 4967745-4967745 on Build GRCh38
rs944441926 Chr.17: 4967794-4967794 on Build GRCh38
rs887532297 Chr.17: 4967918-4967918 on Build GRCh38
rs764369476 Chr.17: 4967813-4967813 on Build GRCh38
rs746946989 Chr.17: 4967699-4967699 on Build GRCh38
rs1055113531 Chr.17: 4968031-4968031 on Build GRCh38
rs750495497 Chr.17: 4967824-4967824 on Build GRCh38
rs765605663 Chr.17: 4967823-4967823 on Build GRCh38
rs914202082 Chr.17: 4968063-4968063 on Build GRCh38
rs960893800 Chr.17: 4967962-4967962 on Build GRCh38
rs965863175 Chr.17: 4967975-4967975 on Build GRCh38
rs770443620 Chr.17: 4967739-4967739 on Build GRCh38
rs545945620 Chr.17: 4967750-4967750 on Build GRCh38
rs996262039 Chr.17: 4967851-4967851 on Build GRCh38
rs373193757 Chr.17: 4967846-4967846 on Build GRCh38
rs993478727 Chr.17: 4967932-4967932 on Build GRCh38
rs986026239 Chr.17: 4967947-4967947 on Build GRCh38
rs761230671 Chr.17: 4967821-4967822 on Build GRCh38
rs946995116 Chr.17: 4968080-4968080 on Build GRCh38
rs934368040 Chr.17: 4968032-4968032 on Build GRCh38
rs570316133 Chr.17: 4968091-4968091 on Build GRCh38
rs753924067 Chr.17: 4967692-4967692 on Build GRCh38
rs769532271 Chr.17: 4967774-4967774 on Build GRCh38
rs768531172 Chr.17: 4967701-4967701 on Build GRCh38
rs369296491 Chr.17: 4967697-4967697 on Build GRCh38
rs952064758 Chr.17: 4967945-4967945 on Build GRCh38
rs773877623 Chr.17: 4967796-4967796 on Build GRCh38
rs772537470 Chr.17: 4967800-4967800 on Build GRCh38
rs753084609 Chr.17: 4967748-4967748 on Build GRCh38

View Full Product Details