Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4970254-4970764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMTA2
Entrez Gene ID
23125
Gene Name
calmodulin binding transcription activator 2
Gene Aliases
-
UniGene
Hs.632242
Gene Chromosome Location
Chr.17: 4967992-4988297 on Build GRCh38
Transcripts
NM_001171166.1 NM_001171167.1 NM_001171168.1 NM_015099.3 XM_006721478.3 XM_006721481.3 XM_006721482.3 XM_011523746.2 XM_011523747.2 XM_011523748.2 XM_011523749.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6195279 c.2923G>C p.E975Q Chr.17: 4970491-4970491 on Build GRCh38
1303035 c.2938G>C p.E980Q Chr.17: 4970407-4970407 on Build GRCh38
4067746 c.2921C>T p.P974L Chr.17: 4970493-4970493 on Build GRCh38
1247291 c.2839A>C p.I947L Chr.17: 4970506-4970506 on Build GRCh38
6894720 c.2940C>T p.L980L Chr.17: 4970411-4970411 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902214706 Chr.17: 4970723-4970723 on Build GRCh38
rs769089277 Chr.17: 4970372-4970372 on Build GRCh38
rs937845123 Chr.17: 4970615-4970615 on Build GRCh38
rs1030544013 Chr.17: 4970283-4970283 on Build GRCh38
rs1024193938 Chr.17: 4970657-4970657 on Build GRCh38
rs765108075 Chr.17: 4970493-4970493 on Build GRCh38
rs997859506 Chr.17: 4970725-4970725 on Build GRCh38
rs777406893 Chr.17: 4970292-4970293 on Build GRCh38
rs756514468 Chr.17: 4970461-4970461 on Build GRCh38
rs1045878144 Chr.17: 4970589-4970589 on Build GRCh38
rs761667523 Chr.17: 4970435-4970435 on Build GRCh38
rs1034355133 Chr.17: 4970610-4970610 on Build GRCh38
rs578203446 Chr.17: 4970569-4970569 on Build GRCh38
rs754591995 Chr.17: 4970530-4970530 on Build GRCh38
rs558284526 Chr.17: 4970562-4970562 on Build GRCh38
rs543675019 Chr.17: 4970665-4970665 on Build GRCh38
rs757988195 Chr.17: 4970355-4970355 on Build GRCh38
rs975152977 Chr.17: 4970728-4970728 on Build GRCh38
rs979186021 Chr.17: 4970394-4970394 on Build GRCh38
rs780355393 Chr.17: 4970359-4970359 on Build GRCh38
rs1014383393 Chr.17: 4970287-4970287 on Build GRCh38
rs138197927 Chr.17: 4970478-4970478 on Build GRCh38
rs202133009 Chr.17: 4970463-4970463 on Build GRCh38
rs897252940 Chr.17: 4970282-4970282 on Build GRCh38
rs542362968 Chr.17: 4970708-4970708 on Build GRCh38
rs992724944 Chr.17: 4970656-4970656 on Build GRCh38
rs773539959 Chr.17: 4970388-4970388 on Build GRCh38
rs17706762 Chr.17: 4970668-4970668 on Build GRCh38
rs764788058 Chr.17: 4970436-4970436 on Build GRCh38
rs1034031872 Chr.17: 4970403-4970403 on Build GRCh38
rs745454226 Chr.17: 4970547-4970549 on Build GRCh38
rs145978879 Chr.17: 4970467-4970467 on Build GRCh38
rs756394510 Chr.17: 4970542-4970542 on Build GRCh38
rs756884989 Chr.17: 4970343-4970343 on Build GRCh38
rs755509808 Chr.17: 4970452-4970452 on Build GRCh38
rs778224388 Chr.17: 4970548-4970548 on Build GRCh38
rs201487930 Chr.17: 4970533-4970533 on Build GRCh38
rs374770503 Chr.17: 4970570-4970570 on Build GRCh38
rs757802028 Chr.17: 4970440-4970440 on Build GRCh38
rs1010816485 Chr.17: 4970707-4970707 on Build GRCh38
rs769665885 Chr.17: 4970582-4970582 on Build GRCh38
rs371109567 Chr.17: 4970451-4970451 on Build GRCh38
rs149597137 Chr.17: 4970492-4970492 on Build GRCh38
rs747548041 Chr.17: 4970367-4970367 on Build GRCh38
rs1037028768 Chr.17: 4970532-4970532 on Build GRCh38
rs753601574 Chr.17: 4970434-4970434 on Build GRCh38
rs1020610514 Chr.17: 4970437-4970437 on Build GRCh38
rs142546361 Chr.17: 4970456-4970456 on Build GRCh38
rs1000248186 Chr.17: 4970591-4970591 on Build GRCh38
rs765934665 Chr.17: 4970450-4970450 on Build GRCh38
rs757763848 Chr.17: 4970678-4970678 on Build GRCh38
rs749665991 Chr.17: 4970550-4970550 on Build GRCh38
rs537322364 Chr.17: 4970431-4970431 on Build GRCh38
rs749555488 Chr.17: 4970472-4970472 on Build GRCh38
rs763907850 Chr.17: 4970331-4970331 on Build GRCh38
rs374249939 Chr.17: 4970516-4970516 on Build GRCh38
rs762884736 Chr.17: 4970407-4970407 on Build GRCh38
rs375211889 Chr.17: 4970504-4970504 on Build GRCh38
rs749900084 Chr.17: 4970354-4970354 on Build GRCh38
rs752694502 Chr.17: 4970309-4970309 on Build GRCh38
rs181795144 Chr.17: 4970673-4970673 on Build GRCh38
rs745939030 Chr.17: 4970568-4970568 on Build GRCh38
rs373840541 Chr.17: 4970344-4970344 on Build GRCh38
rs779176282 Chr.17: 4970567-4970567 on Build GRCh38
rs906337563 Chr.17: 4970593-4970593 on Build GRCh38
rs147242428 Chr.17: 4970551-4970551 on Build GRCh38
rs144421247 Chr.17: 4970525-4970525 on Build GRCh38
rs957687953 Chr.17: 4970406-4970406 on Build GRCh38
rs376425518 Chr.17: 4970378-4970378 on Build GRCh38
rs954851747 Chr.17: 4970288-4970288 on Build GRCh38
rs745990971 Chr.17: 4970487-4970487 on Build GRCh38
rs770869130 Chr.17: 4970476-4970476 on Build GRCh38
rs982611360 Chr.17: 4970468-4970468 on Build GRCh38
rs753261673 Chr.17: 4970540-4970540 on Build GRCh38
rs199749173 Chr.17: 4970384-4970384 on Build GRCh38
rs769901293 Chr.17: 4970387-4970387 on Build GRCh38
rs958683472 Chr.17: 4970632-4970632 on Build GRCh38
rs138067622 Chr.17: 4970430-4970430 on Build GRCh38
rs561288617 Chr.17: 4970455-4970455 on Build GRCh38
rs748914314 Chr.17: 4970320-4970320 on Build GRCh38
rs753441259 Chr.17: 4970332-4970332 on Build GRCh38
rs1010351359 Chr.17: 4970291-4970291 on Build GRCh38
rs771047614 Chr.17: 4970426-4970426 on Build GRCh38
rs772687225 Chr.17: 4970503-4970503 on Build GRCh38

View Full Product Details