Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7075491-7076007 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC10A
Entrez Gene ID
10462
Gene Name
C-type lectin domain family 10 member A
Gene Aliases
CD301, CLECSF13, CLECSF14, HML, HML2, MGL
UniGene
Hs.54403
Gene Chromosome Location
Chr.17: 7074537-7080635 on Build GRCh38
Transcripts
NM_006344.2 NM_182906.2 XM_005256412.1 XM_011523613.1 XM_011523614.2 XM_011523615.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5936284 c.500G>A p.W167* Chr.17: 7075825-7075825 on Build GRCh38
5933540 c.448G>A p.E150K Chr.17: 7075877-7075877 on Build GRCh38
6630080 c.500T>C p.V167A Chr.17: 7076005-7076005 on Build GRCh38
6630075 c.614C>A p.A205D Chr.17: 7075792-7075792 on Build GRCh38
1721713 c.559G>A p.E187K Chr.17: 7075847-7075847 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764011840 Chr.17: 7075746-7075746 on Build GRCh38
rs767754002 Chr.17: 7075567-7075567 on Build GRCh38
rs764796270 Chr.17: 7075921-7075921 on Build GRCh38
rs777397728 Chr.17: 7075731-7075731 on Build GRCh38
rs771486038 Chr.17: 7075626-7075626 on Build GRCh38
rs750011461 Chr.17: 7075924-7075924 on Build GRCh38
rs777313287 Chr.17: 7075614-7075614 on Build GRCh38
rs768972842 Chr.17: 7075660-7075660 on Build GRCh38
rs527762535 Chr.17: 7075767-7075767 on Build GRCh38
rs372609073 Chr.17: 7075736-7075736 on Build GRCh38
rs776921832 Chr.17: 7075664-7075664 on Build GRCh38
rs752411619 Chr.17: 7075716-7075716 on Build GRCh38
rs749873524 Chr.17: 7075855-7075855 on Build GRCh38
rs368232903 Chr.17: 7075698-7075698 on Build GRCh38
rs753521406 Chr.17: 7075834-7075834 on Build GRCh38
rs364569 Chr.17: 7075860-7075860 on Build GRCh38
rs772518823 Chr.17: 7075647-7075647 on Build GRCh38
rs760477667 Chr.17: 7075825-7075825 on Build GRCh38
rs769054359 Chr.17: 7075795-7075795 on Build GRCh38
rs746296487 Chr.17: 7075646-7075646 on Build GRCh38
rs3830425 Chr.17: 7075900-7075904 on Build GRCh38
rs535678341 Chr.17: 7075862-7075862 on Build GRCh38
rs779487769 Chr.17: 7075638-7075638 on Build GRCh38
rs955585888 Chr.17: 7075880-7075880 on Build GRCh38
rs753810962 Chr.17: 7075772-7075773 on Build GRCh38
rs145297270 Chr.17: 7075515-7075515 on Build GRCh38
rs776390757 Chr.17: 7075912-7075912 on Build GRCh38
rs760524840 Chr.17: 7075679-7075679 on Build GRCh38
rs772267288 Chr.17: 7075524-7075524 on Build GRCh38
rs748765114 Chr.17: 7075623-7075623 on Build GRCh38
rs546045571 Chr.17: 7075593-7075593 on Build GRCh38
rs752424354 Chr.17: 7075605-7075605 on Build GRCh38
rs762387836 Chr.17: 7075955-7075955 on Build GRCh38
rs888826704 Chr.17: 7075562-7075562 on Build GRCh38
rs181487113 Chr.17: 7075961-7075961 on Build GRCh38
rs146827384 Chr.17: 7075850-7075850 on Build GRCh38
rs763899730 Chr.17: 7075906-7075906 on Build GRCh38
rs751312500 Chr.17: 7075905-7075907 on Build GRCh38
rs541748001 Chr.17: 7075779-7075779 on Build GRCh38
rs149367958 Chr.17: 7075685-7075685 on Build GRCh38
rs555943706 Chr.17: 7075600-7075600 on Build GRCh38
rs796266016 Chr.17: 7075905-7075909 on Build GRCh38
rs753947899 Chr.17: 7075547-7075547 on Build GRCh38
rs757920247 Chr.17: 7075902-7075904 on Build GRCh38
rs763027849 Chr.17: 7075548-7075548 on Build GRCh38
rs138348118 Chr.17: 7075750-7075750 on Build GRCh38
rs531762386 Chr.17: 7075694-7075694 on Build GRCh38
rs775787119 Chr.17: 7075532-7075532 on Build GRCh38
rs200740525 Chr.17: 7075870-7075870 on Build GRCh38
rs775410503 Chr.17: 7075885-7075885 on Build GRCh38
rs756764711 Chr.17: 7075734-7075734 on Build GRCh38
rs377384027 Chr.17: 7075796-7075796 on Build GRCh38
rs201601339 Chr.17: 7075890-7075890 on Build GRCh38
rs775422957 Chr.17: 7075808-7075808 on Build GRCh38
rs765844461 Chr.17: 7075960-7075960 on Build GRCh38
rs79945463 Chr.17: 7075797-7075797 on Build GRCh38
rs765345852 Chr.17: 7075681-7075681 on Build GRCh38
rs200068112 Chr.17: 7075918-7075918 on Build GRCh38
rs574360159 Chr.17: 7075676-7075676 on Build GRCh38
rs764295074 Chr.17: 7075539-7075539 on Build GRCh38
rs755711960 Chr.17: 7075606-7075606 on Build GRCh38
rs373310764 Chr.17: 7075972-7075972 on Build GRCh38
rs376457612 Chr.17: 7075930-7075930 on Build GRCh38
rs764598849 Chr.17: 7075860-7075860 on Build GRCh38
rs942817482 Chr.17: 7075901-7075901 on Build GRCh38
rs780416542 Chr.17: 7075859-7075859 on Build GRCh38
rs761287121 Chr.17: 7075854-7075855 on Build GRCh38
rs943077516 Chr.17: 7075937-7075937 on Build GRCh38
rs753431237 Chr.17: 7075733-7075733 on Build GRCh38
rs766411547 Chr.17: 7075552-7075552 on Build GRCh38
rs1003094045 Chr.17: 7075747-7075747 on Build GRCh38
rs370695862 Chr.17: 7075529-7075529 on Build GRCh38
rs747376569 Chr.17: 7075772-7075772 on Build GRCh38
rs759014817 Chr.17: 7075856-7075856 on Build GRCh38
rs763810382 Chr.17: 7075828-7075828 on Build GRCh38
rs779768277 Chr.17: 7075943-7075944 on Build GRCh38
rs755158327 Chr.17: 7075555-7075555 on Build GRCh38
rs761029379 Chr.17: 7075659-7075659 on Build GRCh38
rs748400637 Chr.17: 7075869-7075869 on Build GRCh38
rs143134980 Chr.17: 7075884-7075884 on Build GRCh38
rs758811568 Chr.17: 7075766-7075766 on Build GRCh38
rs556452972 Chr.17: 7075806-7075806 on Build GRCh38
rs199687112 Chr.17: 7075722-7075722 on Build GRCh38
rs781722434 Chr.17: 7075973-7075973 on Build GRCh38
rs545560286 Chr.17: 7075697-7075697 on Build GRCh38
rs532880398 Chr.17: 7075535-7075535 on Build GRCh38
rs35101468 Chr.17: 7075798-7075798 on Build GRCh38
rs373819182 Chr.17: 7075653-7075653 on Build GRCh38
rs764854106 Chr.17: 7075852-7075852 on Build GRCh38
rs755394592 Chr.17: 7075861-7075861 on Build GRCh38
rs775671103 Chr.17: 7075649-7075649 on Build GRCh38
rs199833742 Chr.17: 7075872-7075872 on Build GRCh38

View Full Product Details