Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7079960-7080470 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC10A
Entrez Gene ID
10462
Gene Name
C-type lectin domain family 10 member A
Gene Aliases
CD301, CLECSF13, CLECSF14, HML, HML2, MGL
UniGene
Hs.54403
Gene Chromosome Location
Chr.17: 7074537-7080635 on Build GRCh38
Transcripts
NM_006344.2 NM_182906.2 XM_005256412.1 XM_011523613.1 XM_011523614.2 XM_011523615.1

SNP Detail

SNP ID Chromosome Location
rs886292450 Chr.17: 7080302-7080302 on Build GRCh38
rs55835902 Chr.17: 7080148-7080148 on Build GRCh38
rs184242724 Chr.17: 7080137-7080137 on Build GRCh38
rs189052490 Chr.17: 7080175-7080175 on Build GRCh38
rs1016367420 Chr.17: 7079988-7079988 on Build GRCh38
rs758862917 Chr.17: 7080357-7080357 on Build GRCh38
rs989349576 Chr.17: 7080420-7080420 on Build GRCh38
rs369102083 Chr.17: 7080082-7080082 on Build GRCh38
rs8075065 Chr.17: 7080031-7080031 on Build GRCh38
rs11657498 Chr.17: 7079996-7079996 on Build GRCh38
rs181142956 Chr.17: 7080179-7080179 on Build GRCh38
rs1046265861 Chr.17: 7080040-7080040 on Build GRCh38
rs945133911 Chr.17: 7079983-7079983 on Build GRCh38
rs949338973 Chr.17: 7080283-7080283 on Build GRCh38
rs1015579756 Chr.17: 7080081-7080081 on Build GRCh38
rs193271222 Chr.17: 7080133-7080133 on Build GRCh38
rs561663326 Chr.17: 7080069-7080069 on Build GRCh38
rs1019719443 Chr.17: 7080441-7080441 on Build GRCh38
rs766464579 Chr.17: 7080032-7080032 on Build GRCh38
rs547221353 Chr.17: 7080103-7080103 on Build GRCh38
rs575282437 Chr.17: 7080054-7080054 on Build GRCh38
rs77445845 Chr.17: 7080364-7080364 on Build GRCh38
rs761985462 Chr.17: 7080217-7080217 on Build GRCh38
rs1004314105 Chr.17: 7080071-7080071 on Build GRCh38
rs957986897 Chr.17: 7080383-7080383 on Build GRCh38
rs1006366738 Chr.17: 7079986-7079986 on Build GRCh38
rs554651252 Chr.17: 7080167-7080167 on Build GRCh38
rs376150995 Chr.17: 7080051-7080051 on Build GRCh38
rs924966151 Chr.17: 7080014-7080014 on Build GRCh38
rs145466410 Chr.17: 7080038-7080038 on Build GRCh38
rs73242048 Chr.17: 7080186-7080186 on Build GRCh38
rs1037148649 Chr.17: 7080359-7080359 on Build GRCh38
rs777241736 Chr.17: 7080239-7080239 on Build GRCh38
rs374789476 Chr.17: 7080362-7080362 on Build GRCh38
rs148814818 Chr.17: 7080114-7080114 on Build GRCh38
rs1025009327 Chr.17: 7080033-7080033 on Build GRCh38
rs190232137 Chr.17: 7080083-7080083 on Build GRCh38
rs989233830 Chr.17: 7080094-7080094 on Build GRCh38
rs540002663 Chr.17: 7080095-7080095 on Build GRCh38
rs1037915188 Chr.17: 7080292-7080292 on Build GRCh38
rs150102632 Chr.17: 7080012-7080012 on Build GRCh38
rs529740309 Chr.17: 7080304-7080304 on Build GRCh38
rs976892188 Chr.17: 7080158-7080158 on Build GRCh38
rs980680179 Chr.17: 7080059-7080059 on Build GRCh38
rs1008450302 Chr.17: 7080409-7080409 on Build GRCh38
rs11653290 Chr.17: 7080210-7080210 on Build GRCh38
rs890003949 Chr.17: 7080382-7080382 on Build GRCh38
rs552222889 Chr.17: 7080144-7080144 on Build GRCh38
rs538266659 Chr.17: 7080154-7080154 on Build GRCh38
rs993626591 Chr.17: 7080323-7080323 on Build GRCh38
rs889339099 Chr.17: 7080215-7080215 on Build GRCh38
rs1046234367 Chr.17: 7080291-7080291 on Build GRCh38
rs936366038 Chr.17: 7080193-7080193 on Build GRCh38
rs867688986 Chr.17: 7080361-7080361 on Build GRCh38

View Full Product Details