Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28236642-28237138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATPIF1
Entrez Gene ID
93974
Gene Name
ATPase inhibitory factor 1
Gene Aliases
ATPI, ATPIP, IP
UniGene
Hs.744914
Gene Chromosome Location
Chr.1: 28236091-28238105 on Build GRCh38
Transcripts
NM_016311.4 NM_178190.2 NM_178191.2

SNP Detail

SNP ID Chromosome Location
rs987169053 Chr.1: 28236835-28236835 on Build GRCh38
rs1057105723 Chr.1: 28236941-28236941 on Build GRCh38
rs984953734 Chr.1: 28236758-28236758 on Build GRCh38
rs1026751561 Chr.1: 28237057-28237057 on Build GRCh38
rs993573278 Chr.1: 28236738-28236738 on Build GRCh38
rs116668731 Chr.1: 28236887-28236887 on Build GRCh38
rs1026364876 Chr.1: 28236743-28236743 on Build GRCh38
rs187383524 Chr.1: 28236802-28236802 on Build GRCh38
rs878909098 Chr.1: 28236691-28236691 on Build GRCh38
rs968191144 Chr.1: 28237073-28237073 on Build GRCh38
rs955678854 Chr.1: 28236834-28236834 on Build GRCh38
rs1044641762 Chr.1: 28236666-28236666 on Build GRCh38
rs189347730 Chr.1: 28236989-28236989 on Build GRCh38
rs886558094 Chr.1: 28236936-28236936 on Build GRCh38
rs768397263 Chr.1: 28236714-28236714 on Build GRCh38
rs1002789144 Chr.1: 28236927-28236927 on Build GRCh38
rs524651 Chr.1: 28236751-28236751 on Build GRCh38
rs1056331802 Chr.1: 28236700-28236700 on Build GRCh38
rs895355845 Chr.1: 28237030-28237030 on Build GRCh38
rs924381021 Chr.1: 28236813-28236813 on Build GRCh38
rs968846184 Chr.1: 28236774-28236774 on Build GRCh38
rs978527547 Chr.1: 28236776-28236776 on Build GRCh38
rs1035001180 Chr.1: 28236724-28236724 on Build GRCh38
rs964917112 Chr.1: 28236812-28236812 on Build GRCh38
rs867209886 Chr.1: 28236993-28236993 on Build GRCh38
rs922936733 Chr.1: 28236850-28236850 on Build GRCh38
rs559928478 Chr.1: 28236904-28236904 on Build GRCh38
rs117855698 Chr.1: 28236880-28236880 on Build GRCh38
rs1003494926 Chr.1: 28236945-28236945 on Build GRCh38
rs1012515257 Chr.1: 28237042-28237042 on Build GRCh38
rs960737907 Chr.1: 28236733-28236733 on Build GRCh38
rs143516972 Chr.1: 28237064-28237064 on Build GRCh38
rs897082961 Chr.1: 28236956-28236956 on Build GRCh38
rs905752324 Chr.1: 28236684-28236684 on Build GRCh38
rs537633761 Chr.1: 28237056-28237056 on Build GRCh38
rs559770 Chr.1: 28236754-28236754 on Build GRCh38
rs910722576 Chr.1: 28236799-28236799 on Build GRCh38
rs1011371650 Chr.1: 28236665-28236665 on Build GRCh38
rs1039250154 Chr.1: 28236864-28236864 on Build GRCh38
rs11580943 Chr.1: 28236895-28236895 on Build GRCh38
rs528684879 Chr.1: 28236965-28236965 on Build GRCh38
rs976326721 Chr.1: 28236838-28236838 on Build GRCh38
rs182237786 Chr.1: 28236664-28236664 on Build GRCh38
rs952229898 Chr.1: 28236756-28236756 on Build GRCh38
rs914305394 Chr.1: 28236873-28236873 on Build GRCh38
rs939192473 Chr.1: 28236696-28236696 on Build GRCh38
rs1053204354 Chr.1: 28236861-28236861 on Build GRCh38
rs1047789948 Chr.1: 28236910-28236910 on Build GRCh38
rs879229588 Chr.1: 28236852-28236852 on Build GRCh38

View Full Product Details