Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28237413-28237924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATPIF1
Entrez Gene ID
93974
Gene Name
ATPase inhibitory factor 1
Gene Aliases
ATPI, ATPIP, IP
UniGene
Hs.744914
Gene Chromosome Location
Chr.1: 28236091-28238105 on Build GRCh38
Transcripts
NM_016311.4 NM_178190.2 NM_178191.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6063078 c.244A>C p.K82Q Chr.1: 28237901-28237901 on Build GRCh38
6063077 c.246G>T p.K82N Chr.1: 28237903-28237903 on Build GRCh38
5955052 c.193G>A p.E65K Chr.1: 28237850-28237850 on Build GRCh38
6608459 c.235G>A p.V79I Chr.1: 28237892-28237892 on Build GRCh38
6608456 c.211delA p.K72fs*31 Chr.1: 28237868-28237868 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs938431326 Chr.1: 28237580-28237580 on Build GRCh38
rs1008665552 Chr.1: 28237488-28237488 on Build GRCh38
rs766752378 Chr.1: 28237728-28237728 on Build GRCh38
rs533357966 Chr.1: 28237548-28237548 on Build GRCh38
rs757140062 Chr.1: 28237738-28237738 on Build GRCh38
rs752878076 Chr.1: 28237736-28237736 on Build GRCh38
rs948073072 Chr.1: 28237655-28237655 on Build GRCh38
rs527740178 Chr.1: 28237478-28237478 on Build GRCh38
rs113261974 Chr.1: 28237834-28237834 on Build GRCh38
rs769226866 Chr.1: 28237634-28237634 on Build GRCh38
rs777602793 Chr.1: 28237698-28237700 on Build GRCh38
rs141948364 Chr.1: 28237769-28237769 on Build GRCh38
rs774827581 Chr.1: 28237812-28237812 on Build GRCh38
rs1031182458 Chr.1: 28237458-28237458 on Build GRCh38
rs918452391 Chr.1: 28237592-28237592 on Build GRCh38
rs1048829751 Chr.1: 28237608-28237608 on Build GRCh38
rs751735945 Chr.1: 28237731-28237731 on Build GRCh38
rs964394396 Chr.1: 28237517-28237518 on Build GRCh38
rs140763065 Chr.1: 28237445-28237445 on Build GRCh38
rs763478808 Chr.1: 28237800-28237800 on Build GRCh38
rs935581566 Chr.1: 28237737-28237737 on Build GRCh38
rs8559 Chr.1: 28237741-28237741 on Build GRCh38
rs773657021 Chr.1: 28237795-28237795 on Build GRCh38
rs769304298 Chr.1: 28237792-28237792 on Build GRCh38
rs979801388 Chr.1: 28237522-28237522 on Build GRCh38
rs780027111 Chr.1: 28237767-28237767 on Build GRCh38
rs759662061 Chr.1: 28237824-28237824 on Build GRCh38
rs533689697 Chr.1: 28237801-28237801 on Build GRCh38
rs1027287407 Chr.1: 28237898-28237898 on Build GRCh38
rs751431301 Chr.1: 28237899-28237899 on Build GRCh38
rs1039011263 Chr.1: 28237680-28237680 on Build GRCh38
rs767011179 Chr.1: 28237874-28237876 on Build GRCh38
rs760640267 Chr.1: 28237720-28237720 on Build GRCh38
rs568776345 Chr.1: 28237648-28237648 on Build GRCh38
rs747624396 Chr.1: 28237774-28237774 on Build GRCh38
rs767469518 Chr.1: 28237733-28237733 on Build GRCh38
rs745395666 Chr.1: 28237632-28237632 on Build GRCh38
rs939416924 Chr.1: 28237606-28237606 on Build GRCh38
rs904070877 Chr.1: 28237722-28237722 on Build GRCh38
rs778930273 Chr.1: 28237739-28237739 on Build GRCh38
rs541425565 Chr.1: 28237489-28237489 on Build GRCh38
rs112006347 Chr.1: 28237835-28237835 on Build GRCh38
rs780773256 Chr.1: 28237584-28237584 on Build GRCh38
rs758222441 Chr.1: 28237765-28237765 on Build GRCh38
rs760751368 Chr.1: 28237865-28237865 on Build GRCh38
rs1030472897 Chr.1: 28237460-28237460 on Build GRCh38
rs1027523774 Chr.1: 28237544-28237544 on Build GRCh38
rs997599595 Chr.1: 28237450-28237450 on Build GRCh38
rs369695947 Chr.1: 28237715-28237715 on Build GRCh38
rs376753470 Chr.1: 28237702-28237702 on Build GRCh38
rs900567898 Chr.1: 28237683-28237683 on Build GRCh38
rs879748592 Chr.1: 28237876-28237876 on Build GRCh38
rs997588141 Chr.1: 28237706-28237706 on Build GRCh38
rs201956032 Chr.1: 28237897-28237897 on Build GRCh38
rs758401235 Chr.1: 28237892-28237892 on Build GRCh38
rs575585835 Chr.1: 28237518-28237518 on Build GRCh38
rs763390750 Chr.1: 28237833-28237844 on Build GRCh38
rs929889934 Chr.1: 28237594-28237594 on Build GRCh38
rs968824834 Chr.1: 28237513-28237513 on Build GRCh38
rs770600600 Chr.1: 28237568-28237568 on Build GRCh38
rs552218002 Chr.1: 28237637-28237637 on Build GRCh38
rs951938160 Chr.1: 28237900-28237900 on Build GRCh38
rs764292791 Chr.1: 28237884-28237884 on Build GRCh38
rs570378650 Chr.1: 28237845-28237845 on Build GRCh38
rs995815918 Chr.1: 28237891-28237891 on Build GRCh38
rs759838729 Chr.1: 28237732-28237732 on Build GRCh38
rs368919521 Chr.1: 28237825-28237825 on Build GRCh38
rs955745616 Chr.1: 28237470-28237470 on Build GRCh38
rs956280832 Chr.1: 28237468-28237468 on Build GRCh38
rs752869401 Chr.1: 28237846-28237846 on Build GRCh38
rs1034029245 Chr.1: 28237817-28237817 on Build GRCh38
rs9508 Chr.1: 28237768-28237768 on Build GRCh38
rs1008839767 Chr.1: 28237789-28237789 on Build GRCh38
rs1021290557 Chr.1: 28237495-28237495 on Build GRCh38
rs941990529 Chr.1: 28237641-28237641 on Build GRCh38
rs564365688 Chr.1: 28237526-28237526 on Build GRCh38

View Full Product Details