Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32013803-32014302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KHDRBS1
Entrez Gene ID
10657
Gene Name
KH RNA binding domain containing, signal transduction associated 1
Gene Aliases
Sam68, p62, p68
UniGene
Hs.445893
Gene Chromosome Location
Chr.1: 32013694-32060859 on Build GRCh38
Transcripts
NM_001271878.1 NM_006559.2 NR_073498.1 NR_073499.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4633794 c.29G>A p.R10H Chr.1: 32014024-32014024 on Build GRCh38
6371072 c.197T>C p.L66P Chr.1: 32014192-32014192 on Build GRCh38
4772345 c.65A>C p.D22A Chr.1: 32014060-32014060 on Build GRCh38
5807257 c.258G>T p.L86L Chr.1: 32014253-32014253 on Build GRCh38
4645160 c.127C>T p.R43W Chr.1: 32014122-32014122 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752713376 Chr.1: 32014037-32014037 on Build GRCh38
rs951207345 Chr.1: 32013880-32013880 on Build GRCh38
rs1006416638 Chr.1: 32014161-32014161 on Build GRCh38
rs947210162 Chr.1: 32013825-32013825 on Build GRCh38
rs996023411 Chr.1: 32014124-32014124 on Build GRCh38
rs1035775809 Chr.1: 32013886-32013886 on Build GRCh38
rs912579924 Chr.1: 32013991-32013991 on Build GRCh38
rs944005169 Chr.1: 32014026-32014026 on Build GRCh38
rs867196638 Chr.1: 32014276-32014276 on Build GRCh38
rs551108017 Chr.1: 32014262-32014262 on Build GRCh38
rs982518354 Chr.1: 32013885-32013885 on Build GRCh38
rs754077786 Chr.1: 32014049-32014049 on Build GRCh38
rs780943655 Chr.1: 32014097-32014097 on Build GRCh38
rs540743924 Chr.1: 32013887-32013887 on Build GRCh38
rs779236077 Chr.1: 32013947-32013947 on Build GRCh38
rs769652619 Chr.1: 32013988-32013988 on Build GRCh38
rs530783649 Chr.1: 32013984-32013984 on Build GRCh38
rs918062032 Chr.1: 32014217-32014217 on Build GRCh38
rs747814006 Chr.1: 32014109-32014109 on Build GRCh38
rs1004387150 Chr.1: 32014110-32014110 on Build GRCh38
rs1022548026 Chr.1: 32014166-32014166 on Build GRCh38
rs6667294 Chr.1: 32013951-32013951 on Build GRCh38
rs766284175 Chr.1: 32014028-32014028 on Build GRCh38
rs544360586 Chr.1: 32014189-32014189 on Build GRCh38
rs1045035277 Chr.1: 32013865-32013865 on Build GRCh38
rs528464512 Chr.1: 32014228-32014228 on Build GRCh38
rs891610883 Chr.1: 32013892-32013892 on Build GRCh38
rs760696661 Chr.1: 32014236-32014236 on Build GRCh38
rs772931806 Chr.1: 32014016-32014016 on Build GRCh38
rs936680295 Chr.1: 32014261-32014261 on Build GRCh38
rs773057128 Chr.1: 32014202-32014202 on Build GRCh38
rs757465968 Chr.1: 32014053-32014053 on Build GRCh38
rs373256621 Chr.1: 32014235-32014235 on Build GRCh38
rs750816164 Chr.1: 32014062-32014062 on Build GRCh38
rs746456822 Chr.1: 32014077-32014077 on Build GRCh38
rs1012535544 Chr.1: 32014159-32014159 on Build GRCh38
rs896521286 Chr.1: 32013841-32013841 on Build GRCh38
rs76080682 Chr.1: 32013930-32013930 on Build GRCh38
rs918176208 Chr.1: 32014019-32014019 on Build GRCh38
rs968561946 Chr.1: 32013923-32013923 on Build GRCh38
rs772493181 Chr.1: 32013961-32013961 on Build GRCh38
rs1052875512 Chr.1: 32013831-32013831 on Build GRCh38
rs768370132 Chr.1: 32013969-32013969 on Build GRCh38
rs568532115 Chr.1: 32014247-32014247 on Build GRCh38
rs746579121 Chr.1: 32013963-32013963 on Build GRCh38
rs995718490 Chr.1: 32014067-32014067 on Build GRCh38
rs961750842 Chr.1: 32014200-32014200 on Build GRCh38
rs900028415 Chr.1: 32013861-32013861 on Build GRCh38
rs891666290 Chr.1: 32014076-32014076 on Build GRCh38
rs535621173 Chr.1: 32014260-32014260 on Build GRCh38
rs375777010 Chr.1: 32014266-32014266 on Build GRCh38
rs956163539 Chr.1: 32013920-32013920 on Build GRCh38
rs776431312 Chr.1: 32013982-32013982 on Build GRCh38
rs901779668 Chr.1: 32014113-32014113 on Build GRCh38
rs1014672870 Chr.1: 32014190-32014190 on Build GRCh38
rs960009019 Chr.1: 32014147-32014147 on Build GRCh38
rs760747849 Chr.1: 32014039-32014039 on Build GRCh38
rs1052519460 Chr.1: 32014074-32014074 on Build GRCh38
rs574486117 Chr.1: 32013921-32013921 on Build GRCh38
rs750743983 Chr.1: 32014264-32014264 on Build GRCh38
rs986040812 Chr.1: 32014145-32014145 on Build GRCh38
rs542002614 Chr.1: 32013925-32013925 on Build GRCh38
rs757900703 Chr.1: 32013856-32013856 on Build GRCh38
rs951904057 Chr.1: 32013942-32013942 on Build GRCh38
rs912637857 Chr.1: 32014250-32014250 on Build GRCh38
rs1029097851 Chr.1: 32014128-32014128 on Build GRCh38
rs868111502 Chr.1: 32013845-32013845 on Build GRCh38
rs764353405 Chr.1: 32013947-32013957 on Build GRCh38
rs757671366 Chr.1: 32013945-32013945 on Build GRCh38
rs369015624 Chr.1: 32014068-32014068 on Build GRCh38
rs762917162 Chr.1: 32014024-32014024 on Build GRCh38
rs903934148 Chr.1: 32013829-32013829 on Build GRCh38
rs903944179 Chr.1: 32013866-32013866 on Build GRCh38
rs867892253 Chr.1: 32014252-32014252 on Build GRCh38
rs935392778 Chr.1: 32013830-32013830 on Build GRCh38
rs779302687 Chr.1: 32014057-32014057 on Build GRCh38
rs995663670 Chr.1: 32013871-32013871 on Build GRCh38
rs992335624 Chr.1: 32014215-32014215 on Build GRCh38
rs12746359 Chr.1: 32014036-32014036 on Build GRCh38
rs765746078 Chr.1: 32013874-32013874 on Build GRCh38
rs764259849 Chr.1: 32014041-32014041 on Build GRCh38
rs936777260 Chr.1: 32014061-32014061 on Build GRCh38
rs981028991 Chr.1: 32014240-32014240 on Build GRCh38
rs764132945 Chr.1: 32014239-32014239 on Build GRCh38
rs1056718089 Chr.1: 32013836-32013836 on Build GRCh38
rs991394559 Chr.1: 32013903-32013903 on Build GRCh38
rs201069205 Chr.1: 32014274-32014274 on Build GRCh38
rs367965800 Chr.1: 32013889-32013889 on Build GRCh38
rs768317064 Chr.1: 32014085-32014085 on Build GRCh38
rs965725793 Chr.1: 32013905-32013905 on Build GRCh38
rs758697117 Chr.1: 32014273-32014273 on Build GRCh38
rs762667767 Chr.1: 32014204-32014204 on Build GRCh38
rs1043823754 Chr.1: 32013827-32013827 on Build GRCh38
rs112917271 Chr.1: 32014130-32014130 on Build GRCh38
rs540375364 Chr.1: 32013822-32013822 on Build GRCh38
rs891100704 Chr.1: 32013848-32013848 on Build GRCh38
rs924437024 Chr.1: 32013941-32013941 on Build GRCh38
rs923072413 Chr.1: 32014075-32014075 on Build GRCh38
rs925306912 Chr.1: 32014249-32014249 on Build GRCh38
rs1018518241 Chr.1: 32013859-32013859 on Build GRCh38
rs762086680 Chr.1: 32014251-32014251 on Build GRCh38

View Full Product Details