Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32039391-32039910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KHDRBS1
Entrez Gene ID
10657
Gene Name
KH RNA binding domain containing, signal transduction associated 1
Gene Aliases
Sam68, p62, p68
UniGene
Hs.445893
Gene Chromosome Location
Chr.1: 32013694-32060859 on Build GRCh38
Transcripts
NM_001271878.1 NM_006559.2 NR_073498.1 NR_073499.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3741224 c.1196A>G p.Y399C Chr.1: 32039535-32039535 on Build GRCh38
4695090 c.1220C>T p.S407F Chr.1: 32039559-32039559 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1047141118 Chr.1: 32039740-32039740 on Build GRCh38
rs559805039 Chr.1: 32039821-32039821 on Build GRCh38
rs760141128 Chr.1: 32039495-32039495 on Build GRCh38
rs894370273 Chr.1: 32039799-32039799 on Build GRCh38
rs184106170 Chr.1: 32039807-32039807 on Build GRCh38
rs771284439 Chr.1: 32039420-32039420 on Build GRCh38
rs924073355 Chr.1: 32039583-32039583 on Build GRCh38
rs796318294 Chr.1: 32039481-32039482 on Build GRCh38
rs1021680155 Chr.1: 32039818-32039818 on Build GRCh38
rs942996736 Chr.1: 32039643-32039643 on Build GRCh38
rs148435661 Chr.1: 32039885-32039885 on Build GRCh38
rs561023203 Chr.1: 32039586-32039586 on Build GRCh38
rs911458073 Chr.1: 32039642-32039642 on Build GRCh38
rs780059501 Chr.1: 32039437-32039437 on Build GRCh38
rs898512148 Chr.1: 32039715-32039715 on Build GRCh38
rs907005043 Chr.1: 32039753-32039753 on Build GRCh38
rs1002658559 Chr.1: 32039755-32039755 on Build GRCh38
rs766909339 Chr.1: 32039496-32039496 on Build GRCh38
rs371475604 Chr.1: 32039590-32039590 on Build GRCh38
rs977131368 Chr.1: 32039831-32039831 on Build GRCh38
rs752306210 Chr.1: 32039498-32039498 on Build GRCh38
rs753586881 Chr.1: 32039588-32039588 on Build GRCh38
rs543826862 Chr.1: 32039670-32039670 on Build GRCh38
rs977183532 Chr.1: 32039443-32039443 on Build GRCh38
rs779559794 Chr.1: 32039474-32039474 on Build GRCh38
rs368454525 Chr.1: 32039444-32039444 on Build GRCh38
rs1034180124 Chr.1: 32039784-32039784 on Build GRCh38
rs934151825 Chr.1: 32039607-32039607 on Build GRCh38
rs149623609 Chr.1: 32039617-32039617 on Build GRCh38
rs994182055 Chr.1: 32039731-32039731 on Build GRCh38
rs752139265 Chr.1: 32039486-32039486 on Build GRCh38
rs77744758 Chr.1: 32039862-32039862 on Build GRCh38
rs777403559 Chr.1: 32039562-32039562 on Build GRCh38
rs760025225 Chr.1: 32039680-32039680 on Build GRCh38
rs561332951 Chr.1: 32039819-32039819 on Build GRCh38
rs548809920 Chr.1: 32039866-32039866 on Build GRCh38
rs746616601 Chr.1: 32039548-32039548 on Build GRCh38
rs1027396473 Chr.1: 32039874-32039875 on Build GRCh38
rs924067956 Chr.1: 32039669-32039669 on Build GRCh38
rs551653711 Chr.1: 32039655-32039655 on Build GRCh38
rs532574526 Chr.1: 32039785-32039785 on Build GRCh38
rs745759660 Chr.1: 32039598-32039598 on Build GRCh38
rs775686856 Chr.1: 32039423-32039423 on Build GRCh38
rs41263985 Chr.1: 32039546-32039546 on Build GRCh38
rs755661820 Chr.1: 32039518-32039518 on Build GRCh38
rs778728383 Chr.1: 32039593-32039593 on Build GRCh38
rs759187707 Chr.1: 32039484-32039484 on Build GRCh38
rs1030085605 Chr.1: 32039872-32039872 on Build GRCh38
rs565309951 Chr.1: 32039671-32039671 on Build GRCh38
rs763416113 Chr.1: 32039471-32039471 on Build GRCh38
rs772112549 Chr.1: 32039616-32039616 on Build GRCh38
rs890352546 Chr.1: 32039746-32039746 on Build GRCh38

View Full Product Details