Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 29566000-29566498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD15
Entrez Gene ID
116236
Gene Name
abhydrolase domain containing 15
Gene Aliases
-
UniGene
Hs.106510
Gene Chromosome Location
Chr.17: 29560661-29567024 on Build GRCh38
Transcripts
NM_198147.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4886276 c.672C>A p.F224L Chr.17: 29566295-29566295 on Build GRCh38
3766056 c.816C>T p.F272F Chr.17: 29566151-29566151 on Build GRCh38
3795414 c.878G>T p.S293I Chr.17: 29566089-29566089 on Build GRCh38
3795415 c.716G>C p.G239A Chr.17: 29566251-29566251 on Build GRCh38
5385960 c.744C>T p.G248G Chr.17: 29566223-29566223 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201682778 Chr.17: 29566229-29566229 on Build GRCh38
rs367565913 Chr.17: 29566228-29566228 on Build GRCh38
rs772192321 Chr.17: 29566253-29566253 on Build GRCh38
rs770149003 Chr.17: 29566372-29566372 on Build GRCh38
rs758605343 Chr.17: 29566327-29566327 on Build GRCh38
rs528883814 Chr.17: 29566286-29566286 on Build GRCh38
rs759609923 Chr.17: 29566395-29566395 on Build GRCh38
rs757452693 Chr.17: 29566294-29566294 on Build GRCh38
rs777088711 Chr.17: 29566271-29566271 on Build GRCh38
rs761016582 Chr.17: 29566140-29566140 on Build GRCh38
rs376693483 Chr.17: 29566295-29566295 on Build GRCh38
rs769028784 Chr.17: 29566452-29566452 on Build GRCh38
rs770347389 Chr.17: 29566441-29566441 on Build GRCh38
rs748267420 Chr.17: 29566127-29566127 on Build GRCh38
rs777887363 Chr.17: 29566420-29566420 on Build GRCh38
rs763721775 Chr.17: 29566037-29566037 on Build GRCh38
rs747097784 Chr.17: 29566332-29566332 on Build GRCh38
rs749963392 Chr.17: 29566289-29566289 on Build GRCh38
rs62070805 Chr.17: 29566363-29566363 on Build GRCh38
rs773779279 Chr.17: 29566131-29566131 on Build GRCh38
rs777037440 Chr.17: 29566163-29566163 on Build GRCh38
rs751175217 Chr.17: 29566181-29566181 on Build GRCh38
rs897868393 Chr.17: 29566234-29566234 on Build GRCh38
rs376765845 Chr.17: 29566220-29566220 on Build GRCh38
rs772892541 Chr.17: 29566386-29566386 on Build GRCh38
rs942606917 Chr.17: 29566417-29566417 on Build GRCh38
rs564476147 Chr.17: 29566044-29566044 on Build GRCh38
rs745944951 Chr.17: 29566440-29566440 on Build GRCh38
rs945711372 Chr.17: 29566063-29566063 on Build GRCh38
rs757344588 Chr.17: 29566433-29566433 on Build GRCh38
rs770956455 Chr.17: 29566334-29566334 on Build GRCh38
rs892946174 Chr.17: 29566208-29566208 on Build GRCh38
rs774006255 Chr.17: 29566284-29566284 on Build GRCh38
rs758342598 Chr.17: 29566376-29566376 on Build GRCh38
rs141623302 Chr.17: 29566299-29566299 on Build GRCh38
rs749657873 Chr.17: 29566442-29566442 on Build GRCh38
rs200675764 Chr.17: 29566282-29566282 on Build GRCh38
rs773230637 Chr.17: 29566256-29566256 on Build GRCh38
rs751758531 Chr.17: 29566423-29566423 on Build GRCh38
rs781222893 Chr.17: 29566066-29566066 on Build GRCh38
rs747097434 Chr.17: 29566261-29566261 on Build GRCh38
rs918336064 Chr.17: 29566369-29566369 on Build GRCh38
rs372213562 Chr.17: 29566123-29566123 on Build GRCh38
rs781681840 Chr.17: 29566352-29566352 on Build GRCh38
rs779932999 Chr.17: 29566086-29566086 on Build GRCh38
rs759870826 Chr.17: 29566168-29566168 on Build GRCh38
rs147908255 Chr.17: 29566151-29566151 on Build GRCh38
rs879860619 Chr.17: 29566375-29566375 on Build GRCh38
rs758180259 Chr.17: 29566407-29566407 on Build GRCh38
rs371803665 Chr.17: 29566233-29566233 on Build GRCh38
rs749147531 Chr.17: 29566232-29566232 on Build GRCh38
rs763365790 Chr.17: 29566384-29566384 on Build GRCh38
rs755950188 Chr.17: 29566076-29566076 on Build GRCh38
rs771674609 Chr.17: 29566385-29566385 on Build GRCh38
rs752528705 Chr.17: 29566323-29566323 on Build GRCh38
rs765251819 Chr.17: 29566402-29566402 on Build GRCh38
rs778740691 Chr.17: 29566356-29566377 on Build GRCh38
rs778032525 Chr.17: 29566331-29566331 on Build GRCh38
rs200715985 Chr.17: 29566389-29566389 on Build GRCh38
rs746069586 Chr.17: 29566224-29566224 on Build GRCh38
rs902543102 Chr.17: 29566254-29566254 on Build GRCh38
rs748256915 Chr.17: 29566247-29566247 on Build GRCh38
rs760335631 Chr.17: 29566461-29566461 on Build GRCh38
rs754052040 Chr.17: 29566125-29566126 on Build GRCh38
rs779298051 Chr.17: 29566244-29566244 on Build GRCh38
rs974256801 Chr.17: 29566329-29566329 on Build GRCh38
rs374244109 Chr.17: 29566042-29566042 on Build GRCh38
rs775785487 Chr.17: 29566376-29566376 on Build GRCh38
rs752483462 Chr.17: 29566406-29566406 on Build GRCh38
rs766462600 Chr.17: 29566298-29566298 on Build GRCh38
rs143659545 Chr.17: 29566453-29566453 on Build GRCh38
rs912927481 Chr.17: 29566049-29566049 on Build GRCh38
rs780130012 Chr.17: 29566230-29566230 on Build GRCh38
rs781182639 Chr.17: 29566438-29566438 on Build GRCh38
rs746318522 Chr.17: 29566354-29566354 on Build GRCh38
rs749094319 Chr.17: 29566096-29566096 on Build GRCh38
rs745745137 Chr.17: 29566070-29566070 on Build GRCh38
rs771993394 Chr.17: 29566130-29566130 on Build GRCh38
rs750217093 Chr.17: 29566374-29566374 on Build GRCh38
rs771475575 Chr.17: 29566270-29566270 on Build GRCh38
rs759926283 Chr.17: 29566278-29566278 on Build GRCh38
rs769060665 Chr.17: 29566106-29566106 on Build GRCh38
rs758113373 Chr.17: 29566036-29566036 on Build GRCh38
rs916713762 Chr.17: 29566029-29566029 on Build GRCh38
rs763774809 Chr.17: 29566169-29566169 on Build GRCh38
rs778780742 Chr.17: 29566314-29566314 on Build GRCh38
rs756815944 Chr.17: 29566194-29566194 on Build GRCh38
rs770698577 Chr.17: 29566473-29566473 on Build GRCh38
rs761304957 Chr.17: 29566285-29566285 on Build GRCh38
rs369157808 Chr.17: 29566387-29566387 on Build GRCh38
rs754831242 Chr.17: 29566305-29566305 on Build GRCh38

View Full Product Details