Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 17260026-17260525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS3
Entrez Gene ID
8533
Gene Name
COP9 signalosome subunit 3
Gene Aliases
CSN3, SGN3
UniGene
Hs.6076
Gene Chromosome Location
Chr.17: 17246624-17281303 on Build GRCh38
Transcripts
NM_001199125.1 NM_001316354.1 NM_001316355.1 NM_001316356.1 NM_001316357.1 NM_001316358.1 NM_003653.3 XM_005256837.4 XM_005256840.3 XM_005256842.4 XM_017025246.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
436130 c.797T>C p.V266A Chr.17: 17260440-17260440 on Build GRCh38
4674189 c.783C>T p.H261H Chr.17: 17260454-17260454 on Build GRCh38
2740092 c.863G>A p.R288H Chr.17: 17260374-17260374 on Build GRCh38
436129 c.842A>G p.K281R Chr.17: 17260395-17260395 on Build GRCh38
4674188 c.915_917delGAA p.K305delK Chr.17: 17260320-17260322 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368452318 Chr.17: 17260454-17260454 on Build GRCh38
rs777413669 Chr.17: 17260291-17260291 on Build GRCh38
rs752254806 Chr.17: 17260493-17260494 on Build GRCh38
rs754877501 Chr.17: 17260250-17260250 on Build GRCh38
rs140397176 Chr.17: 17260372-17260372 on Build GRCh38
rs575149632 Chr.17: 17260157-17260157 on Build GRCh38
rs766627921 Chr.17: 17260386-17260386 on Build GRCh38
rs760854832 Chr.17: 17260072-17260072 on Build GRCh38
rs770154004 Chr.17: 17260485-17260485 on Build GRCh38
rs938149571 Chr.17: 17260165-17260165 on Build GRCh38
rs921360002 Chr.17: 17260054-17260054 on Build GRCh38
rs552239580 Chr.17: 17260445-17260445 on Build GRCh38
rs767434942 Chr.17: 17260410-17260410 on Build GRCh38
rs1006145068 Chr.17: 17260274-17260274 on Build GRCh38
rs370425975 Chr.17: 17260373-17260373 on Build GRCh38
rs777454485 Chr.17: 17260423-17260423 on Build GRCh38
rs748909016 Chr.17: 17260298-17260298 on Build GRCh38
rs1046391496 Chr.17: 17260089-17260089 on Build GRCh38
rs752579367 Chr.17: 17260411-17260411 on Build GRCh38
rs375625767 Chr.17: 17260346-17260346 on Build GRCh38
rs912730832 Chr.17: 17260179-17260179 on Build GRCh38
rs888649184 Chr.17: 17260242-17260242 on Build GRCh38
rs748622935 Chr.17: 17260482-17260482 on Build GRCh38
rs745358654 Chr.17: 17260461-17260461 on Build GRCh38
rs368053525 Chr.17: 17260286-17260286 on Build GRCh38
rs561587944 Chr.17: 17260120-17260120 on Build GRCh38
rs945258387 Chr.17: 17260071-17260071 on Build GRCh38
rs954308105 Chr.17: 17260494-17260494 on Build GRCh38
rs778601165 Chr.17: 17260330-17260330 on Build GRCh38
rs199633470 Chr.17: 17260415-17260415 on Build GRCh38
rs541391295 Chr.17: 17260119-17260119 on Build GRCh38
rs781153390 Chr.17: 17260269-17260269 on Build GRCh38
rs998402570 Chr.17: 17260289-17260289 on Build GRCh38
rs77161056 Chr.17: 17260057-17260057 on Build GRCh38
rs759666547 Chr.17: 17260406-17260406 on Build GRCh38
rs1009920789 Chr.17: 17260424-17260424 on Build GRCh38
rs151026691 Chr.17: 17260316-17260316 on Build GRCh38
rs367575161 Chr.17: 17260400-17260400 on Build GRCh38
rs755614179 Chr.17: 17260237-17260237 on Build GRCh38
rs560095139 Chr.17: 17260050-17260050 on Build GRCh38
rs773417129 Chr.17: 17260489-17260489 on Build GRCh38
rs531825543 Chr.17: 17260427-17260427 on Build GRCh38
rs780842313 Chr.17: 17260115-17260115 on Build GRCh38
rs772040887 Chr.17: 17260195-17260195 on Build GRCh38
rs927961661 Chr.17: 17260101-17260101 on Build GRCh38
rs1037274553 Chr.17: 17260221-17260221 on Build GRCh38
rs756868131 Chr.17: 17260055-17260055 on Build GRCh38
rs745415469 Chr.17: 17260362-17260362 on Build GRCh38
rs771124317 Chr.17: 17260493-17260493 on Build GRCh38
rs543449184 Chr.17: 17260213-17260213 on Build GRCh38
rs770059818 Chr.17: 17260374-17260374 on Build GRCh38
rs374083241 Chr.17: 17260279-17260279 on Build GRCh38
rs989246951 Chr.17: 17260053-17260053 on Build GRCh38
rs563225822 Chr.17: 17260363-17260363 on Build GRCh38
rs745396331 Chr.17: 17260173-17260173 on Build GRCh38
rs768703155 Chr.17: 17260469-17260469 on Build GRCh38
rs532153875 Chr.17: 17260428-17260428 on Build GRCh38
rs758050667 Chr.17: 17260464-17260464 on Build GRCh38
rs749701721 Chr.17: 17260491-17260491 on Build GRCh38
rs1007331463 Chr.17: 17260334-17260334 on Build GRCh38
rs373742862 Chr.17: 17260375-17260375 on Build GRCh38
rs763114048 Chr.17: 17260376-17260376 on Build GRCh38
rs1048731204 Chr.17: 17260287-17260287 on Build GRCh38
rs767503113 Chr.17: 17260259-17260259 on Build GRCh38

View Full Product Details