Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18744154-18744658 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW10
Entrez Gene ID
10517
Gene Name
F-box and WD repeat domain containing 10
Gene Aliases
Fbw10, HREP, SM25H2, SM2SH2
UniGene
Hs.592128
Gene Chromosome Location
Chr.17: 18744013-18779349 on Build GRCh38
Transcripts
NM_001267585.1 NM_001267586.1 NR_051988.1 XM_011523625.1 XM_011523626.2 XM_017024020.1 XM_017024021.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4064547 c.17C>T p.S6L Chr.17: 18744261-18744261 on Build GRCh38
1252286 c.214T>A p.Y72N Chr.17: 18744458-18744458 on Build GRCh38
4684055 c.90G>T p.E30D Chr.17: 18744334-18744334 on Build GRCh38
4579501 c.408T>C p.Y136Y Chr.17: 18744652-18744652 on Build GRCh38
5454518 c.58A>G p.T20A Chr.17: 18744302-18744302 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768357844 Chr.17: 18744590-18744590 on Build GRCh38
rs186119518 Chr.17: 18744210-18744210 on Build GRCh38
rs148027955 Chr.17: 18744512-18744512 on Build GRCh38
rs1055730216 Chr.17: 18744385-18744385 on Build GRCh38
rs751820849 Chr.17: 18744313-18744313 on Build GRCh38
rs767063943 Chr.17: 18744269-18744269 on Build GRCh38
rs776223797 Chr.17: 18744461-18744461 on Build GRCh38
rs759800252 Chr.17: 18744513-18744513 on Build GRCh38
rs200553727 Chr.17: 18744324-18744324 on Build GRCh38
rs143573197 Chr.17: 18744476-18744476 on Build GRCh38
rs778797983 Chr.17: 18744558-18744558 on Build GRCh38
rs377476079 Chr.17: 18744548-18744548 on Build GRCh38
rs1013644169 Chr.17: 18744188-18744188 on Build GRCh38
rs557257349 Chr.17: 18744212-18744212 on Build GRCh38
rs764249402 Chr.17: 18744626-18744626 on Build GRCh38
rs765612082 Chr.17: 18744297-18744297 on Build GRCh38
rs756298001 Chr.17: 18744530-18744530 on Build GRCh38
rs769617817 Chr.17: 18744200-18744200 on Build GRCh38
rs764573852 Chr.17: 18744211-18744211 on Build GRCh38
rs775852531 Chr.17: 18744622-18744622 on Build GRCh38
rs757644169 Chr.17: 18744216-18744216 on Build GRCh38
rs528488880 Chr.17: 18744410-18744410 on Build GRCh38
rs769553087 Chr.17: 18744250-18744250 on Build GRCh38
rs369742505 Chr.17: 18744195-18744195 on Build GRCh38
rs780225956 Chr.17: 18744403-18744405 on Build GRCh38
rs778707969 Chr.17: 18744446-18744446 on Build GRCh38
rs748130978 Chr.17: 18744249-18744249 on Build GRCh38
rs750777371 Chr.17: 18744218-18744218 on Build GRCh38
rs568555990 Chr.17: 18744457-18744457 on Build GRCh38
rs771803969 Chr.17: 18744455-18744455 on Build GRCh38
rs752311721 Chr.17: 18744190-18744190 on Build GRCh38
rs1025099572 Chr.17: 18744194-18744199 on Build GRCh38
rs780882213 Chr.17: 18744595-18744595 on Build GRCh38
rs149323817 Chr.17: 18744453-18744453 on Build GRCh38
rs753966727 Chr.17: 18744628-18744628 on Build GRCh38
rs545784792 Chr.17: 18744399-18744399 on Build GRCh38
rs531699302 Chr.17: 18744613-18744613 on Build GRCh38
rs755097685 Chr.17: 18744323-18744323 on Build GRCh38
rs764159611 Chr.17: 18744547-18744547 on Build GRCh38
rs530588304 Chr.17: 18744464-18744464 on Build GRCh38
rs773849873 Chr.17: 18744363-18744363 on Build GRCh38
rs747124829 Chr.17: 18744237-18744237 on Build GRCh38
rs772332798 Chr.17: 18744621-18744621 on Build GRCh38
rs1032488908 Chr.17: 18744473-18744473 on Build GRCh38
rs752076802 Chr.17: 18744279-18744279 on Build GRCh38
rs201935353 Chr.17: 18744489-18744489 on Build GRCh38
rs777742132 Chr.17: 18744424-18744424 on Build GRCh38
rs11544711 Chr.17: 18744312-18744312 on Build GRCh38
rs762413363 Chr.17: 18744289-18744289 on Build GRCh38
rs757117927 Chr.17: 18744445-18744445 on Build GRCh38
rs959365809 Chr.17: 18744187-18744187 on Build GRCh38
rs760016525 Chr.17: 18744620-18744620 on Build GRCh38
rs772319592 Chr.17: 18744281-18744281 on Build GRCh38
rs745535174 Chr.17: 18744341-18744341 on Build GRCh38
rs751095242 Chr.17: 18744208-18744208 on Build GRCh38
rs751908282 Chr.17: 18744398-18744398 on Build GRCh38
rs572883925 Chr.17: 18744304-18744304 on Build GRCh38
rs775087357 Chr.17: 18744355-18744355 on Build GRCh38
rs553036479 Chr.17: 18744252-18744252 on Build GRCh38
rs1033648983 Chr.17: 18744186-18744186 on Build GRCh38
rs758122859 Chr.17: 18744561-18744561 on Build GRCh38
rs772826093 Chr.17: 18744201-18744201 on Build GRCh38
rs765980822 Chr.17: 18744204-18744204 on Build GRCh38
rs757205842 Chr.17: 18744556-18744556 on Build GRCh38
rs765811738 Chr.17: 18744360-18744360 on Build GRCh38
rs573539157 Chr.17: 18744535-18744536 on Build GRCh38
rs758716432 Chr.17: 18744220-18744220 on Build GRCh38
rs748140429 Chr.17: 18744330-18744330 on Build GRCh38
rs752873990 Chr.17: 18744417-18744417 on Build GRCh38
rs779337212 Chr.17: 18744217-18744217 on Build GRCh38
rs759102264 Chr.17: 18744262-18744262 on Build GRCh38
rs779813238 Chr.17: 18744566-18744566 on Build GRCh38
rs774975314 Chr.17: 18744456-18744456 on Build GRCh38
rs761849181 Chr.17: 18744630-18744630 on Build GRCh38
rs62073745 Chr.17: 18744226-18744226 on Build GRCh38
rs143662391 Chr.17: 18744338-18744338 on Build GRCh38
rs781052077 Chr.17: 18744246-18744246 on Build GRCh38
rs770621487 Chr.17: 18744261-18744261 on Build GRCh38
rs762601352 Chr.17: 18744202-18744202 on Build GRCh38
rs200806553 Chr.17: 18744409-18744409 on Build GRCh38
rs762483019 Chr.17: 18744192-18744192 on Build GRCh38
rs774691557 Chr.17: 18744614-18744614 on Build GRCh38
rs747613611 Chr.17: 18744609-18744609 on Build GRCh38
rs550819237 Chr.17: 18744467-18744467 on Build GRCh38
rs138647311 Chr.17: 18744350-18744350 on Build GRCh38
rs762689214 Chr.17: 18744260-18744260 on Build GRCh38
rs760915440 Chr.17: 18744625-18744625 on Build GRCh38
rs142528410 Chr.17: 18744587-18744587 on Build GRCh38
rs539416490 Chr.17: 18744245-18744245 on Build GRCh38
rs559368390 Chr.17: 18744406-18744406 on Build GRCh38
rs532702039 Chr.17: 18744396-18744396 on Build GRCh38
rs1013632642 Chr.17: 18744408-18744408 on Build GRCh38
rs574904473 Chr.17: 18744337-18744337 on Build GRCh38
rs965011265 Chr.17: 18744578-18744578 on Build GRCh38
rs568114049 Chr.17: 18744194-18744194 on Build GRCh38
rs758802594 Chr.17: 18744305-18744305 on Build GRCh38
rs749232563 Chr.17: 18744427-18744427 on Build GRCh38
rs760081035 Chr.17: 18744359-18744359 on Build GRCh38
rs763469446 Chr.17: 18744364-18744364 on Build GRCh38
rs745713216 Chr.17: 18744560-18744560 on Build GRCh38
rs769796646 Chr.17: 18744336-18744336 on Build GRCh38
rs752861486 Chr.17: 18744520-18744520 on Build GRCh38
rs767824789 Chr.17: 18744515-18744515 on Build GRCh38

View Full Product Details