Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18749815-18750359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW10
Entrez Gene ID
10517
Gene Name
F-box and WD repeat domain containing 10
Gene Aliases
Fbw10, HREP, SM25H2, SM2SH2
UniGene
Hs.592128
Gene Chromosome Location
Chr.17: 18744013-18779349 on Build GRCh38
Transcripts
NM_001267585.1 NM_001267586.1 NR_051988.1 XM_011523625.1 XM_011523626.2 XM_017024020.1 XM_017024021.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3988832 c.773T>A p.L258* Chr.17: 18749824-18749824 on Build GRCh38
386784 c.835C>A p.R279S Chr.17: 18749886-18749886 on Build GRCh38
5779976 c.958T>A p.S320T Chr.17: 18750096-18750096 on Build GRCh38
6458294 c.996_997delTG p.V333fs*37 Chr.17: 18750047-18750048 on Build GRCh38
6662424 c.883A>G p.R295G Chr.17: 18750021-18750021 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774102601 Chr.17: 18749997-18749997 on Build GRCh38
rs755009683 Chr.17: 18749887-18749887 on Build GRCh38
rs771417449 Chr.17: 18750111-18750111 on Build GRCh38
rs777652149 Chr.17: 18750007-18750007 on Build GRCh38
rs545382552 Chr.17: 18749861-18749862 on Build GRCh38
rs779830992 Chr.17: 18750100-18750100 on Build GRCh38
rs563388788 Chr.17: 18749912-18749912 on Build GRCh38
rs1028236667 Chr.17: 18750207-18750207 on Build GRCh38
rs772936873 Chr.17: 18749840-18749840 on Build GRCh38
rs770535291 Chr.17: 18749850-18749850 on Build GRCh38
rs779743177 Chr.17: 18750042-18750042 on Build GRCh38
rs574377467 Chr.17: 18749858-18749858 on Build GRCh38
rs879417070 Chr.17: 18750188-18750188 on Build GRCh38
rs773841637 Chr.17: 18750060-18750060 on Build GRCh38
rs762523622 Chr.17: 18749843-18749844 on Build GRCh38
rs1007587839 Chr.17: 18750269-18750269 on Build GRCh38
rs368213609 Chr.17: 18749886-18749886 on Build GRCh38
rs773938679 Chr.17: 18749854-18749854 on Build GRCh38
rs763387114 Chr.17: 18749975-18749975 on Build GRCh38
rs748093008 Chr.17: 18749900-18749900 on Build GRCh38
rs145368701 Chr.17: 18750105-18750105 on Build GRCh38
rs746575161 Chr.17: 18750104-18750104 on Build GRCh38
rs903697760 Chr.17: 18750161-18750161 on Build GRCh38
rs773866777 Chr.17: 18749924-18749924 on Build GRCh38
rs780780178 Chr.17: 18750106-18750106 on Build GRCh38
rs756073865 Chr.17: 18750005-18750005 on Build GRCh38
rs767736420 Chr.17: 18750075-18750075 on Build GRCh38
rs758028438 Chr.17: 18750097-18750097 on Build GRCh38
rs531915116 Chr.17: 18749931-18749931 on Build GRCh38
rs878858216 Chr.17: 18750176-18750176 on Build GRCh38
rs755982110 Chr.17: 18749910-18749910 on Build GRCh38
rs745522658 Chr.17: 18750038-18750038 on Build GRCh38
rs889097720 Chr.17: 18750247-18750247 on Build GRCh38
rs754384552 Chr.17: 18750047-18750048 on Build GRCh38
rs752721490 Chr.17: 18749990-18749990 on Build GRCh38
rs560771896 Chr.17: 18749839-18749839 on Build GRCh38
rs772325828 Chr.17: 18750149-18750149 on Build GRCh38
rs1042179363 Chr.17: 18750115-18750115 on Build GRCh38
rs752811151 Chr.17: 18750077-18750077 on Build GRCh38
rs150057313 Chr.17: 18750090-18750090 on Build GRCh38
rs770652948 Chr.17: 18749918-18749918 on Build GRCh38
rs547763481 Chr.17: 18750065-18750065 on Build GRCh38
rs1000731650 Chr.17: 18750197-18750197 on Build GRCh38
rs565557139 Chr.17: 18750009-18750009 on Build GRCh38
rs930844094 Chr.17: 18749996-18749996 on Build GRCh38
rs753781961 Chr.17: 18750087-18750087 on Build GRCh38
rs17852531 Chr.17: 18749875-18749875 on Build GRCh38
rs763441089 Chr.17: 18750062-18750062 on Build GRCh38
rs760735331 Chr.17: 18750080-18750080 on Build GRCh38
rs758716390 Chr.17: 18749882-18749882 on Build GRCh38
rs776725881 Chr.17: 18750172-18750172 on Build GRCh38
rs757764850 Chr.17: 18750147-18750148 on Build GRCh38
rs771707792 Chr.17: 18750041-18750041 on Build GRCh38
rs200711350 Chr.17: 18749963-18749963 on Build GRCh38
rs773751826 Chr.17: 18749971-18749971 on Build GRCh38
rs538432795 Chr.17: 18750198-18750198 on Build GRCh38
rs759696705 Chr.17: 18750066-18750066 on Build GRCh38
rs762433594 Chr.17: 18749968-18749968 on Build GRCh38
rs534570378 Chr.17: 18750335-18750335 on Build GRCh38
rs751724596 Chr.17: 18749981-18749981 on Build GRCh38
rs757001235 Chr.17: 18750026-18750026 on Build GRCh38
rs770473344 Chr.17: 18750058-18750058 on Build GRCh38
rs569844204 Chr.17: 18750187-18750187 on Build GRCh38
rs746209573 Chr.17: 18750136-18750136 on Build GRCh38
rs776987029 Chr.17: 18749874-18749874 on Build GRCh38
rs749037300 Chr.17: 18749914-18749914 on Build GRCh38
rs764300127 Chr.17: 18750023-18750026 on Build GRCh38
rs926267527 Chr.17: 18750052-18750052 on Build GRCh38
rs764725786 Chr.17: 18749865-18749865 on Build GRCh38
rs750193700 Chr.17: 18750095-18750095 on Build GRCh38
rs766680128 Chr.17: 18749980-18749980 on Build GRCh38
rs748947198 Chr.17: 18749844-18749844 on Build GRCh38
rs541014989 Chr.17: 18749838-18749838 on Build GRCh38
rs559008663 Chr.17: 18749996-18749997 on Build GRCh38
rs528013240 Chr.17: 18750028-18750028 on Build GRCh38
rs766487517 Chr.17: 18749883-18749883 on Build GRCh38
rs558512330 Chr.17: 18750202-18750202 on Build GRCh38
rs746487298 Chr.17: 18750044-18750044 on Build GRCh38
rs144428015 Chr.17: 18750160-18750160 on Build GRCh38
rs775688571 Chr.17: 18750150-18750150 on Build GRCh38
rs771494320 Chr.17: 18749933-18749933 on Build GRCh38
rs372514251 Chr.17: 18750170-18750170 on Build GRCh38
rs767505981 Chr.17: 18749989-18749989 on Build GRCh38
rs937728779 Chr.17: 18750079-18750079 on Build GRCh38
rs753694558 Chr.17: 18750008-18750008 on Build GRCh38
rs200034399 Chr.17: 18750113-18750113 on Build GRCh38
rs372345142 Chr.17: 18750081-18750081 on Build GRCh38
rs759734557 Chr.17: 18749986-18749986 on Build GRCh38
rs781249754 Chr.17: 18749896-18749896 on Build GRCh38
rs778597832 Chr.17: 18750093-18750093 on Build GRCh38
rs749770488 Chr.17: 18750107-18750107 on Build GRCh38
rs572302093 Chr.17: 18750203-18750203 on Build GRCh38
rs761689141 Chr.17: 18750178-18750178 on Build GRCh38

View Full Product Details