Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 21416743-21417249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNJ12
Entrez Gene ID
3768
Gene Name
potassium voltage-gated channel subfamily J member 12
Gene Aliases
IRK-2, IRK2, KCNJN1, Kir2.2, Kir2.2v, hIRK, hIRK1, hkir2.2x, kcnj12x
UniGene
Hs.200629
Gene Chromosome Location
Chr.17: 21376387-21419870 on Build GRCh38
Transcripts
NM_021012.4 XM_005256625.4 XM_011523831.2

SNP Detail

SNP ID Chromosome Location
rs542033485 Chr.17: 21416770-21416770 on Build GRCh38
rs551866283 Chr.17: 21417180-21417180 on Build GRCh38
rs73313941 Chr.17: 21417187-21417187 on Build GRCh38
rs375616058 Chr.17: 21417106-21417107 on Build GRCh38
rs572467810 Chr.17: 21416829-21416829 on Build GRCh38
rs67199304 Chr.17: 21417128-21417128 on Build GRCh38
rs34010550 Chr.17: 21417081-21417082 on Build GRCh38
rs77646945 Chr.17: 21417081-21417081 on Build GRCh38
rs62049520 Chr.17: 21416898-21416898 on Build GRCh38
rs5021705 Chr.17: 21416900-21416900 on Build GRCh38
rs570101403 Chr.17: 21417194-21417194 on Build GRCh38
rs561725120 Chr.17: 21417157-21417157 on Build GRCh38
rs5028197 Chr.17: 21417011-21417011 on Build GRCh38
rs73981226 Chr.17: 21417038-21417038 on Build GRCh38
rs67680166 Chr.17: 21417217-21417217 on Build GRCh38
rs562434065 Chr.17: 21416973-21416973 on Build GRCh38
rs5021704 Chr.17: 21416802-21416802 on Build GRCh38
rs75437577 Chr.17: 21417114-21417114 on Build GRCh38
rs113939610 Chr.17: 21417117-21417117 on Build GRCh38
rs75497256 Chr.17: 21417107-21417107 on Build GRCh38
rs556160072 Chr.17: 21417207-21417207 on Build GRCh38
rs73313935 Chr.17: 21417113-21417113 on Build GRCh38
rs62049521 Chr.17: 21416940-21416940 on Build GRCh38
rs537492715 Chr.17: 21417202-21417202 on Build GRCh38
rs62049522 Chr.17: 21417053-21417053 on Build GRCh38
rs369195245 Chr.17: 21416991-21416991 on Build GRCh38
rs73313932 Chr.17: 21416967-21416967 on Build GRCh38
rs550492787 Chr.17: 21416957-21416957 on Build GRCh38
rs546339922 Chr.17: 21416839-21416839 on Build GRCh38
rs368249953 Chr.17: 21417113-21417113 on Build GRCh38
rs5021703 Chr.17: 21416790-21416790 on Build GRCh38
rs199796804 Chr.17: 21417094-21417094 on Build GRCh38
rs560368469 Chr.17: 21416771-21416771 on Build GRCh38
rs201692849 Chr.17: 21417093-21417093 on Build GRCh38
rs78347298 Chr.17: 21417082-21417082 on Build GRCh38
rs74366214 Chr.17: 21417080-21417080 on Build GRCh38
rs78953330 Chr.17: 21417115-21417115 on Build GRCh38
rs78164125 Chr.17: 21417109-21417109 on Build GRCh38
rs564424188 Chr.17: 21416897-21416897 on Build GRCh38
rs79945769 Chr.17: 21416885-21416885 on Build GRCh38
rs74591721 Chr.17: 21417139-21417139 on Build GRCh38
rs111237557 Chr.17: 21417124-21417124 on Build GRCh38
rs566166138 Chr.17: 21417112-21417112 on Build GRCh38
rs5021706 Chr.17: 21416951-21416951 on Build GRCh38
rs74336563 Chr.17: 21417168-21417168 on Build GRCh38
rs547939123 Chr.17: 21416994-21416994 on Build GRCh38

View Full Product Details