Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 27878540-27879031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LYRM9
Entrez Gene ID
201229
Gene Name
LYR motif containing 9
Gene Aliases
C17orf108, HSD24
UniGene
Hs.687951
Gene Chromosome Location
Chr.17: 27878314-27893383 on Build GRCh38
Transcripts
NM_001076680.1 XM_011524491.2 XM_017024325.1

SNP Detail

SNP ID Chromosome Location
rs15133 Chr.17: 27878616-27878616 on Build GRCh38
rs954496167 Chr.17: 27878671-27878671 on Build GRCh38
rs963517841 Chr.17: 27878919-27878919 on Build GRCh38
rs77880338 Chr.17: 27878947-27878947 on Build GRCh38
rs974338500 Chr.17: 27878960-27878960 on Build GRCh38
rs531425579 Chr.17: 27878863-27878863 on Build GRCh38
rs560458962 Chr.17: 27878680-27878680 on Build GRCh38
rs915383330 Chr.17: 27878743-27878743 on Build GRCh38
rs987105502 Chr.17: 27878673-27878673 on Build GRCh38
rs142337150 Chr.17: 27878905-27878905 on Build GRCh38
rs1043009055 Chr.17: 27878767-27878767 on Build GRCh38
rs945588556 Chr.17: 27878755-27878755 on Build GRCh38
rs1023981291 Chr.17: 27878956-27878956 on Build GRCh38
rs1023528684 Chr.17: 27878687-27878687 on Build GRCh38
rs73986075 Chr.17: 27878686-27878686 on Build GRCh38
rs929710467 Chr.17: 27878818-27878818 on Build GRCh38
rs549545339 Chr.17: 27878900-27878900 on Build GRCh38
rs74504364 Chr.17: 27878940-27878940 on Build GRCh38
rs1026763390 Chr.17: 27878962-27878962 on Build GRCh38
rs535051444 Chr.17: 27878933-27878933 on Build GRCh38
rs910211259 Chr.17: 27878971-27878971 on Build GRCh38
rs1053502322 Chr.17: 27878695-27878695 on Build GRCh38
rs542123963 Chr.17: 27878659-27878659 on Build GRCh38
rs903932315 Chr.17: 27878632-27878632 on Build GRCh38
rs900720751 Chr.17: 27878901-27878901 on Build GRCh38
rs554057289 Chr.17: 27878937-27878937 on Build GRCh38
rs773566657 Chr.17: 27878650-27878650 on Build GRCh38
rs903960170 Chr.17: 27878918-27878918 on Build GRCh38
rs906799743 Chr.17: 27878865-27878865 on Build GRCh38
rs1005625376 Chr.17: 27878906-27878906 on Build GRCh38
rs374021153 Chr.17: 27878858-27878858 on Build GRCh38
rs558329286 Chr.17: 27878978-27878978 on Build GRCh38
rs987387134 Chr.17: 27878959-27878959 on Build GRCh38
rs763593182 Chr.17: 27878979-27878979 on Build GRCh38
rs552301451 Chr.17: 27878688-27878688 on Build GRCh38
rs1003922783 Chr.17: 27878899-27878899 on Build GRCh38
rs866588409 Chr.17: 27878635-27878635 on Build GRCh38
rs369790842 Chr.17: 27878662-27878662 on Build GRCh38
rs760236868 Chr.17: 27878969-27878969 on Build GRCh38
rs1017051401 Chr.17: 27878915-27878915 on Build GRCh38
rs1030988125 Chr.17: 27878661-27878661 on Build GRCh38
rs900688536 Chr.17: 27878574-27878574 on Build GRCh38
rs191021438 Chr.17: 27878597-27878597 on Build GRCh38
rs991283811 Chr.17: 27878596-27878596 on Build GRCh38
rs762292090 Chr.17: 27878885-27878886 on Build GRCh38
rs114073755 Chr.17: 27878610-27878610 on Build GRCh38
rs963269140 Chr.17: 27878609-27878609 on Build GRCh38
rs895189725 Chr.17: 27878903-27878903 on Build GRCh38
rs570500121 Chr.17: 27878720-27878720 on Build GRCh38

View Full Product Details