Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 43853550-43854044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD300LG
Entrez Gene ID
146894
Gene Name
CD300 molecule like family member g
Gene Aliases
CLM-9, CLM9, NEPMUCIN, TREM-4, TREM4
UniGene
Hs.147313
Gene Chromosome Location
Chr.17: 43846777-43863639 on Build GRCh38
Transcripts
NM_001168322.1 NM_001168323.1 NM_001168324.1 NM_145273.3 XM_005257075.2 XM_005257076.2 XM_006721721.2 XM_006721722.2 XM_011524384.2 XM_017024235.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
472865 c.552_553GC>TA p.L184>? Chr.17: 43853877-43853878 on Build GRCh38
5893913 c.557G>A p.G186E Chr.17: 43853882-43853882 on Build GRCh38
472867 c.647C>G p.P216R Chr.17: 43853972-43853972 on Build GRCh38
3691603 c.613C>A p.P205T Chr.17: 43853938-43853938 on Build GRCh38
979808 c.570C>T p.Y190Y Chr.17: 43853895-43853895 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760923345 Chr.17: 43853969-43853970 on Build GRCh38
rs956752295 Chr.17: 43854019-43854019 on Build GRCh38
rs777125264 Chr.17: 43853822-43853822 on Build GRCh38
rs201495676 Chr.17: 43853958-43853958 on Build GRCh38
rs149330626 Chr.17: 43853895-43853895 on Build GRCh38
rs540762451 Chr.17: 43853746-43853746 on Build GRCh38
rs748579528 Chr.17: 43853961-43853961 on Build GRCh38
rs201969253 Chr.17: 43853817-43853817 on Build GRCh38
rs552093596 Chr.17: 43853848-43853848 on Build GRCh38
rs760716131 Chr.17: 43853942-43853942 on Build GRCh38
rs114915030 Chr.17: 43853723-43853723 on Build GRCh38
rs375677756 Chr.17: 43853771-43853771 on Build GRCh38
rs775684712 Chr.17: 43853900-43853900 on Build GRCh38
rs9911625 Chr.17: 43853874-43853874 on Build GRCh38
rs778160205 Chr.17: 43853792-43853792 on Build GRCh38
rs764809817 Chr.17: 43853969-43853969 on Build GRCh38
rs565818799 Chr.17: 43853974-43853974 on Build GRCh38
rs752402778 Chr.17: 43853971-43853971 on Build GRCh38
rs12453522 Chr.17: 43854007-43854007 on Build GRCh38
rs761707909 Chr.17: 43853850-43853850 on Build GRCh38
rs757979028 Chr.17: 43854008-43854008 on Build GRCh38
rs968010374 Chr.17: 43853731-43853731 on Build GRCh38
rs772312024 Chr.17: 43853968-43853968 on Build GRCh38
rs150185239 Chr.17: 43853854-43853854 on Build GRCh38
rs776444960 Chr.17: 43854011-43854011 on Build GRCh38
rs1012670241 Chr.17: 43853703-43853703 on Build GRCh38
rs763272617 Chr.17: 43853836-43853836 on Build GRCh38
rs138728662 Chr.17: 43853869-43853869 on Build GRCh38
rs775570666 Chr.17: 43853832-43853832 on Build GRCh38
rs994308590 Chr.17: 43853673-43853673 on Build GRCh38
rs771389686 Chr.17: 43853821-43853821 on Build GRCh38
rs267604895 Chr.17: 43853902-43853902 on Build GRCh38
rs759083134 Chr.17: 43853979-43853979 on Build GRCh38
rs368997901 Chr.17: 43854001-43854001 on Build GRCh38
rs775688553 Chr.17: 43853926-43853943 on Build GRCh38
rs764133105 Chr.17: 43853970-43853970 on Build GRCh38
rs765955166 Chr.17: 43853861-43853861 on Build GRCh38
rs17852267 Chr.17: 43853986-43853986 on Build GRCh38
rs1050576909 Chr.17: 43853892-43853892 on Build GRCh38
rs748424993 Chr.17: 43853996-43853996 on Build GRCh38
rs758660052 Chr.17: 43853786-43853786 on Build GRCh38
rs201288918 Chr.17: 43853928-43853928 on Build GRCh38
rs781576192 Chr.17: 43853975-43853975 on Build GRCh38
rs778570647 Chr.17: 43853872-43853872 on Build GRCh38
rs1056751011 Chr.17: 43853683-43853683 on Build GRCh38
rs1023672635 Chr.17: 43853704-43853704 on Build GRCh38
rs866367835 Chr.17: 43853985-43853985 on Build GRCh38
rs73985460 Chr.17: 43853626-43853626 on Build GRCh38
rs749888524 Chr.17: 43853760-43853760 on Build GRCh38
rs935564914 Chr.17: 43853790-43853790 on Build GRCh38
rs769171631 Chr.17: 43853859-43853859 on Build GRCh38
rs769727412 Chr.17: 43853899-43853899 on Build GRCh38
rs908861648 Chr.17: 43853624-43853624 on Build GRCh38
rs756592250 Chr.17: 43853976-43853976 on Build GRCh38
rs953353692 Chr.17: 43853776-43853776 on Build GRCh38
rs991279272 Chr.17: 43853696-43853696 on Build GRCh38
rs968451673 Chr.17: 43853710-43853710 on Build GRCh38
rs774236298 Chr.17: 43853847-43853847 on Build GRCh38
rs146326351 Chr.17: 43853937-43853937 on Build GRCh38
rs781654438 Chr.17: 43853891-43853891 on Build GRCh38
rs941815064 Chr.17: 43853901-43853901 on Build GRCh38
rs763771839 Chr.17: 43853841-43853841 on Build GRCh38
rs373769899 Chr.17: 43853759-43853759 on Build GRCh38
rs764772064 Chr.17: 43853737-43853737 on Build GRCh38
rs779966692 Chr.17: 43853687-43853687 on Build GRCh38
rs374685266 Chr.17: 43853915-43853915 on Build GRCh38
rs761689655 Chr.17: 43853932-43853932 on Build GRCh38
rs779563629 Chr.17: 43853767-43853767 on Build GRCh38
rs746235278 Chr.17: 43853781-43853782 on Build GRCh38
rs530715075 Chr.17: 43853896-43853896 on Build GRCh38
rs558909154 Chr.17: 43853667-43853667 on Build GRCh38
rs769974140 Chr.17: 43853831-43853831 on Build GRCh38
rs563058490 Chr.17: 43853766-43853766 on Build GRCh38
rs372449182 Chr.17: 43853761-43853761 on Build GRCh38
rs773012123 Chr.17: 43854003-43854003 on Build GRCh38
rs138369136 Chr.17: 43853982-43853982 on Build GRCh38
rs140396225 Chr.17: 43853826-43853826 on Build GRCh38
rs552417566 Chr.17: 43853956-43853956 on Build GRCh38
rs772486596 Chr.17: 43853829-43853829 on Build GRCh38
rs749040721 Chr.17: 43853768-43853768 on Build GRCh38
rs760420111 Chr.17: 43854006-43854006 on Build GRCh38
rs757631036 Chr.17: 43853882-43853882 on Build GRCh38
rs1002769862 Chr.17: 43853657-43853657 on Build GRCh38
rs767507369 Chr.17: 43853852-43853852 on Build GRCh38
rs758974683 Chr.17: 43853959-43853959 on Build GRCh38
rs576653804 Chr.17: 43853625-43853625 on Build GRCh38
rs772995550 Chr.17: 43853939-43853939 on Build GRCh38

View Full Product Details