Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44075573-44076069 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G6PC3
Entrez Gene ID
92579
Gene Name
glucose-6-phosphatase catalytic subunit 3
Gene Aliases
SCN4, UGRP
UniGene
Hs.294005
Gene Chromosome Location
Chr.17: 44070700-44076344 on Build GRCh38
Transcripts
NM_001319945.1 NM_138387.3 NR_028581.1 NR_028582.1 XM_011525473.2 XM_011525474.2 XM_017025335.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7117538 c.811G>T p.A271S Chr.17: 44075813-44075813 on Build GRCh38
1383592 c.906delC p.P304fs*20 Chr.17: 44075908-44075908 on Build GRCh38
1252917 c.758G>A p.R253H Chr.17: 44075760-44075760 on Build GRCh38
5386450 c.927C>T p.F309F Chr.17: 44075929-44075929 on Build GRCh38
360827 c.846G>C p.Q282H Chr.17: 44075848-44075848 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751895520 Chr.17: 44075767-44075767 on Build GRCh38
rs755083701 Chr.17: 44075969-44075969 on Build GRCh38
rs978510802 Chr.17: 44075926-44075926 on Build GRCh38
rs771873137 Chr.17: 44075905-44075905 on Build GRCh38
rs142944376 Chr.17: 44075896-44075896 on Build GRCh38
rs113416399 Chr.17: 44076045-44076045 on Build GRCh38
rs889605875 Chr.17: 44076014-44076014 on Build GRCh38
rs765927570 Chr.17: 44075759-44075759 on Build GRCh38
rs879194442 Chr.17: 44076043-44076043 on Build GRCh38
rs775177185 Chr.17: 44075914-44075914 on Build GRCh38
rs1043122698 Chr.17: 44075651-44075651 on Build GRCh38
rs774344287 Chr.17: 44075717-44075717 on Build GRCh38
rs201268618 Chr.17: 44075869-44075869 on Build GRCh38
rs992652990 Chr.17: 44075647-44075647 on Build GRCh38
rs776556492 Chr.17: 44075910-44075910 on Build GRCh38
rs202238577 Chr.17: 44075885-44075885 on Build GRCh38
rs118203971 Chr.17: 44075786-44075786 on Build GRCh38
rs151105942 Chr.17: 44075999-44075999 on Build GRCh38
rs746410997 Chr.17: 44075791-44075791 on Build GRCh38
rs768125842 Chr.17: 44076025-44076025 on Build GRCh38
rs748931188 Chr.17: 44075767-44075768 on Build GRCh38
rs865977529 Chr.17: 44075770-44075770 on Build GRCh38
rs118203968 Chr.17: 44075760-44075760 on Build GRCh38
rs752981586 Chr.17: 44075686-44075686 on Build GRCh38
rs140785361 Chr.17: 44075729-44075729 on Build GRCh38
rs762579612 Chr.17: 44075922-44075922 on Build GRCh38
rs774716362 Chr.17: 44075819-44075819 on Build GRCh38
rs757593798 Chr.17: 44075848-44075848 on Build GRCh38
rs773752221 Chr.17: 44076000-44076001 on Build GRCh38
rs755580450 Chr.17: 44075645-44075645 on Build GRCh38
rs761219348 Chr.17: 44076044-44076046 on Build GRCh38
rs1025496655 Chr.17: 44076001-44076001 on Build GRCh38
rs141431274 Chr.17: 44076008-44076008 on Build GRCh38
rs762536153 Chr.17: 44076024-44076024 on Build GRCh38
rs201561483 Chr.17: 44075636-44075636 on Build GRCh38
rs1013640910 Chr.17: 44075997-44075997 on Build GRCh38
rs1049604897 Chr.17: 44076009-44076009 on Build GRCh38
rs780718603 Chr.17: 44075881-44075881 on Build GRCh38
rs200478425 Chr.17: 44075780-44075780 on Build GRCh38
rs1037358150 Chr.17: 44075859-44075859 on Build GRCh38
rs754361488 Chr.17: 44076046-44076046 on Build GRCh38
rs888765058 Chr.17: 44075824-44075824 on Build GRCh38
rs904160246 Chr.17: 44075670-44075670 on Build GRCh38
rs373207529 Chr.17: 44075838-44075838 on Build GRCh38
rs529935686 Chr.17: 44075701-44075701 on Build GRCh38
rs746741551 Chr.17: 44076003-44076003 on Build GRCh38
rs747931007 Chr.17: 44075821-44075821 on Build GRCh38
rs143791785 Chr.17: 44075700-44075700 on Build GRCh38
rs148559256 Chr.17: 44075831-44075831 on Build GRCh38
rs764750000 Chr.17: 44075913-44075913 on Build GRCh38
rs777475388 Chr.17: 44075757-44075780 on Build GRCh38
rs778098924 Chr.17: 44075766-44075766 on Build GRCh38
rs749505455 Chr.17: 44075811-44075811 on Build GRCh38
rs200971807 Chr.17: 44075762-44075762 on Build GRCh38
rs761874618 Chr.17: 44075633-44075633 on Build GRCh38
rs569262371 Chr.17: 44075755-44075755 on Build GRCh38
rs753515098 Chr.17: 44075672-44075672 on Build GRCh38
rs751548067 Chr.17: 44075972-44075972 on Build GRCh38
rs756740955 Chr.17: 44075873-44075873 on Build GRCh38
rs770799037 Chr.17: 44075908-44075908 on Build GRCh38
rs995618283 Chr.17: 44075601-44075601 on Build GRCh38
rs202163545 Chr.17: 44075940-44075940 on Build GRCh38
rs34491309 Chr.17: 44075817-44075817 on Build GRCh38
rs773183765 Chr.17: 44075823-44075823 on Build GRCh38
rs752964083 Chr.17: 44075983-44075983 on Build GRCh38
rs760179845 Chr.17: 44075740-44075740 on Build GRCh38
rs34019455 Chr.17: 44075913-44075914 on Build GRCh38
rs765872526 Chr.17: 44075826-44075826 on Build GRCh38
rs780218874 Chr.17: 44075643-44075643 on Build GRCh38
rs951492056 Chr.17: 44075624-44075624 on Build GRCh38
rs769341704 Chr.17: 44076016-44076016 on Build GRCh38
rs750695182 Chr.17: 44075928-44075928 on Build GRCh38
rs144587192 Chr.17: 44076019-44076019 on Build GRCh38
rs767908887 Chr.17: 44075923-44075923 on Build GRCh38
rs756548836 Chr.17: 44075929-44075929 on Build GRCh38
rs201244794 Chr.17: 44076037-44076037 on Build GRCh38
rs1025801526 Chr.17: 44075606-44075606 on Build GRCh38
rs1025726441 Chr.17: 44075953-44075953 on Build GRCh38
rs887279817 Chr.17: 44075966-44075966 on Build GRCh38
rs781564534 Chr.17: 44075698-44075698 on Build GRCh38
rs749887112 Chr.17: 44075640-44075640 on Build GRCh38
rs377343204 Chr.17: 44075689-44075689 on Build GRCh38
rs779143230 Chr.17: 44075889-44075889 on Build GRCh38
rs747469767 Chr.17: 44075687-44075687 on Build GRCh38
rs1052762189 Chr.17: 44075984-44075984 on Build GRCh38
rs1016698344 Chr.17: 44075775-44075775 on Build GRCh38
rs1028155953 Chr.17: 44075709-44075709 on Build GRCh38
rs768565958 Chr.17: 44075712-44075712 on Build GRCh38
rs200982052 Chr.17: 44075674-44075674 on Build GRCh38
rs746938030 Chr.17: 44075907-44075907 on Build GRCh38
rs548087372 Chr.17: 44075702-44075702 on Build GRCh38
rs758609108 Chr.17: 44075994-44075994 on Build GRCh38
rs145779116 Chr.17: 44075822-44075822 on Build GRCh38
rs752323869 Chr.17: 44075846-44075846 on Build GRCh38
rs199992248 Chr.17: 44075863-44075863 on Build GRCh38
rs201700844 Chr.17: 44075802-44075802 on Build GRCh38
rs200393998 Chr.17: 44075825-44075825 on Build GRCh38
rs763307095 Chr.17: 44075711-44075711 on Build GRCh38
rs770448782 Chr.17: 44075901-44075902 on Build GRCh38
rs934342273 Chr.17: 44075974-44075974 on Build GRCh38
rs770745949 Chr.17: 44076004-44076004 on Build GRCh38
rs995753400 Chr.17: 44075883-44075883 on Build GRCh38
rs797044567 Chr.17: 44075937-44075938 on Build GRCh38
rs781059301 Chr.17: 44076007-44076007 on Build GRCh38
rs767810900 Chr.17: 44075851-44075851 on Build GRCh38
rs779637877 Chr.17: 44075649-44075649 on Build GRCh38
rs147246080 Chr.17: 44075728-44075728 on Build GRCh38
rs558164927 Chr.17: 44075800-44075800 on Build GRCh38
rs750355531 Chr.17: 44075739-44075739 on Build GRCh38
rs774174457 Chr.17: 44076034-44076034 on Build GRCh38
rs960279172 Chr.17: 44075818-44075818 on Build GRCh38
rs934590407 Chr.17: 44075671-44075671 on Build GRCh38
rs757568546 Chr.17: 44075774-44075774 on Build GRCh38
rs554064887 Chr.17: 44075828-44075828 on Build GRCh38

View Full Product Details