Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44350924-44351428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRN
Entrez Gene ID
2896
Gene Name
granulin
Gene Aliases
CLN11, GEP, GP88, PCDGF, PEPI, PGRN
UniGene
Hs.514220
Gene Chromosome Location
Chr.17: 44345086-44353106 on Build GRCh38
Transcripts
NM_002087.3 XM_005257253.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
979937 c.869G>T p.C290F Chr.17: 44351396-44351396 on Build GRCh38
4935904 c.786C>T p.S262S Chr.17: 44351114-44351114 on Build GRCh38
6676269 c.795C>T p.N265N Chr.17: 44351123-44351123 on Build GRCh38
4512211 c.895C>T p.L299L Chr.17: 44351422-44351422 on Build GRCh38
4689027 c.796G>A p.A266T Chr.17: 44351124-44351124 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778066666 Chr.17: 44350957-44350957 on Build GRCh38
rs755779122 Chr.17: 44350993-44350993 on Build GRCh38
rs779615581 Chr.17: 44350999-44350999 on Build GRCh38
rs950604255 Chr.17: 44351175-44351175 on Build GRCh38
rs63751035 Chr.17: 44351087-44351088 on Build GRCh38
rs63750344 Chr.17: 44351071-44351071 on Build GRCh38
rs35759647 Chr.17: 44351318-44351318 on Build GRCh38
rs1051797384 Chr.17: 44351349-44351349 on Build GRCh38
rs776977351 Chr.17: 44351176-44351178 on Build GRCh38
rs63751296 Chr.17: 44351362-44351362 on Build GRCh38
rs63750229 Chr.17: 44351132-44351132 on Build GRCh38
rs1045288242 Chr.17: 44351223-44351223 on Build GRCh38
rs530854692 Chr.17: 44351220-44351220 on Build GRCh38
rs771907059 Chr.17: 44351360-44351360 on Build GRCh38
rs151050549 Chr.17: 44351141-44351141 on Build GRCh38
rs773431127 Chr.17: 44351016-44351016 on Build GRCh38
rs764968816 Chr.17: 44351406-44351406 on Build GRCh38
rs747593674 Chr.17: 44351021-44351021 on Build GRCh38
rs776705304 Chr.17: 44350984-44350985 on Build GRCh38
rs1054073758 Chr.17: 44351023-44351024 on Build GRCh38
rs761770604 Chr.17: 44351193-44351194 on Build GRCh38
rs63750435 Chr.17: 44351109-44351109 on Build GRCh38
rs753580360 Chr.17: 44351105-44351105 on Build GRCh38
rs936915984 Chr.17: 44351264-44351264 on Build GRCh38
rs995227941 Chr.17: 44351162-44351162 on Build GRCh38
rs200408271 Chr.17: 44351051-44351051 on Build GRCh38
rs747538590 Chr.17: 44350958-44350958 on Build GRCh38
rs552184990 Chr.17: 44351247-44351247 on Build GRCh38
rs754558857 Chr.17: 44351118-44351118 on Build GRCh38
rs202178902 Chr.17: 44351155-44351155 on Build GRCh38
rs374280545 Chr.17: 44351184-44351184 on Build GRCh38
rs765556715 Chr.17: 44351041-44351041 on Build GRCh38
rs1053119478 Chr.17: 44351274-44351274 on Build GRCh38
rs570501654 Chr.17: 44351311-44351311 on Build GRCh38
rs745639522 Chr.17: 44351080-44351081 on Build GRCh38
rs775409489 Chr.17: 44351145-44351145 on Build GRCh38
rs63750565 Chr.17: 44351388-44351388 on Build GRCh38
rs992569096 Chr.17: 44351168-44351168 on Build GRCh38
rs63751000 Chr.17: 44351101-44351101 on Build GRCh38
rs1001261540 Chr.17: 44350948-44350948 on Build GRCh38
rs757652171 Chr.17: 44351207-44351207 on Build GRCh38
rs376153436 Chr.17: 44351080-44351080 on Build GRCh38
rs556825114 Chr.17: 44351209-44351209 on Build GRCh38
rs202006119 Chr.17: 44351131-44351131 on Build GRCh38
rs780245492 Chr.17: 44351325-44351325 on Build GRCh38
rs997508256 Chr.17: 44351315-44351315 on Build GRCh38
rs778271207 Chr.17: 44351123-44351123 on Build GRCh38
rs1045338022 Chr.17: 44350945-44350945 on Build GRCh38
rs746425997 Chr.17: 44351215-44351215 on Build GRCh38
rs935706835 Chr.17: 44351330-44351330 on Build GRCh38
rs772437404 Chr.17: 44351013-44351013 on Build GRCh38
rs979891508 Chr.17: 44351294-44351294 on Build GRCh38
rs750312897 Chr.17: 44351173-44351173 on Build GRCh38
rs1016120447 Chr.17: 44351100-44351100 on Build GRCh38
rs606231221 Chr.17: 44351164-44351164 on Build GRCh38
rs377020182 Chr.17: 44351391-44351391 on Build GRCh38
rs529849967 Chr.17: 44351152-44351152 on Build GRCh38
rs112294997 Chr.17: 44351139-44351139 on Build GRCh38
rs772521950 Chr.17: 44350997-44350997 on Build GRCh38
rs770167919 Chr.17: 44351318-44351318 on Build GRCh38
rs63749877 Chr.17: 44351141-44351144 on Build GRCh38
rs971616593 Chr.17: 44351258-44351258 on Build GRCh38
rs983331801 Chr.17: 44351203-44351203 on Build GRCh38
rs761620336 Chr.17: 44351358-44351358 on Build GRCh38
rs63750976 Chr.17: 44351163-44351164 on Build GRCh38
rs758900589 Chr.17: 44351397-44351397 on Build GRCh38
rs760675928 Chr.17: 44351378-44351378 on Build GRCh38
rs904129782 Chr.17: 44350947-44350947 on Build GRCh38
rs779598691 Chr.17: 44351091-44351091 on Build GRCh38
rs755582839 Chr.17: 44351088-44351088 on Build GRCh38
rs374661973 Chr.17: 44351366-44351366 on Build GRCh38
rs894043737 Chr.17: 44351044-44351044 on Build GRCh38
rs536035764 Chr.17: 44351309-44351309 on Build GRCh38
rs72824736 Chr.17: 44351170-44351170 on Build GRCh38
rs576913209 Chr.17: 44351000-44351000 on Build GRCh38
rs751278565 Chr.17: 44351056-44351056 on Build GRCh38
rs761530797 Chr.17: 44351156-44351156 on Build GRCh38
rs953208851 Chr.17: 44351078-44351078 on Build GRCh38
rs759627977 Chr.17: 44351040-44351040 on Build GRCh38
rs376226985 Chr.17: 44351355-44351355 on Build GRCh38
rs201528305 Chr.17: 44351190-44351190 on Build GRCh38
rs904033884 Chr.17: 44351252-44351252 on Build GRCh38
rs768929056 Chr.17: 44351328-44351328 on Build GRCh38
rs142805340 Chr.17: 44351081-44351081 on Build GRCh38
rs775740192 Chr.17: 44351047-44351047 on Build GRCh38
rs776002425 Chr.17: 44351024-44351024 on Build GRCh38
rs772963921 Chr.17: 44351368-44351368 on Build GRCh38
rs63750548 Chr.17: 44351035-44351035 on Build GRCh38
rs373210564 Chr.17: 44351029-44351029 on Build GRCh38
rs749940570 Chr.17: 44351087-44351087 on Build GRCh38
rs761798446 Chr.17: 44351065-44351065 on Build GRCh38
rs754715236 Chr.17: 44351187-44351187 on Build GRCh38
rs747362041 Chr.17: 44351124-44351124 on Build GRCh38
rs771557106 Chr.17: 44351025-44351025 on Build GRCh38
rs377103616 Chr.17: 44351042-44351042 on Build GRCh38
rs764208053 Chr.17: 44351166-44351166 on Build GRCh38
rs781207112 Chr.17: 44351133-44351133 on Build GRCh38
rs749270742 Chr.17: 44351327-44351327 on Build GRCh38
rs753286440 Chr.17: 44351177-44351177 on Build GRCh38
rs892435487 Chr.17: 44351310-44351310 on Build GRCh38
rs997573237 Chr.17: 44351032-44351032 on Build GRCh38
rs763996390 Chr.17: 44351053-44351053 on Build GRCh38
rs1030973406 Chr.17: 44350977-44350977 on Build GRCh38
rs760586487 Chr.17: 44351174-44351174 on Build GRCh38
rs757755025 Chr.17: 44351020-44351020 on Build GRCh38
rs765408713 Chr.17: 44351334-44351338 on Build GRCh38

View Full Product Details