Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45132508-45133012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACBD4
Entrez Gene ID
79777
Gene Name
acyl-CoA binding domain containing 4
Gene Aliases
HMFT0700
UniGene
Hs.110298
Gene Chromosome Location
Chr.17: 45132600-45144181 on Build GRCh38
Transcripts
NM_001321352.1 NM_001321353.1 XM_006722085.2 XM_011525259.2 XM_011525261.2 XM_017025084.1 XM_017025085.1 XM_017025086.1 XM_017025087.1 XM_017025088.1 XM_017025089.1 XM_017025090.1 XM_017025091.1 XM_017025092.1 XM_017025093.1 XM_017025095.1 XM_017025096.1 XM_017025097.1 XM_017025098.1 XM_017025099.1 XM_017025100.1 XM_017025101.1 NM_133373.4 XM_011524253.2

Gene Symbol
PLCD3
Entrez Gene ID
113026
Gene Name
phospholipase C delta 3
Gene Aliases
PLC-delta-3
UniGene
Hs.380094
Gene Chromosome Location
Chr.17: 45111640-45132533 on Build GRCh38
Transcripts
NM_001321352.1 NM_001321353.1 XM_006722085.2 XM_011525259.2 XM_011525261.2 XM_017025084.1 XM_017025085.1 XM_017025086.1 XM_017025087.1 XM_017025088.1 XM_017025089.1 XM_017025090.1 XM_017025091.1 XM_017025092.1 XM_017025093.1 XM_017025095.1 XM_017025096.1 XM_017025097.1 XM_017025098.1 XM_017025099.1 XM_017025100.1 XM_017025101.1 NM_133373.4 XM_011524253.2

SNP Detail

SNP ID Chromosome Location
rs1039517433 Chr.17: 45132547-45132547 on Build GRCh38
rs951439079 Chr.17: 45132589-45132589 on Build GRCh38
rs989603427 Chr.17: 45132747-45132747 on Build GRCh38
rs563287639 Chr.17: 45132950-45132950 on Build GRCh38
rs1024854639 Chr.17: 45132877-45132877 on Build GRCh38
rs1051940058 Chr.17: 45132647-45132647 on Build GRCh38
rs916963393 Chr.17: 45132631-45132631 on Build GRCh38
rs922028452 Chr.17: 45132754-45132754 on Build GRCh38
rs866456198 Chr.17: 45132595-45132595 on Build GRCh38
rs568297310 Chr.17: 45132773-45132773 on Build GRCh38
rs555734442 Chr.17: 45132977-45132977 on Build GRCh38
rs1048116904 Chr.17: 45132853-45132853 on Build GRCh38
rs1051611622 Chr.17: 45132674-45132674 on Build GRCh38
rs930994310 Chr.17: 45132828-45132828 on Build GRCh38
rs960860125 Chr.17: 45132734-45132734 on Build GRCh38
rs1012101404 Chr.17: 45132687-45132687 on Build GRCh38
rs998239158 Chr.17: 45132585-45132585 on Build GRCh38
rs540572788 Chr.17: 45132658-45132658 on Build GRCh38
rs970636532 Chr.17: 45132713-45132713 on Build GRCh38
rs535001643 Chr.17: 45132716-45132716 on Build GRCh38
rs1054355765 Chr.17: 45132628-45132628 on Build GRCh38
rs987605891 Chr.17: 45132771-45132771 on Build GRCh38
rs958041449 Chr.17: 45132919-45132919 on Build GRCh38
rs1036701276 Chr.17: 45132660-45132660 on Build GRCh38
rs906441458 Chr.17: 45132889-45132889 on Build GRCh38
rs1014171978 Chr.17: 45132933-45132933 on Build GRCh38
rs546149193 Chr.17: 45132626-45132626 on Build GRCh38
rs896845291 Chr.17: 45132821-45132821 on Build GRCh38
rs1042608040 Chr.17: 45132612-45132612 on Build GRCh38
rs890720151 Chr.17: 45132650-45132650 on Build GRCh38
rs898486324 Chr.17: 45132867-45132867 on Build GRCh38
rs370617588 Chr.17: 45132639-45132639 on Build GRCh38
rs569286303 Chr.17: 45132861-45132861 on Build GRCh38
rs1033835045 Chr.17: 45132725-45132725 on Build GRCh38
rs1009992532 Chr.17: 45132590-45132590 on Build GRCh38
rs1016913380 Chr.17: 45132698-45132698 on Build GRCh38
rs958350534 Chr.17: 45132733-45132733 on Build GRCh38
rs562570165 Chr.17: 45132714-45132714 on Build GRCh38
rs72832865 Chr.17: 45132970-45132970 on Build GRCh38
rs751214539 Chr.17: 45132856-45132856 on Build GRCh38
rs558122974 Chr.17: 45132934-45132934 on Build GRCh38
rs958478275 Chr.17: 45132627-45132627 on Build GRCh38
rs149949892 Chr.17: 45132805-45132805 on Build GRCh38
rs896892584 Chr.17: 45132661-45132661 on Build GRCh38
rs1038805439 Chr.17: 45132858-45132858 on Build GRCh38
rs923982713 Chr.17: 45132623-45132623 on Build GRCh38
rs917860033 Chr.17: 45132536-45132536 on Build GRCh38
rs187549251 Chr.17: 45132788-45132788 on Build GRCh38
rs899936580 Chr.17: 45132559-45132560 on Build GRCh38
rs866799318 Chr.17: 45132596-45132596 on Build GRCh38
rs1029689644 Chr.17: 45132586-45132586 on Build GRCh38
rs544309180 Chr.17: 45132691-45132691 on Build GRCh38
rs1027218145 Chr.17: 45132676-45132676 on Build GRCh38
rs970690336 Chr.17: 45132607-45132607 on Build GRCh38
rs1004625354 Chr.17: 45132694-45132694 on Build GRCh38
rs373079502 Chr.17: 45132679-45132679 on Build GRCh38
rs957023310 Chr.17: 45132986-45132986 on Build GRCh38
rs887210855 Chr.17: 45132840-45132840 on Build GRCh38
rs1000037052 Chr.17: 45132545-45132545 on Build GRCh38
rs999478290 Chr.17: 45132909-45132909 on Build GRCh38
rs146231156 Chr.17: 45132646-45132646 on Build GRCh38
rs925610123 Chr.17: 45132767-45132767 on Build GRCh38
rs953465565 Chr.17: 45132949-45132949 on Build GRCh38
rs916992108 Chr.17: 45132744-45132744 on Build GRCh38
rs1053821036 Chr.17: 45132583-45132583 on Build GRCh38
rs528754042 Chr.17: 45132757-45132757 on Build GRCh38
rs770835106 Chr.17: 45132937-45132937 on Build GRCh38
rs575663760 Chr.17: 45132622-45132622 on Build GRCh38
rs1031402295 Chr.17: 45132912-45132912 on Build GRCh38
rs893802396 Chr.17: 45132584-45132584 on Build GRCh38
rs932724107 Chr.17: 45132578-45132578 on Build GRCh38
rs183380710 Chr.17: 45132668-45132668 on Build GRCh38
rs139426383 Chr.17: 45132760-45132760 on Build GRCh38
rs922152936 Chr.17: 45132630-45132630 on Build GRCh38
rs573322395 Chr.17: 45132936-45132936 on Build GRCh38
rs936809763 Chr.17: 45132625-45132625 on Build GRCh38
rs551311435 Chr.17: 45132718-45132718 on Build GRCh38
rs539558782 Chr.17: 45132862-45132862 on Build GRCh38
rs940984299 Chr.17: 45132792-45132792 on Build GRCh38
rs921137800 Chr.17: 45132791-45132791 on Build GRCh38
rs894906773 Chr.17: 45132859-45132859 on Build GRCh38
rs935878156 Chr.17: 45132651-45132651 on Build GRCh38
rs559905117 Chr.17: 45132741-45132741 on Build GRCh38

View Full Product Details