Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 46923038-46923541 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOSR2
Entrez Gene ID
9570
Gene Name
golgi SNAP receptor complex member 2
Gene Aliases
Bos1, EPM6, GS27
UniGene
Hs.463278
Gene Chromosome Location
Chr.17: 46923086-46975519 on Build GRCh38
Transcripts
NM_001012511.2 NM_001321133.1 NM_001321134.1 NM_004287.4 NM_054022.3 XM_005257844.3 XM_006722190.3 XM_011525501.2 XM_011525502.2 XM_017025378.1 XM_017025379.1 XM_017025380.1 XM_017025381.1 XM_017025382.1 XM_017025383.1 XM_017025384.1 XM_017025385.1 XM_017025386.1 XM_017025387.1 XM_017025388.1 XM_017025389.1 XM_017025390.1 XM_017025391.1 XM_017025392.1 XM_017025393.1 XM_017025394.1 XM_017025395.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5983089 c.7C>A p.P3T Chr.17: 46923199-46923199 on Build GRCh38
5983090 c.7C>A p.P3T Chr.17: 46923199-46923199 on Build GRCh38
5983088 c.7C>A p.P3T Chr.17: 46923199-46923199 on Build GRCh38
5641294 c.17A>T p.Q6L Chr.17: 46923209-46923209 on Build GRCh38
3983509 c.15C>G p.F5L Chr.17: 46923207-46923207 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768312081 Chr.17: 46923228-46923228 on Build GRCh38
rs971902224 Chr.17: 46923077-46923077 on Build GRCh38
rs779892743 Chr.17: 46923155-46923156 on Build GRCh38
rs895199968 Chr.17: 46923387-46923387 on Build GRCh38
rs934383775 Chr.17: 46923160-46923160 on Build GRCh38
rs949715024 Chr.17: 46923105-46923105 on Build GRCh38
rs975759441 Chr.17: 46923063-46923063 on Build GRCh38
rs886053077 Chr.17: 46923173-46923173 on Build GRCh38
rs374396285 Chr.17: 46923236-46923236 on Build GRCh38
rs944286811 Chr.17: 46923331-46923331 on Build GRCh38
rs751712181 Chr.17: 46923194-46923194 on Build GRCh38
rs1050325997 Chr.17: 46923313-46923313 on Build GRCh38
rs386797400 Chr.17: 46923163-46923181 on Build GRCh38
rs530684599 Chr.17: 46923496-46923496 on Build GRCh38
rs1022901930 Chr.17: 46923275-46923275 on Build GRCh38
rs959903813 Chr.17: 46923238-46923238 on Build GRCh38
rs578223403 Chr.17: 46923347-46923347 on Build GRCh38
rs960134672 Chr.17: 46923500-46923500 on Build GRCh38
rs1035618725 Chr.17: 46923492-46923492 on Build GRCh38
rs926715302 Chr.17: 46923125-46923125 on Build GRCh38
rs1056571567 Chr.17: 46923152-46923152 on Build GRCh38
rs984640215 Chr.17: 46923083-46923083 on Build GRCh38
rs756276963 Chr.17: 46923202-46923202 on Build GRCh38
rs924377872 Chr.17: 46923154-46923154 on Build GRCh38
rs746855352 Chr.17: 46923211-46923212 on Build GRCh38
rs545533018 Chr.17: 46923361-46923361 on Build GRCh38
rs774363772 Chr.17: 46923190-46923190 on Build GRCh38
rs530179478 Chr.17: 46923079-46923079 on Build GRCh38
rs12944167 Chr.17: 46923199-46923199 on Build GRCh38
rs939404257 Chr.17: 46923134-46923134 on Build GRCh38
rs548152029 Chr.17: 46923443-46923443 on Build GRCh38
rs897281647 Chr.17: 46923206-46923206 on Build GRCh38
rs952096287 Chr.17: 46923088-46923088 on Build GRCh38
rs891407619 Chr.17: 46923319-46923319 on Build GRCh38
rs995435315 Chr.17: 46923366-46923366 on Build GRCh38
rs561373587 Chr.17: 46923461-46923461 on Build GRCh38
rs1015854832 Chr.17: 46923423-46923423 on Build GRCh38
rs989917587 Chr.17: 46923306-46923306 on Build GRCh38
rs183199 Chr.17: 46923181-46923181 on Build GRCh38
rs1023204527 Chr.17: 46923418-46923418 on Build GRCh38
rs747791818 Chr.17: 46923234-46923234 on Build GRCh38
rs794726915 Chr.17: 46923224-46923224 on Build GRCh38
rs997731224 Chr.17: 46923359-46923359 on Build GRCh38
rs770509830 Chr.17: 46923254-46923254 on Build GRCh38
rs934436033 Chr.17: 46923300-46923300 on Build GRCh38
rs547920808 Chr.17: 46923090-46923090 on Build GRCh38
rs765601291 Chr.17: 46923187-46923187 on Build GRCh38
rs562701294 Chr.17: 46923312-46923312 on Build GRCh38
rs886053076 Chr.17: 46923129-46923129 on Build GRCh38
rs1026370281 Chr.17: 46923068-46923068 on Build GRCh38
rs879328855 Chr.17: 46923193-46923193 on Build GRCh38
rs374064799 Chr.17: 46923100-46923100 on Build GRCh38
rs764879964 Chr.17: 46923417-46923417 on Build GRCh38
rs559045365 Chr.17: 46923082-46923082 on Build GRCh38
rs914271265 Chr.17: 46923308-46923308 on Build GRCh38
rs997650392 Chr.17: 46923179-46923179 on Build GRCh38
rs760018200 Chr.17: 46923186-46923186 on Build GRCh38
rs773823844 Chr.17: 46923174-46923174 on Build GRCh38
rs369892885 Chr.17: 46923167-46923167 on Build GRCh38
rs185341564 Chr.17: 46923280-46923280 on Build GRCh38
rs769918763 Chr.17: 46923164-46923164 on Build GRCh38
rs971288143 Chr.17: 46923277-46923277 on Build GRCh38
rs897166068 Chr.17: 46923357-46923357 on Build GRCh38
rs980056593 Chr.17: 46923219-46923219 on Build GRCh38
rs1045403860 Chr.17: 46923150-46923150 on Build GRCh38
rs995547601 Chr.17: 46923215-46923215 on Build GRCh38
rs951923133 Chr.17: 46923230-46923230 on Build GRCh38
rs771767913 Chr.17: 46923245-46923245 on Build GRCh38
rs757594881 Chr.17: 46923198-46923198 on Build GRCh38
rs1001819742 Chr.17: 46923485-46923485 on Build GRCh38
rs779815521 Chr.17: 46923203-46923203 on Build GRCh38
rs189899 Chr.17: 46923163-46923163 on Build GRCh38
rs920432230 Chr.17: 46923085-46923085 on Build GRCh38
rs768526397 Chr.17: 46923171-46923171 on Build GRCh38
rs763191781 Chr.17: 46923191-46923191 on Build GRCh38
rs981352878 Chr.17: 46923464-46923464 on Build GRCh38
rs189458855 Chr.17: 46923377-46923377 on Build GRCh38
rs562237021 Chr.17: 46923170-46923170 on Build GRCh38
rs909169794 Chr.17: 46923089-46923089 on Build GRCh38
rs111850322 Chr.17: 46923335-46923335 on Build GRCh38
rs749270151 Chr.17: 46923209-46923209 on Build GRCh38
rs767675224 Chr.17: 46923298-46923298 on Build GRCh38
rs12949902 Chr.17: 46923479-46923479 on Build GRCh38
rs992617000 Chr.17: 46923268-46923269 on Build GRCh38
rs574045746 Chr.17: 46923192-46923192 on Build GRCh38
rs373192791 Chr.17: 46923177-46923177 on Build GRCh38
rs573156486 Chr.17: 46923084-46923084 on Build GRCh38
rs981131543 Chr.17: 46923278-46923278 on Build GRCh38
rs950391474 Chr.17: 46923080-46923080 on Build GRCh38
rs886039603 Chr.17: 46923222-46923222 on Build GRCh38
rs573306680 Chr.17: 46923229-46923229 on Build GRCh38
rs12150668 Chr.17: 46923370-46923370 on Build GRCh38
rs148390346 Chr.17: 46923250-46923250 on Build GRCh38
rs1031551677 Chr.17: 46923502-46923502 on Build GRCh38
rs750589437 Chr.17: 46923201-46923201 on Build GRCh38

View Full Product Details