Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48841590-48842100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALCOCO2
Entrez Gene ID
10241
Gene Name
calcium binding and coiled-coil domain 2
Gene Aliases
NDP52
UniGene
Hs.514920
Gene Chromosome Location
Chr.17: 48830988-48865245 on Build GRCh38
Transcripts
NM_001261390.1 NM_001261391.1 NM_001261393.1 NM_001261395.1 NM_005831.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5972257 c.111C>G p.D37E Chr.17: 48841818-48841818 on Build GRCh38
7174093 c.165G>T p.W55C Chr.17: 48841872-48841872 on Build GRCh38
4151656 c.124T>C p.Y42H Chr.17: 48841831-48841831 on Build GRCh38
6464954 c.154C>A p.R52R Chr.17: 48841861-48841861 on Build GRCh38
6464955 c.155G>T p.R52L Chr.17: 48841862-48841862 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765125675 Chr.17: 48841862-48841862 on Build GRCh38
rs138961879 Chr.17: 48841857-48841857 on Build GRCh38
rs749812006 Chr.17: 48841792-48841792 on Build GRCh38
rs1002795275 Chr.17: 48841987-48841987 on Build GRCh38
rs775932832 Chr.17: 48841787-48841787 on Build GRCh38
rs1012532680 Chr.17: 48841654-48841654 on Build GRCh38
rs918305387 Chr.17: 48841955-48841955 on Build GRCh38
rs766294612 Chr.17: 48841677-48841677 on Build GRCh38
rs971247378 Chr.17: 48841675-48841675 on Build GRCh38
rs761609853 Chr.17: 48841942-48841942 on Build GRCh38
rs929661106 Chr.17: 48841979-48841979 on Build GRCh38
rs1033816344 Chr.17: 48842003-48842003 on Build GRCh38
rs200318833 Chr.17: 48841839-48841839 on Build GRCh38
rs780772905 Chr.17: 48841913-48841913 on Build GRCh38
rs754941587 Chr.17: 48841861-48841861 on Build GRCh38
rs1038842882 Chr.17: 48841800-48841800 on Build GRCh38
rs762797808 Chr.17: 48841666-48841667 on Build GRCh38
rs552522249 Chr.17: 48841824-48841824 on Build GRCh38
rs753819624 Chr.17: 48841689-48841689 on Build GRCh38
rs767819806 Chr.17: 48841664-48841664 on Build GRCh38
rs769234486 Chr.17: 48841802-48841802 on Build GRCh38
rs1031018864 Chr.17: 48841706-48841706 on Build GRCh38
rs758174944 Chr.17: 48841885-48841885 on Build GRCh38
rs751191089 Chr.17: 48841884-48841885 on Build GRCh38
rs770466837 Chr.17: 48841785-48841785 on Build GRCh38
rs769338590 Chr.17: 48841916-48841916 on Build GRCh38
rs768386881 Chr.17: 48841648-48841648 on Build GRCh38
rs905596640 Chr.17: 48841965-48841965 on Build GRCh38
rs746558307 Chr.17: 48841739-48841739 on Build GRCh38
rs373600469 Chr.17: 48841715-48841715 on Build GRCh38
rs746673859 Chr.17: 48841909-48841909 on Build GRCh38
rs756798651 Chr.17: 48841911-48841911 on Build GRCh38
rs375681296 Chr.17: 48841818-48841818 on Build GRCh38
rs752535564 Chr.17: 48841863-48841863 on Build GRCh38
rs554115615 Chr.17: 48842024-48842024 on Build GRCh38
rs778703939 Chr.17: 48841719-48841719 on Build GRCh38
rs944841629 Chr.17: 48841881-48841881 on Build GRCh38
rs750520631 Chr.17: 48841666-48841666 on Build GRCh38
rs35668544 Chr.17: 48841836-48841837 on Build GRCh38
rs997409675 Chr.17: 48841845-48841845 on Build GRCh38
rs747896111 Chr.17: 48841729-48841729 on Build GRCh38
rs528158396 Chr.17: 48842009-48842009 on Build GRCh38
rs7211475 Chr.17: 48841616-48841616 on Build GRCh38
rs202108136 Chr.17: 48841836-48841836 on Build GRCh38
rs773392344 Chr.17: 48841928-48841928 on Build GRCh38
rs766155654 Chr.17: 48841734-48841735 on Build GRCh38
rs758085076 Chr.17: 48841730-48841730 on Build GRCh38
rs753813766 Chr.17: 48841859-48841859 on Build GRCh38
rs992574366 Chr.17: 48841643-48841643 on Build GRCh38
rs534044078 Chr.17: 48841954-48841954 on Build GRCh38
rs762249535 Chr.17: 48841819-48841819 on Build GRCh38
rs772573437 Chr.17: 48841925-48841925 on Build GRCh38
rs369990612 Chr.17: 48841669-48841669 on Build GRCh38
rs147070596 Chr.17: 48841695-48841695 on Build GRCh38
rs966689232 Chr.17: 48842046-48842046 on Build GRCh38
rs777354852 Chr.17: 48841895-48841895 on Build GRCh38
rs371490739 Chr.17: 48841922-48841922 on Build GRCh38
rs111868404 Chr.17: 48842038-48842038 on Build GRCh38
rs745327976 Chr.17: 48841915-48841915 on Build GRCh38
rs866560115 Chr.17: 48841746-48841746 on Build GRCh38
rs774988415 Chr.17: 48841921-48841921 on Build GRCh38
rs1021001976 Chr.17: 48842042-48842042 on Build GRCh38
rs748594391 Chr.17: 48841923-48841923 on Build GRCh38
rs777380934 Chr.17: 48841734-48841734 on Build GRCh38
rs766494137 Chr.17: 48841858-48841858 on Build GRCh38

View Full Product Details