Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48845971-48846505 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALCOCO2
Entrez Gene ID
10241
Gene Name
calcium binding and coiled-coil domain 2
Gene Aliases
NDP52
UniGene
Hs.514920
Gene Chromosome Location
Chr.17: 48830988-48865245 on Build GRCh38
Transcripts
NM_001261390.1 NM_001261391.1 NM_001261393.1 NM_001261395.1 NM_005831.4

SNP Detail

SNP ID Chromosome Location
rs985320418 Chr.17: 48846046-48846046 on Build GRCh38
rs575635330 Chr.17: 48846063-48846063 on Build GRCh38
rs1025528482 Chr.17: 48846441-48846441 on Build GRCh38
rs1022012458 Chr.17: 48846361-48846361 on Build GRCh38
rs1048505245 Chr.17: 48846206-48846206 on Build GRCh38
rs765215574 Chr.17: 48846418-48846418 on Build GRCh38
rs1041529609 Chr.17: 48846065-48846065 on Build GRCh38
rs189795766 Chr.17: 48846247-48846247 on Build GRCh38
rs551402638 Chr.17: 48846384-48846384 on Build GRCh38
rs936071884 Chr.17: 48846102-48846102 on Build GRCh38
rs573822794 Chr.17: 48845997-48845997 on Build GRCh38
rs899832863 Chr.17: 48846108-48846108 on Build GRCh38
rs1004405819 Chr.17: 48846224-48846224 on Build GRCh38
rs761984633 Chr.17: 48846316-48846316 on Build GRCh38
rs1019433488 Chr.17: 48846391-48846391 on Build GRCh38
rs774349370 Chr.17: 48846268-48846268 on Build GRCh38
rs773464618 Chr.17: 48846371-48846371 on Build GRCh38
rs759588702 Chr.17: 48846071-48846071 on Build GRCh38
rs369153718 Chr.17: 48846325-48846325 on Build GRCh38
rs924743966 Chr.17: 48846080-48846080 on Build GRCh38
rs931011794 Chr.17: 48846179-48846179 on Build GRCh38
rs1021732663 Chr.17: 48846036-48846036 on Build GRCh38
rs62621985 Chr.17: 48846470-48846470 on Build GRCh38
rs977254107 Chr.17: 48846364-48846364 on Build GRCh38
rs918273549 Chr.17: 48846160-48846160 on Build GRCh38
rs956053409 Chr.17: 48846020-48846020 on Build GRCh38
rs917205879 Chr.17: 48846349-48846350 on Build GRCh38
rs192432955 Chr.17: 48846266-48846266 on Build GRCh38
rs1053346960 Chr.17: 48846319-48846319 on Build GRCh38
rs916339926 Chr.17: 48846208-48846208 on Build GRCh38
rs1011593462 Chr.17: 48846356-48846356 on Build GRCh38
rs533842 Chr.17: 48846022-48846022 on Build GRCh38
rs965297773 Chr.17: 48846120-48846120 on Build GRCh38
rs752625620 Chr.17: 48846429-48846429 on Build GRCh38
rs972514586 Chr.17: 48846424-48846424 on Build GRCh38
rs947754599 Chr.17: 48846246-48846246 on Build GRCh38
rs958624586 Chr.17: 48846283-48846283 on Build GRCh38
rs767590032 Chr.17: 48846342-48846342 on Build GRCh38
rs972609653 Chr.17: 48846131-48846131 on Build GRCh38
rs201271700 Chr.17: 48846204-48846205 on Build GRCh38
rs1051380585 Chr.17: 48846177-48846177 on Build GRCh38
rs954242580 Chr.17: 48846043-48846043 on Build GRCh38
rs949924126 Chr.17: 48846060-48846060 on Build GRCh38

View Full Product Details