Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 49796839-49797388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KAT7
Entrez Gene ID
11143
Gene Name
lysine acetyltransferase 7
Gene Aliases
HBO1, HBOA, MYST2, ZC2HC7
UniGene
Hs.21907
Gene Chromosome Location
Chr.17: 49788619-49829096 on Build GRCh38
Transcripts
NM_001199155.1 NM_001199156.1 NM_001199157.1 NM_001199158.1 NM_007067.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5747873 c.262A>G p.T88A Chr.17: 49796848-49796848 on Build GRCh38
3421676 c.266C>T p.P89L Chr.17: 49796852-49796852 on Build GRCh38
6277399 c.339A>T p.R113S Chr.17: 49796925-49796925 on Build GRCh38
4933708 c.322G>T p.V108F Chr.17: 49796908-49796908 on Build GRCh38
6690238 c.283C>T p.R95W Chr.17: 49796869-49796869 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs944064740 Chr.17: 49797306-49797306 on Build GRCh38
rs55883907 Chr.17: 49797237-49797237 on Build GRCh38
rs891046106 Chr.17: 49797100-49797100 on Build GRCh38
rs545981026 Chr.17: 49796898-49796898 on Build GRCh38
rs1035632489 Chr.17: 49797209-49797209 on Build GRCh38
rs1047442797 Chr.17: 49797157-49797157 on Build GRCh38
rs956364185 Chr.17: 49797140-49797140 on Build GRCh38
rs750980612 Chr.17: 49797340-49797341 on Build GRCh38
rs573040426 Chr.17: 49796963-49796963 on Build GRCh38
rs1041267031 Chr.17: 49797309-49797309 on Build GRCh38
rs985394800 Chr.17: 49797284-49797284 on Build GRCh38
rs928445354 Chr.17: 49796940-49796940 on Build GRCh38
rs562052321 Chr.17: 49797007-49797007 on Build GRCh38
rs548565167 Chr.17: 49797204-49797204 on Build GRCh38
rs750842619 Chr.17: 49796924-49796924 on Build GRCh38
rs530011485 Chr.17: 49797034-49797034 on Build GRCh38
rs373781117 Chr.17: 49797035-49797036 on Build GRCh38
rs574522622 Chr.17: 49797156-49797156 on Build GRCh38
rs1050210346 Chr.17: 49797009-49797009 on Build GRCh38
rs555218442 Chr.17: 49796905-49796905 on Build GRCh38
rs1022644460 Chr.17: 49797165-49797165 on Build GRCh38
rs758919960 Chr.17: 49796937-49796937 on Build GRCh38
rs532458031 Chr.17: 49797060-49797060 on Build GRCh38
rs528566887 Chr.17: 49797012-49797012 on Build GRCh38
rs981928407 Chr.17: 49797126-49797126 on Build GRCh38
rs780469886 Chr.17: 49796939-49796939 on Build GRCh38
rs568756653 Chr.17: 49797220-49797220 on Build GRCh38
rs970062876 Chr.17: 49797083-49797083 on Build GRCh38
rs887500483 Chr.17: 49797359-49797359 on Build GRCh38
rs777510718 Chr.17: 49796953-49796953 on Build GRCh38
rs887542212 Chr.17: 49797159-49797159 on Build GRCh38
rs1041152250 Chr.17: 49797280-49797280 on Build GRCh38
rs888301137 Chr.17: 49797362-49797362 on Build GRCh38
rs952643177 Chr.17: 49797063-49797063 on Build GRCh38
rs1018140123 Chr.17: 49797114-49797114 on Build GRCh38
rs149083262 Chr.17: 49796943-49796943 on Build GRCh38
rs1027280568 Chr.17: 49797014-49797014 on Build GRCh38
rs1041319027 Chr.17: 49797125-49797125 on Build GRCh38
rs765611916 Chr.17: 49797026-49797026 on Build GRCh38
rs553165492 Chr.17: 49797046-49797046 on Build GRCh38
rs565913463 Chr.17: 49797132-49797132 on Build GRCh38
rs1049042307 Chr.17: 49797343-49797343 on Build GRCh38
rs947003468 Chr.17: 49796901-49796901 on Build GRCh38
rs182125771 Chr.17: 49797253-49797253 on Build GRCh38
rs181443956 Chr.17: 49797086-49797086 on Build GRCh38
rs533377454 Chr.17: 49797009-49797010 on Build GRCh38
rs930133716 Chr.17: 49797341-49797341 on Build GRCh38
rs1022904323 Chr.17: 49797066-49797066 on Build GRCh38
rs747691369 Chr.17: 49796947-49796947 on Build GRCh38
rs989002202 Chr.17: 49797158-49797158 on Build GRCh38
rs1006774321 Chr.17: 49797103-49797103 on Build GRCh38
rs889999593 Chr.17: 49797028-49797028 on Build GRCh38
rs774264789 Chr.17: 49796973-49796973 on Build GRCh38
rs955886869 Chr.17: 49797216-49797216 on Build GRCh38
rs186464490 Chr.17: 49797164-49797164 on Build GRCh38
rs1006153346 Chr.17: 49797064-49797064 on Build GRCh38
rs550229243 Chr.17: 49797067-49797068 on Build GRCh38
rs745690093 Chr.17: 49796974-49796974 on Build GRCh38
rs563399821 Chr.17: 49797055-49797055 on Build GRCh38
rs541131895 Chr.17: 49797037-49797037 on Build GRCh38
rs918851070 Chr.17: 49797314-49797314 on Build GRCh38
rs74332240 Chr.17: 49797080-49797080 on Build GRCh38
rs947383069 Chr.17: 49797006-49797006 on Build GRCh38
rs988893693 Chr.17: 49796983-49796983 on Build GRCh38
rs191629861 Chr.17: 49796997-49796997 on Build GRCh38
rs755641140 Chr.17: 49796952-49796952 on Build GRCh38
rs911215948 Chr.17: 49797298-49797298 on Build GRCh38
rs374837770 Chr.17: 49797067-49797067 on Build GRCh38
rs994771009 Chr.17: 49797142-49797142 on Build GRCh38
rs969744400 Chr.17: 49797188-49797188 on Build GRCh38
rs557006013 Chr.17: 49797249-49797249 on Build GRCh38
rs201468778 Chr.17: 49796915-49796915 on Build GRCh38
rs539536839 Chr.17: 49797340-49797340 on Build GRCh38
rs757537872 Chr.17: 49797299-49797299 on Build GRCh38
rs985737257 Chr.17: 49797226-49797226 on Build GRCh38
rs969965491 Chr.17: 49796879-49796879 on Build GRCh38
rs754069524 Chr.17: 49796876-49796876 on Build GRCh38
rs763360657 Chr.17: 49797066-49797067 on Build GRCh38

View Full Product Details