Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 49846845-49847341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ45513
Entrez Gene ID
729220
Gene Name
uncharacterized LOC729220
Gene Aliases
-
UniGene
Hs.434138
Gene Chromosome Location
Chr.17: 49845910-49848837 on Build GRCh38
Transcripts
NM_001242791.1 NM_001077503.1 NM_001077504.1 NM_001077505.1 NM_001077506.1 NM_170685.2

Gene Symbol
TAC4
Entrez Gene ID
255061
Gene Name
tachykinin 4 (hemokinin)
Gene Aliases
EK, HK-1, HK1, PPT-C
UniGene
Hs.652130
Gene Chromosome Location
Chr.17: 49838309-49848017 on Build GRCh38
Transcripts
NM_001242791.1 NM_001077503.1 NM_001077504.1 NM_001077505.1 NM_001077506.1 NM_170685.2

SNP Detail

SNP ID Chromosome Location
rs938287084 Chr.17: 49847114-49847114 on Build GRCh38
rs757598740 Chr.17: 49847172-49847172 on Build GRCh38
rs1004251523 Chr.17: 49847227-49847227 on Build GRCh38
rs185701868 Chr.17: 49846927-49846927 on Build GRCh38
rs956132222 Chr.17: 49846908-49846908 on Build GRCh38
rs752811007 Chr.17: 49847112-49847112 on Build GRCh38
rs566282886 Chr.17: 49847118-49847118 on Build GRCh38
rs1029678015 Chr.17: 49847043-49847043 on Build GRCh38
rs188977489 Chr.17: 49847292-49847292 on Build GRCh38
rs752716780 Chr.17: 49846999-49846999 on Build GRCh38
rs532695191 Chr.17: 49847102-49847102 on Build GRCh38
rs1044306551 Chr.17: 49847110-49847110 on Build GRCh38
rs769324776 Chr.17: 49847224-49847224 on Build GRCh38
rs911483449 Chr.17: 49846977-49846977 on Build GRCh38
rs552766558 Chr.17: 49847115-49847115 on Build GRCh38
rs891159709 Chr.17: 49847223-49847223 on Build GRCh38
rs1042198127 Chr.17: 49847140-49847140 on Build GRCh38
rs765131915 Chr.17: 49847281-49847281 on Build GRCh38
rs201896080 Chr.17: 49847213-49847213 on Build GRCh38
rs780364865 Chr.17: 49847293-49847293 on Build GRCh38
rs955620320 Chr.17: 49847062-49847062 on Build GRCh38
rs1047972284 Chr.17: 49847096-49847096 on Build GRCh38
rs944185889 Chr.17: 49846984-49846984 on Build GRCh38
rs1056201681 Chr.17: 49846922-49846922 on Build GRCh38
rs772854574 Chr.17: 49847030-49847030 on Build GRCh38
rs746282282 Chr.17: 49847205-49847205 on Build GRCh38
rs546964368 Chr.17: 49847004-49847004 on Build GRCh38
rs371403490 Chr.17: 49847243-49847243 on Build GRCh38
rs1000778850 Chr.17: 49846990-49846990 on Build GRCh38
rs1021618851 Chr.17: 49846923-49846923 on Build GRCh38
rs535530178 Chr.17: 49847156-49847156 on Build GRCh38
rs528767390 Chr.17: 49847127-49847127 on Build GRCh38
rs772493155 Chr.17: 49846957-49846957 on Build GRCh38
rs762695205 Chr.17: 49847057-49847057 on Build GRCh38
rs146522271 Chr.17: 49847079-49847079 on Build GRCh38
rs887409121 Chr.17: 49847098-49847098 on Build GRCh38
rs941682614 Chr.17: 49847109-49847109 on Build GRCh38
rs541777177 Chr.17: 49846919-49846919 on Build GRCh38
rs530025843 Chr.17: 49846980-49846980 on Build GRCh38
rs747590190 Chr.17: 49847214-49847214 on Build GRCh38
rs746359170 Chr.17: 49846945-49846945 on Build GRCh38
rs767693667 Chr.17: 49847146-49847146 on Build GRCh38
rs779302729 Chr.17: 49847203-49847203 on Build GRCh38
rs985443813 Chr.17: 49847217-49847217 on Build GRCh38
rs779236027 Chr.17: 49846928-49846928 on Build GRCh38
rs749664702 Chr.17: 49847207-49847207 on Build GRCh38
rs756407796 Chr.17: 49847279-49847279 on Build GRCh38
rs770531887 Chr.17: 49847281-49847281 on Build GRCh38
rs1002816845 Chr.17: 49847120-49847120 on Build GRCh38
rs936513495 Chr.17: 49847154-49847154 on Build GRCh38
rs997284717 Chr.17: 49847032-49847032 on Build GRCh38
rs926289173 Chr.17: 49846906-49846906 on Build GRCh38
rs1002721783 Chr.17: 49846877-49846877 on Build GRCh38
rs761708258 Chr.17: 49847176-49847176 on Build GRCh38
rs903688351 Chr.17: 49847145-49847145 on Build GRCh38
rs545411474 Chr.17: 49847058-49847058 on Build GRCh38
rs766183946 Chr.17: 49847078-49847078 on Build GRCh38
rs776197744 Chr.17: 49846988-49846988 on Build GRCh38
rs1010318733 Chr.17: 49847196-49847196 on Build GRCh38
rs142464631 Chr.17: 49847015-49847015 on Build GRCh38
rs568771063 Chr.17: 49847230-49847230 on Build GRCh38
rs751503039 Chr.17: 49847128-49847128 on Build GRCh38
rs779216577 Chr.17: 49847315-49847315 on Build GRCh38
rs977005481 Chr.17: 49847302-49847302 on Build GRCh38
rs56186849 Chr.17: 49846905-49846905 on Build GRCh38
rs536638786 Chr.17: 49847267-49847267 on Build GRCh38
rs1017575141 Chr.17: 49847019-49847019 on Build GRCh38
rs941580735 Chr.17: 49846903-49846903 on Build GRCh38
rs200699277 Chr.17: 49847087-49847087 on Build GRCh38
rs982837145 Chr.17: 49847071-49847071 on Build GRCh38

View Full Product Details