Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 49973092-49973588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLX4
Entrez Gene ID
1748
Gene Name
distal-less homeobox 4
Gene Aliases
BP1, DLX7, DLX8, DLX9, OFC15
UniGene
Hs.591167
Gene Chromosome Location
Chr.17: 49968970-49974959 on Build GRCh38
Transcripts
NM_001934.3 NM_138281.2 XM_017024291.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6645122 c.106C>T p.R36W Chr.17: 49973111-49973111 on Build GRCh38
3889859 c.102C>T p.S34S Chr.17: 49973107-49973107 on Build GRCh38
6869925 c.342C>A p.A114A Chr.17: 49973131-49973131 on Build GRCh38
6645124 c.174G>A p.Q58Q Chr.17: 49973179-49973179 on Build GRCh38
6645121 c.322C>T p.R108W Chr.17: 49973111-49973111 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1044222265 Chr.17: 49973484-49973484 on Build GRCh38
rs911372543 Chr.17: 49973346-49973346 on Build GRCh38
rs749670037 Chr.17: 49973114-49973114 on Build GRCh38
rs370138317 Chr.17: 49973131-49973132 on Build GRCh38
rs777442358 Chr.17: 49973302-49973302 on Build GRCh38
rs914843640 Chr.17: 49973269-49973269 on Build GRCh38
rs745721685 Chr.17: 49973329-49973329 on Build GRCh38
rs780544447 Chr.17: 49973282-49973282 on Build GRCh38
rs746238219 Chr.17: 49973348-49973351 on Build GRCh38
rs767672602 Chr.17: 49973158-49973158 on Build GRCh38
rs754160098 Chr.17: 49973175-49973175 on Build GRCh38
rs774177303 Chr.17: 49973146-49973146 on Build GRCh38
rs769494752 Chr.17: 49973139-49973139 on Build GRCh38
rs748859909 Chr.17: 49973307-49973307 on Build GRCh38
rs372168267 Chr.17: 49973395-49973395 on Build GRCh38
rs768591855 Chr.17: 49973342-49973342 on Build GRCh38
rs1053396168 Chr.17: 49973426-49973426 on Build GRCh38
rs1036224708 Chr.17: 49973521-49973521 on Build GRCh38
rs756852050 Chr.17: 49973397-49973397 on Build GRCh38
rs779305699 Chr.17: 49973120-49973120 on Build GRCh38
rs765499246 Chr.17: 49973265-49973265 on Build GRCh38
rs771446065 Chr.17: 49973287-49973288 on Build GRCh38
rs778257713 Chr.17: 49973113-49973113 on Build GRCh38
rs766822070 Chr.17: 49973370-49973370 on Build GRCh38
rs973289126 Chr.17: 49973531-49973531 on Build GRCh38
rs772064013 Chr.17: 49973332-49973332 on Build GRCh38
rs902773730 Chr.17: 49973132-49973132 on Build GRCh38
rs1014726649 Chr.17: 49973500-49973500 on Build GRCh38
rs150742579 Chr.17: 49973251-49973251 on Build GRCh38
rs768188135 Chr.17: 49973124-49973124 on Build GRCh38
rs778949467 Chr.17: 49973486-49973486 on Build GRCh38
rs747724378 Chr.17: 49973205-49973205 on Build GRCh38
rs775577618 Chr.17: 49973340-49973340 on Build GRCh38
rs764473165 Chr.17: 49973173-49973173 on Build GRCh38
rs560535958 Chr.17: 49973393-49973393 on Build GRCh38
rs374194687 Chr.17: 49973276-49973276 on Build GRCh38
rs755235771 Chr.17: 49973371-49973371 on Build GRCh38
rs774779056 Chr.17: 49973306-49973306 on Build GRCh38
rs759620239 Chr.17: 49973150-49973150 on Build GRCh38
rs752145159 Chr.17: 49973283-49973283 on Build GRCh38
rs760963288 Chr.17: 49973160-49973160 on Build GRCh38
rs775704162 Chr.17: 49973253-49973253 on Build GRCh38
rs757773828 Chr.17: 49973117-49973117 on Build GRCh38
rs540857506 Chr.17: 49973377-49973377 on Build GRCh38
rs575249063 Chr.17: 49973111-49973111 on Build GRCh38
rs956141580 Chr.17: 49973530-49973530 on Build GRCh38
rs777303729 Chr.17: 49973225-49973225 on Build GRCh38
rs753166489 Chr.17: 49973492-49973492 on Build GRCh38
rs769369952 Chr.17: 49973224-49973224 on Build GRCh38
rs151318731 Chr.17: 49973193-49973193 on Build GRCh38
rs755517636 Chr.17: 49973378-49973378 on Build GRCh38
rs887714053 Chr.17: 49973466-49973466 on Build GRCh38
rs779438741 Chr.17: 49973187-49973187 on Build GRCh38
rs776823926 Chr.17: 49973349-49973349 on Build GRCh38
rs543347418 Chr.17: 49973475-49973475 on Build GRCh38
rs758955111 Chr.17: 49973192-49973192 on Build GRCh38
rs1029197898 Chr.17: 49973477-49973477 on Build GRCh38
rs780722446 Chr.17: 49973129-49973129 on Build GRCh38
rs756981590 Chr.17: 49973319-49973319 on Build GRCh38
rs574249255 Chr.17: 49973409-49973409 on Build GRCh38
rs999152692 Chr.17: 49973183-49973183 on Build GRCh38
rs754041857 Chr.17: 49973259-49973259 on Build GRCh38
rs763321942 Chr.17: 49973364-49973364 on Build GRCh38
rs773499716 Chr.17: 49973361-49973361 on Build GRCh38
rs753916651 Chr.17: 49973352-49973352 on Build GRCh38
rs757589150 Chr.17: 49973182-49973182 on Build GRCh38
rs928995864 Chr.17: 49973445-49973445 on Build GRCh38
rs370377249 Chr.17: 49973248-49973248 on Build GRCh38
rs765563815 Chr.17: 49973356-49973356 on Build GRCh38
rs753442669 Chr.17: 49973396-49973396 on Build GRCh38
rs1010682951 Chr.17: 49973558-49973558 on Build GRCh38
rs746483420 Chr.17: 49973123-49973123 on Build GRCh38
rs1035702966 Chr.17: 49973252-49973252 on Build GRCh38
rs762661270 Chr.17: 49973143-49973143 on Build GRCh38
rs1027562048 Chr.17: 49973551-49973551 on Build GRCh38
rs139118373 Chr.17: 49973260-49973260 on Build GRCh38
rs770843158 Chr.17: 49973144-49973144 on Build GRCh38
rs778557043 Chr.17: 49973323-49973323 on Build GRCh38
rs893572771 Chr.17: 49973217-49973217 on Build GRCh38
rs747049496 Chr.17: 49973341-49973341 on Build GRCh38
rs760838498 Chr.17: 49973255-49973255 on Build GRCh38
rs372156240 Chr.17: 49973142-49973142 on Build GRCh38
rs749719388 Chr.17: 49973129-49973129 on Build GRCh38
rs747779313 Chr.17: 49973131-49973131 on Build GRCh38
rs1559829 Chr.17: 49973448-49973448 on Build GRCh38
rs954724802 Chr.17: 49973233-49973233 on Build GRCh38
rs764366790 Chr.17: 49973257-49973257 on Build GRCh38
rs750860246 Chr.17: 49973275-49973275 on Build GRCh38
rs1048145603 Chr.17: 49973157-49973157 on Build GRCh38
rs1047376323 Chr.17: 49973453-49973453 on Build GRCh38
rs750866926 Chr.17: 49973191-49973191 on Build GRCh38
rs755644161 Chr.17: 49973284-49973284 on Build GRCh38
rs562426934 Chr.17: 49973524-49973524 on Build GRCh38
rs749061325 Chr.17: 49973244-49973244 on Build GRCh38

View Full Product Details