Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50056257-50056761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA3
Entrez Gene ID
3675
Gene Name
integrin subunit alpha 3
Gene Aliases
CD49C, FRP-2, GAP-B3, GAPB3, ILNEB, MSK18, VCA-2, VL3A, VLA3a
UniGene
Hs.265829
Gene Chromosome Location
Chr.17: 50055968-50090485 on Build GRCh38
Transcripts
NM_002204.3 XM_005257308.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4524409 c.126G>A p.V42V Chr.17: 50056565-50056565 on Build GRCh38
4943882 c.200G>C p.R67P Chr.17: 50056639-50056639 on Build GRCh38
1302990 c.187G>A p.E63K Chr.17: 50056626-50056626 on Build GRCh38
5972266 c.121G>T p.V41L Chr.17: 50056560-50056560 on Build GRCh38
6523673 c.87C>A p.V29V Chr.17: 50056526-50056526 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765222002 Chr.17: 50056604-50056604 on Build GRCh38
rs908827953 Chr.17: 50056527-50056527 on Build GRCh38
rs776549135 Chr.17: 50056680-50056680 on Build GRCh38
rs564360365 Chr.17: 50056384-50056384 on Build GRCh38
rs922827589 Chr.17: 50056491-50056491 on Build GRCh38
rs908322767 Chr.17: 50056653-50056653 on Build GRCh38
rs975654305 Chr.17: 50056483-50056483 on Build GRCh38
rs941215209 Chr.17: 50056377-50056377 on Build GRCh38
rs975509582 Chr.17: 50056463-50056463 on Build GRCh38
rs967488958 Chr.17: 50056467-50056467 on Build GRCh38
rs750587035 Chr.17: 50056616-50056616 on Build GRCh38
rs982779323 Chr.17: 50056526-50056526 on Build GRCh38
rs767963814 Chr.17: 50056632-50056632 on Build GRCh38
rs374174059 Chr.17: 50056508-50056508 on Build GRCh38
rs1041349892 Chr.17: 50056727-50056727 on Build GRCh38
rs964139493 Chr.17: 50056478-50056478 on Build GRCh38
rs868126432 Chr.17: 50056513-50056513 on Build GRCh38
rs757986965 Chr.17: 50056661-50056661 on Build GRCh38
rs376671926 Chr.17: 50056599-50056599 on Build GRCh38
rs938146569 Chr.17: 50056601-50056601 on Build GRCh38
rs963276455 Chr.17: 50056553-50056553 on Build GRCh38
rs779701860 Chr.17: 50056663-50056663 on Build GRCh38
rs751166737 Chr.17: 50056525-50056526 on Build GRCh38
rs776431279 Chr.17: 50056418-50056418 on Build GRCh38
rs776404147 Chr.17: 50056584-50056584 on Build GRCh38
rs748793535 Chr.17: 50056421-50056421 on Build GRCh38
rs754656724 Chr.17: 50056678-50056679 on Build GRCh38
rs1000803746 Chr.17: 50056468-50056468 on Build GRCh38
rs1038489414 Chr.17: 50056724-50056724 on Build GRCh38
rs540556912 Chr.17: 50056639-50056639 on Build GRCh38
rs759211080 Chr.17: 50056676-50056676 on Build GRCh38
rs903122561 Chr.17: 50056737-50056737 on Build GRCh38
rs560069882 Chr.17: 50056732-50056732 on Build GRCh38
rs955361533 Chr.17: 50056503-50056503 on Build GRCh38
rs895492866 Chr.17: 50056337-50056337 on Build GRCh38
rs1000060536 Chr.17: 50056398-50056398 on Build GRCh38
rs771778302 Chr.17: 50056518-50056518 on Build GRCh38
rs778533113 Chr.17: 50056492-50056492 on Build GRCh38
rs753121051 Chr.17: 50056633-50056633 on Build GRCh38
rs746764263 Chr.17: 50056672-50056672 on Build GRCh38
rs766723131 Chr.17: 50056628-50056628 on Build GRCh38
rs903088789 Chr.17: 50056389-50056389 on Build GRCh38
rs199862946 Chr.17: 50056473-50056473 on Build GRCh38
rs1020196479 Chr.17: 50056372-50056372 on Build GRCh38
rs1000541321 Chr.17: 50056738-50056738 on Build GRCh38
rs768316065 Chr.17: 50056677-50056677 on Build GRCh38
rs763201490 Chr.17: 50056618-50056618 on Build GRCh38
rs933633966 Chr.17: 50056321-50056321 on Build GRCh38
rs868149945 Chr.17: 50056305-50056305 on Build GRCh38
rs768529626 Chr.17: 50056581-50056581 on Build GRCh38
rs778281378 Chr.17: 50056647-50056647 on Build GRCh38
rs982443386 Chr.17: 50056338-50056338 on Build GRCh38
rs775195327 Chr.17: 50056575-50056575 on Build GRCh38
rs982451007 Chr.17: 50056523-50056523 on Build GRCh38
rs745493795 Chr.17: 50056499-50056499 on Build GRCh38
rs895834773 Chr.17: 50056437-50056439 on Build GRCh38
rs1020814034 Chr.17: 50056466-50056468 on Build GRCh38
rs751924860 Chr.17: 50056629-50056629 on Build GRCh38
rs760473716 Chr.17: 50056577-50056577 on Build GRCh38
rs1055440940 Chr.17: 50056334-50056334 on Build GRCh38
rs1002124626 Chr.17: 50056408-50056408 on Build GRCh38
rs916993007 Chr.17: 50056658-50056658 on Build GRCh38
rs1033243260 Chr.17: 50056407-50056407 on Build GRCh38
rs745385761 Chr.17: 50056656-50056656 on Build GRCh38
rs868296100 Chr.17: 50056416-50056416 on Build GRCh38
rs1013834700 Chr.17: 50056339-50056339 on Build GRCh38
rs73330528 Chr.17: 50056292-50056292 on Build GRCh38

View Full Product Details